Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6199838-6200332 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR52W1
Entrez Gene ID
120787
Gene Name
olfactory receptor family 52 subfamily W member 1
Gene Aliases
OR11-71, OR52W1P
UniGene
Hs.553570
Gene Chromosome Location
Chr.11: 6199224-6200186 on Build GRCh38
Transcripts
NM_001005178.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
322366 c.803G>A p.R268H Chr.11: 6200026-6200026 on Build GRCh38
6133792 c.931G>A p.E311K Chr.11: 6200154-6200154 on Build GRCh38
6282387 c.791A>G p.Y264C Chr.11: 6200014-6200014 on Build GRCh38
337286 c.701C>A p.P234H Chr.11: 6199924-6199924 on Build GRCh38
358496 c.662G>T p.G221V Chr.11: 6199885-6199885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768280451 Chr.11: 6200094-6200094 on Build GRCh38
rs201282683 Chr.11: 6200220-6200220 on Build GRCh38
rs376530772 Chr.11: 6199900-6199900 on Build GRCh38
rs780153200 Chr.11: 6200277-6200277 on Build GRCh38
rs773980390 Chr.11: 6199890-6199890 on Build GRCh38
rs748574406 Chr.11: 6199931-6199931 on Build GRCh38
rs752330733 Chr.11: 6199895-6199895 on Build GRCh38
rs371022304 Chr.11: 6199937-6199937 on Build GRCh38
rs113131236 Chr.11: 6200068-6200068 on Build GRCh38
rs938344976 Chr.11: 6200107-6200107 on Build GRCh38
rs779347603 Chr.11: 6200182-6200182 on Build GRCh38
rs754223253 Chr.11: 6200179-6200179 on Build GRCh38
rs374054864 Chr.11: 6200075-6200075 on Build GRCh38
rs10839531 Chr.11: 6199939-6199939 on Build GRCh38
rs753614040 Chr.11: 6199907-6199907 on Build GRCh38
rs780701943 Chr.11: 6200111-6200111 on Build GRCh38
rs529509371 Chr.11: 6200202-6200202 on Build GRCh38
rs747205909 Chr.11: 6199927-6199927 on Build GRCh38
rs749821038 Chr.11: 6199935-6199935 on Build GRCh38
rs772266778 Chr.11: 6199864-6199864 on Build GRCh38
rs74053127 Chr.11: 6199949-6199949 on Build GRCh38
rs115082738 Chr.11: 6199970-6199970 on Build GRCh38
rs1003628764 Chr.11: 6200176-6200176 on Build GRCh38
rs749716734 Chr.11: 6200091-6200091 on Build GRCh38
rs780010069 Chr.11: 6199925-6199925 on Build GRCh38
rs963591571 Chr.11: 6200209-6200209 on Build GRCh38
rs747759167 Chr.11: 6200116-6200116 on Build GRCh38
rs768792357 Chr.11: 6199929-6199929 on Build GRCh38
rs11040799 Chr.11: 6199984-6199984 on Build GRCh38
rs751648338 Chr.11: 6200056-6200056 on Build GRCh38
rs560734680 Chr.11: 6199959-6199959 on Build GRCh38
rs752914128 Chr.11: 6200074-6200074 on Build GRCh38
rs773927346 Chr.11: 6199934-6199934 on Build GRCh38
rs769008496 Chr.11: 6199872-6199872 on Build GRCh38
rs749294958 Chr.11: 6200307-6200307 on Build GRCh38
rs756704818 Chr.11: 6200070-6200071 on Build GRCh38
rs325609 Chr.11: 6200019-6200019 on Build GRCh38
rs778879292 Chr.11: 6199917-6199917 on Build GRCh38
rs770414159 Chr.11: 6199932-6199932 on Build GRCh38
rs767076899 Chr.11: 6199893-6199893 on Build GRCh38
rs139467264 Chr.11: 6200128-6200128 on Build GRCh38
rs973344982 Chr.11: 6200227-6200227 on Build GRCh38
rs982281939 Chr.11: 6200283-6200283 on Build GRCh38
rs762716587 Chr.11: 6200137-6200137 on Build GRCh38
rs183591651 Chr.11: 6200309-6200309 on Build GRCh38
rs540552724 Chr.11: 6200165-6200165 on Build GRCh38
rs767713682 Chr.11: 6200071-6200071 on Build GRCh38
rs780390546 Chr.11: 6199868-6199868 on Build GRCh38
rs757494744 Chr.11: 6200180-6200180 on Build GRCh38
rs779354546 Chr.11: 6200097-6200097 on Build GRCh38
rs767993008 Chr.11: 6200110-6200110 on Build GRCh38
rs538344497 Chr.11: 6200203-6200203 on Build GRCh38
rs767062220 Chr.11: 6200158-6200158 on Build GRCh38
rs755083328 Chr.11: 6200057-6200057 on Build GRCh38
rs769477980 Chr.11: 6200125-6200125 on Build GRCh38
rs751269901 Chr.11: 6200009-6200011 on Build GRCh38
rs761468618 Chr.11: 6200026-6200026 on Build GRCh38
rs746389643 Chr.11: 6200106-6200106 on Build GRCh38
rs776882569 Chr.11: 6199879-6199879 on Build GRCh38
rs772182219 Chr.11: 6200238-6200238 on Build GRCh38
rs1023762810 Chr.11: 6200261-6200261 on Build GRCh38
rs201702362 Chr.11: 6200023-6200023 on Build GRCh38
rs749769760 Chr.11: 6199992-6199992 on Build GRCh38
rs756207052 Chr.11: 6200205-6200205 on Build GRCh38
rs987220253 Chr.11: 6200029-6200029 on Build GRCh38
rs912051600 Chr.11: 6200055-6200055 on Build GRCh38
rs762651736 Chr.11: 6199986-6199987 on Build GRCh38
rs748977480 Chr.11: 6200212-6200212 on Build GRCh38
rs781590097 Chr.11: 6199930-6199930 on Build GRCh38
rs750496379 Chr.11: 6199920-6199920 on Build GRCh38
rs763806189 Chr.11: 6199988-6199990 on Build GRCh38
rs766442779 Chr.11: 6199968-6199968 on Build GRCh38
rs756987307 Chr.11: 6200041-6200041 on Build GRCh38
rs201841977 Chr.11: 6199938-6199938 on Build GRCh38
rs901118984 Chr.11: 6200192-6200192 on Build GRCh38
rs915843131 Chr.11: 6199989-6199989 on Build GRCh38
rs778249225 Chr.11: 6200085-6200085 on Build GRCh38
rs748521105 Chr.11: 6199981-6199981 on Build GRCh38
rs745995266 Chr.11: 6199860-6199860 on Build GRCh38
rs112509421 Chr.11: 6199965-6199965 on Build GRCh38
rs762372181 Chr.11: 6199882-6199882 on Build GRCh38
rs770900105 Chr.11: 6200142-6200142 on Build GRCh38
rs371030571 Chr.11: 6200051-6200051 on Build GRCh38
rs138922985 Chr.11: 6199997-6199997 on Build GRCh38
rs760953246 Chr.11: 6200169-6200169 on Build GRCh38
rs781331080 Chr.11: 6199979-6199979 on Build GRCh38
rs763906051 Chr.11: 6199904-6199904 on Build GRCh38
rs759357115 Chr.11: 6200235-6200235 on Build GRCh38
rs965374198 Chr.11: 6199996-6199996 on Build GRCh38
rs758934751 Chr.11: 6200189-6200189 on Build GRCh38
rs878942747 Chr.11: 6200093-6200093 on Build GRCh38
rs947004361 Chr.11: 6200124-6200124 on Build GRCh38
rs770465457 Chr.11: 6199885-6199885 on Build GRCh38
rs573085053 Chr.11: 6200163-6200163 on Build GRCh38
rs894023438 Chr.11: 6200098-6200098 on Build GRCh38
rs943566988 Chr.11: 6200059-6200059 on Build GRCh38
rs140475656 Chr.11: 6199911-6199911 on Build GRCh38
rs764464918 Chr.11: 6200172-6200172 on Build GRCh38
rs1006056488 Chr.11: 6200247-6200247 on Build GRCh38
rs142946621 Chr.11: 6200014-6200014 on Build GRCh38
rs150530085 Chr.11: 6200164-6200164 on Build GRCh38
rs12417049 Chr.11: 6200197-6200197 on Build GRCh38
rs928310053 Chr.11: 6199897-6199897 on Build GRCh38
rs764962797 Chr.11: 6200036-6200036 on Build GRCh38
rs750846382 Chr.11: 6200185-6200185 on Build GRCh38
rs149476132 Chr.11: 6200146-6200146 on Build GRCh38
rs73392966 Chr.11: 6199966-6199966 on Build GRCh38
rs578041524 Chr.11: 6200296-6200296 on Build GRCh38
rs191891210 Chr.11: 6199871-6199871 on Build GRCh38
rs775446638 Chr.11: 6199985-6199986 on Build GRCh38
rs756485245 Chr.11: 6199983-6199983 on Build GRCh38
rs755206993 Chr.11: 6199971-6199971 on Build GRCh38
rs138813300 Chr.11: 6199891-6199891 on Build GRCh38
rs982772137 Chr.11: 6200090-6200090 on Build GRCh38
rs761661371 Chr.11: 6199941-6199941 on Build GRCh38
rs767660421 Chr.11: 6200147-6200147 on Build GRCh38
rs969955154 Chr.11: 6200273-6200273 on Build GRCh38
rs1007233607 Chr.11: 6199977-6199977 on Build GRCh38
rs776198176 Chr.11: 6200025-6200025 on Build GRCh38
rs765065858 Chr.11: 6199980-6199981 on Build GRCh38
rs774227633 Chr.11: 6200234-6200234 on Build GRCh38
rs774091686 Chr.11: 6200145-6200145 on Build GRCh38

View Full Product Details