Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 32091180-32091686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RCN1
Entrez Gene ID
5954
Gene Name
reticulocalbin 1
Gene Aliases
HEL-S-84, PIG20, RCAL, RCN
UniGene
Hs.97887
Gene Chromosome Location
Chr.11: 32090931-32105726 on Build GRCh38
Transcripts
NM_002901.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
307547 c.210G>C p.K70N Chr.11: 32091406-32091406 on Build GRCh38
6416589 c.101G>A p.R34H Chr.11: 32091297-32091297 on Build GRCh38
1223447 c.205T>C p.S69P Chr.11: 32091401-32091401 on Build GRCh38
6408764 c.19G>T p.G7C Chr.11: 32091215-32091215 on Build GRCh38
4805356 c.235G>A p.E79K Chr.11: 32091431-32091431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113010055 Chr.11: 32091433-32091433 on Build GRCh38
rs555252762 Chr.11: 32091540-32091540 on Build GRCh38
rs534701713 Chr.11: 32091477-32091477 on Build GRCh38
rs1006325565 Chr.11: 32091321-32091321 on Build GRCh38
rs757128727 Chr.11: 32091214-32091214 on Build GRCh38
rs981590237 Chr.11: 32091261-32091261 on Build GRCh38
rs901993473 Chr.11: 32091291-32091291 on Build GRCh38
rs912524746 Chr.11: 32091641-32091641 on Build GRCh38
rs975399374 Chr.11: 32091469-32091469 on Build GRCh38
rs943593939 Chr.11: 32091289-32091289 on Build GRCh38
rs1039761213 Chr.11: 32091659-32091659 on Build GRCh38
rs745569609 Chr.11: 32091236-32091236 on Build GRCh38
rs929956167 Chr.11: 32091235-32091235 on Build GRCh38
rs537294360 Chr.11: 32091664-32091664 on Build GRCh38
rs895173058 Chr.11: 32091344-32091344 on Build GRCh38
rs774394053 Chr.11: 32091426-32091426 on Build GRCh38
rs751802902 Chr.11: 32091574-32091574 on Build GRCh38
rs914202429 Chr.11: 32091446-32091446 on Build GRCh38
rs1047899631 Chr.11: 32091484-32091484 on Build GRCh38
rs955758921 Chr.11: 32091430-32091430 on Build GRCh38
rs924403005 Chr.11: 32091272-32091272 on Build GRCh38
rs926375414 Chr.11: 32091584-32091584 on Build GRCh38
rs763126358 Chr.11: 32091386-32091386 on Build GRCh38
rs1001045520 Chr.11: 32091324-32091324 on Build GRCh38
rs1016282152 Chr.11: 32091204-32091204 on Build GRCh38
rs1031472386 Chr.11: 32091390-32091390 on Build GRCh38
rs943076611 Chr.11: 32091451-32091451 on Build GRCh38
rs1047502590 Chr.11: 32091631-32091631 on Build GRCh38
rs1022916631 Chr.11: 32091377-32091377 on Build GRCh38
rs944044743 Chr.11: 32091648-32091648 on Build GRCh38
rs145095137 Chr.11: 32091364-32091364 on Build GRCh38
rs867797073 Chr.11: 32091617-32091617 on Build GRCh38
rs929254321 Chr.11: 32091629-32091629 on Build GRCh38
rs771721732 Chr.11: 32091242-32091242 on Build GRCh38
rs963153994 Chr.11: 32091585-32091585 on Build GRCh38
rs970882942 Chr.11: 32091560-32091560 on Build GRCh38
rs375873456 Chr.11: 32091501-32091501 on Build GRCh38
rs1002758379 Chr.11: 32091570-32091570 on Build GRCh38
rs1037547608 Chr.11: 32091209-32091209 on Build GRCh38
rs767646275 Chr.11: 32091458-32091458 on Build GRCh38
rs113591704 Chr.11: 32091452-32091452 on Build GRCh38
rs367867618 Chr.11: 32091657-32091657 on Build GRCh38
rs971634786 Chr.11: 32091378-32091378 on Build GRCh38
rs1012287435 Chr.11: 32091558-32091558 on Build GRCh38
rs919158750 Chr.11: 32091610-32091610 on Build GRCh38
rs759665438 Chr.11: 32091448-32091448 on Build GRCh38
rs372792725 Chr.11: 32091419-32091419 on Build GRCh38
rs768116592 Chr.11: 32091280-32091280 on Build GRCh38
rs1023202519 Chr.11: 32091240-32091240 on Build GRCh38
rs919189802 Chr.11: 32091230-32091230 on Build GRCh38
rs1046832033 Chr.11: 32091559-32091559 on Build GRCh38
rs1037819393 Chr.11: 32091523-32091523 on Build GRCh38
rs1016250873 Chr.11: 32091322-32091322 on Build GRCh38
rs535060992 Chr.11: 32091528-32091528 on Build GRCh38
rs530769533 Chr.11: 32091279-32091279 on Build GRCh38
rs989739995 Chr.11: 32091287-32091287 on Build GRCh38
rs955893278 Chr.11: 32091577-32091577 on Build GRCh38
rs552400957 Chr.11: 32091288-32091288 on Build GRCh38
rs1047930083 Chr.11: 32091294-32091294 on Build GRCh38
rs891196060 Chr.11: 32091248-32091248 on Build GRCh38
rs1040194746 Chr.11: 32091286-32091286 on Build GRCh38
rs941873687 Chr.11: 32091207-32091207 on Build GRCh38
rs757251568 Chr.11: 32091475-32091475 on Build GRCh38
rs755142923 Chr.11: 32091666-32091666 on Build GRCh38
rs1804281 Chr.11: 32091413-32091413 on Build GRCh38
rs746555362 Chr.11: 32091262-32091262 on Build GRCh38
rs778835942 Chr.11: 32091233-32091233 on Build GRCh38
rs770204229 Chr.11: 32091345-32091345 on Build GRCh38
rs762250805 Chr.11: 32091295-32091295 on Build GRCh38
rs576319046 Chr.11: 32091546-32091546 on Build GRCh38
rs765161590 Chr.11: 32091478-32091478 on Build GRCh38
rs764801279 Chr.11: 32091354-32091354 on Build GRCh38
rs971284337 Chr.11: 32091250-32091250 on Build GRCh38
rs779482224 Chr.11: 32091255-32091255 on Build GRCh38
rs899932104 Chr.11: 32091661-32091661 on Build GRCh38
rs749942641 Chr.11: 32091245-32091246 on Build GRCh38
rs758082045 Chr.11: 32091481-32091481 on Build GRCh38
rs973228773 Chr.11: 32091593-32091593 on Build GRCh38
rs223045 Chr.11: 32091511-32091511 on Build GRCh38
rs1039309206 Chr.11: 32091290-32091290 on Build GRCh38

View Full Product Details