Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 36633395-36633922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf74
Entrez Gene ID
119710
Gene Name
chromosome 11 open reading frame 74
Gene Aliases
HEPIS
UniGene
Hs.159376
Gene Chromosome Location
Chr.11: 36594245-36659291 on Build GRCh38
Transcripts
NM_001276722.1 NM_001276723.1 NM_001276724.1 NM_001276725.1 NM_001276726.1 NM_001276727.1 NM_138787.3 NR_077243.1 XM_006718138.3 XM_011519885.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1475407 c.266C>A p.T89N Chr.11: 36633413-36633413 on Build GRCh38
3869127 c.262C>T p.R88C Chr.11: 36633409-36633409 on Build GRCh38
6614107 c.263G>A p.R88H Chr.11: 36633410-36633410 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs571727656 Chr.11: 36633547-36633547 on Build GRCh38
rs558669041 Chr.11: 36633871-36633871 on Build GRCh38
rs567586306 Chr.11: 36633613-36633613 on Build GRCh38
rs1030263818 Chr.11: 36633619-36633619 on Build GRCh38
rs375563958 Chr.11: 36633489-36633489 on Build GRCh38
rs941915165 Chr.11: 36633661-36633661 on Build GRCh38
rs534433861 Chr.11: 36633590-36633590 on Build GRCh38
rs556899780 Chr.11: 36633816-36633816 on Build GRCh38
rs780809673 Chr.11: 36633442-36633442 on Build GRCh38
rs948262315 Chr.11: 36633543-36633543 on Build GRCh38
rs887976342 Chr.11: 36633597-36633597 on Build GRCh38
rs1047982294 Chr.11: 36633634-36633634 on Build GRCh38
rs750834019 Chr.11: 36633423-36633423 on Build GRCh38
rs1055036220 Chr.11: 36633694-36633694 on Build GRCh38
rs752041535 Chr.11: 36633433-36633433 on Build GRCh38
rs779890419 Chr.11: 36633859-36633859 on Build GRCh38
rs968619729 Chr.11: 36633655-36633655 on Build GRCh38
rs897867259 Chr.11: 36633610-36633610 on Build GRCh38
rs910523434 Chr.11: 36633512-36633512 on Build GRCh38
rs527657366 Chr.11: 36633570-36633570 on Build GRCh38
rs929163029 Chr.11: 36633603-36633603 on Build GRCh38
rs977617935 Chr.11: 36633673-36633673 on Build GRCh38
rs1017793926 Chr.11: 36633878-36633878 on Build GRCh38
rs530144679 Chr.11: 36633516-36633516 on Build GRCh38
rs992525766 Chr.11: 36633727-36633727 on Build GRCh38
rs889094884 Chr.11: 36633625-36633625 on Build GRCh38
rs770645920 Chr.11: 36633481-36633481 on Build GRCh38
rs757712205 Chr.11: 36633576-36633576 on Build GRCh38
rs755534432 Chr.11: 36633439-36633439 on Build GRCh38
rs1031929799 Chr.11: 36633453-36633453 on Build GRCh38
rs992870509 Chr.11: 36633538-36633538 on Build GRCh38
rs199551949 Chr.11: 36633461-36633461 on Build GRCh38
rs1032231381 Chr.11: 36633854-36633854 on Build GRCh38
rs10836581 Chr.11: 36633849-36633849 on Build GRCh38
rs942187671 Chr.11: 36633600-36633600 on Build GRCh38
rs1021488872 Chr.11: 36633642-36633642 on Build GRCh38
rs758790282 Chr.11: 36633424-36633424 on Build GRCh38
rs144097691 Chr.11: 36633499-36633500 on Build GRCh38
rs867581828 Chr.11: 36633716-36633716 on Build GRCh38
rs909464555 Chr.11: 36633826-36633826 on Build GRCh38
rs902228326 Chr.11: 36633824-36633824 on Build GRCh38
rs762178798 Chr.11: 36633773-36633773 on Build GRCh38
rs1031426239 Chr.11: 36633698-36633698 on Build GRCh38
rs866904675 Chr.11: 36633786-36633786 on Build GRCh38
rs747586551 Chr.11: 36633460-36633460 on Build GRCh38
rs750959992 Chr.11: 36633870-36633870 on Build GRCh38
rs749103508 Chr.11: 36633477-36633477 on Build GRCh38
rs1013257172 Chr.11: 36633740-36633740 on Build GRCh38
rs780716378 Chr.11: 36633431-36633431 on Build GRCh38
rs948355417 Chr.11: 36633752-36633752 on Build GRCh38
rs10836580 Chr.11: 36633738-36633738 on Build GRCh38
rs572131584 Chr.11: 36633880-36633880 on Build GRCh38
rs547747025 Chr.11: 36633604-36633604 on Build GRCh38
rs1046211485 Chr.11: 36633753-36633753 on Build GRCh38
rs957656721 Chr.11: 36633867-36633867 on Build GRCh38
rs748942297 Chr.11: 36633898-36633898 on Build GRCh38
rs867004084 Chr.11: 36633582-36633582 on Build GRCh38
rs1007458457 Chr.11: 36633628-36633628 on Build GRCh38
rs964715763 Chr.11: 36633879-36633880 on Build GRCh38
rs570259957 Chr.11: 36633825-36633825 on Build GRCh38
rs3758874 Chr.11: 36633539-36633539 on Build GRCh38

View Full Product Details