Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44599974-44600466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD82
Entrez Gene ID
3732
Gene Name
CD82 molecule
Gene Aliases
4F9, C33, GR15, IA4, KAI1, R2, SAR2, ST6, TSPAN27
UniGene
Hs.527778
Gene Chromosome Location
Chr.11: 44565591-44620363 on Build GRCh38
Transcripts
NM_001024844.1 NM_002231.3 XM_006718223.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1979335 c.135G>T p.L45L Chr.11: 44600229-44600229 on Build GRCh38
3967504 c.117C>T p.S39S Chr.11: 44600211-44600211 on Build GRCh38
7331707 c.72C>T p.G24G Chr.11: 44600166-44600166 on Build GRCh38
5838530 c.73G>A p.A25T Chr.11: 44600167-44600167 on Build GRCh38
355978 c.123C>A p.F41L Chr.11: 44600217-44600217 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs138833231 Chr.11: 44600202-44600202 on Build GRCh38
rs761468393 Chr.11: 44600187-44600187 on Build GRCh38
rs545728893 Chr.11: 44600312-44600312 on Build GRCh38
rs1050155633 Chr.11: 44600104-44600104 on Build GRCh38
rs758337930 Chr.11: 44600203-44600203 on Build GRCh38
rs3217345 Chr.11: 44600149-44600151 on Build GRCh38
rs774964064 Chr.11: 44600167-44600167 on Build GRCh38
rs756411683 Chr.11: 44600218-44600218 on Build GRCh38
rs746415347 Chr.11: 44600254-44600254 on Build GRCh38
rs989641328 Chr.11: 44600435-44600435 on Build GRCh38
rs149293292 Chr.11: 44600434-44600434 on Build GRCh38
rs945193697 Chr.11: 44600161-44600161 on Build GRCh38
rs377535419 Chr.11: 44600198-44600198 on Build GRCh38
rs920858289 Chr.11: 44600351-44600351 on Build GRCh38
rs972286056 Chr.11: 44600350-44600350 on Build GRCh38
rs781480823 Chr.11: 44600213-44600213 on Build GRCh38
rs1036646430 Chr.11: 44600009-44600009 on Build GRCh38
rs778085878 Chr.11: 44600224-44600224 on Build GRCh38
rs771239010 Chr.11: 44600242-44600242 on Build GRCh38
rs371613857 Chr.11: 44600257-44600257 on Build GRCh38
rs372427368 Chr.11: 44600335-44600335 on Build GRCh38
rs897991407 Chr.11: 44600328-44600328 on Build GRCh38
rs554121820 Chr.11: 44600185-44600185 on Build GRCh38
rs370003971 Chr.11: 44600288-44600288 on Build GRCh38
rs1000615015 Chr.11: 44600216-44600216 on Build GRCh38
rs942133977 Chr.11: 44600243-44600243 on Build GRCh38
rs150533529 Chr.11: 44600230-44600230 on Build GRCh38
rs1025223025 Chr.11: 44600075-44600075 on Build GRCh38
rs1001892328 Chr.11: 44600390-44600390 on Build GRCh38
rs1037757502 Chr.11: 44600273-44600273 on Build GRCh38
rs920257157 Chr.11: 44600076-44600076 on Build GRCh38
rs917463541 Chr.11: 44600001-44600001 on Build GRCh38
rs771489038 Chr.11: 44600155-44600155 on Build GRCh38
rs1025681764 Chr.11: 44600342-44600342 on Build GRCh38
rs930324594 Chr.11: 44600085-44600085 on Build GRCh38
rs535717592 Chr.11: 44600166-44600166 on Build GRCh38
rs892281407 Chr.11: 44600442-44600442 on Build GRCh38
rs74886044 Chr.11: 44600114-44600114 on Build GRCh38
rs930293793 Chr.11: 44600376-44600376 on Build GRCh38
rs373431121 Chr.11: 44600150-44600150 on Build GRCh38
rs760055528 Chr.11: 44600164-44600164 on Build GRCh38
rs543117766 Chr.11: 44600362-44600362 on Build GRCh38
rs763666572 Chr.11: 44600180-44600180 on Build GRCh38
rs772563836 Chr.11: 44600256-44600256 on Build GRCh38
rs992948488 Chr.11: 44600287-44600287 on Build GRCh38
rs940917315 Chr.11: 44600006-44600006 on Build GRCh38
rs779313004 Chr.11: 44600244-44600244 on Build GRCh38
rs199846636 Chr.11: 44600116-44600116 on Build GRCh38
rs778904702 Chr.11: 44600428-44600428 on Build GRCh38
rs888871804 Chr.11: 44600111-44600111 on Build GRCh38
rs913735286 Chr.11: 44600145-44600145 on Build GRCh38
rs989203900 Chr.11: 44600139-44600139 on Build GRCh38
rs749539429 Chr.11: 44600171-44600171 on Build GRCh38
rs753062672 Chr.11: 44600033-44600033 on Build GRCh38
rs148396643 Chr.11: 44600189-44600189 on Build GRCh38
rs970384489 Chr.11: 44600088-44600088 on Build GRCh38
rs569011124 Chr.11: 44600319-44600319 on Build GRCh38
rs774679121 Chr.11: 44600160-44600160 on Build GRCh38
rs760383801 Chr.11: 44600169-44600169 on Build GRCh38
rs760360335 Chr.11: 44600038-44600038 on Build GRCh38
rs576017432 Chr.11: 44600357-44600357 on Build GRCh38
rs764433314 Chr.11: 44600128-44600129 on Build GRCh38
rs994189300 Chr.11: 44600002-44600002 on Build GRCh38
rs372637755 Chr.11: 44600112-44600112 on Build GRCh38
rs910257337 Chr.11: 44600403-44600403 on Build GRCh38
rs906770543 Chr.11: 44600381-44600381 on Build GRCh38

View Full Product Details