Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47179146-47179641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR6745
Entrez Gene ID
102466726
Gene Name
microRNA 6745
Gene Aliases
hsa-mir-6745
UniGene
Gene Chromosome Location
Chr.11: 47179611-47179737 on Build GRCh38
Transcripts
NR_106803.1 NM_001184974.1 NM_001184975.1 NM_016223.4

Gene Symbol
PACSIN3
Entrez Gene ID
29763
Gene Name
protein kinase C and casein kinase substrate in neurons 3
Gene Aliases
SDPIII
UniGene
Hs.334639
Gene Chromosome Location
Chr.11: 47177522-47186459 on Build GRCh38
Transcripts
NR_106803.1 NM_001184974.1 NM_001184975.1 NM_016223.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5469612 c.854G>A p.R285H Chr.11: 47179205-47179205 on Build GRCh38
6736812 c.897C>T p.F299F Chr.11: 47179162-47179162 on Build GRCh38
3809302 c.633A>C p.A211A Chr.11: 47179557-47179557 on Build GRCh38
3809301 c.634G>A p.E212K Chr.11: 47179556-47179556 on Build GRCh38
3809300 c.839A>G p.D280G Chr.11: 47179220-47179220 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763237797 Chr.11: 47179255-47179255 on Build GRCh38
rs1002689739 Chr.11: 47179378-47179378 on Build GRCh38
rs368292408 Chr.11: 47179302-47179302 on Build GRCh38
rs754923304 Chr.11: 47179505-47179505 on Build GRCh38
rs756655757 Chr.11: 47179214-47179214 on Build GRCh38
rs34915644 Chr.11: 47179478-47179478 on Build GRCh38
rs373614899 Chr.11: 47179262-47179262 on Build GRCh38
rs370846491 Chr.11: 47179297-47179297 on Build GRCh38
rs535287709 Chr.11: 47179617-47179617 on Build GRCh38
rs549826440 Chr.11: 47179534-47179534 on Build GRCh38
rs147818380 Chr.11: 47179263-47179263 on Build GRCh38
rs774414693 Chr.11: 47179410-47179410 on Build GRCh38
rs202065451 Chr.11: 47179483-47179483 on Build GRCh38
rs995829702 Chr.11: 47179350-47179350 on Build GRCh38
rs771187396 Chr.11: 47179409-47179409 on Build GRCh38
rs748047182 Chr.11: 47179482-47179482 on Build GRCh38
rs756957083 Chr.11: 47179307-47179307 on Build GRCh38
rs746976937 Chr.11: 47179377-47179377 on Build GRCh38
rs201662110 Chr.11: 47179241-47179241 on Build GRCh38
rs770240765 Chr.11: 47179580-47179580 on Build GRCh38
rs773730979 Chr.11: 47179585-47179585 on Build GRCh38
rs769245550 Chr.11: 47179491-47179491 on Build GRCh38
rs765047382 Chr.11: 47179467-47179467 on Build GRCh38
rs749992719 Chr.11: 47179324-47179324 on Build GRCh38
rs765990359 Chr.11: 47179477-47179477 on Build GRCh38
rs377108623 Chr.11: 47179195-47179195 on Build GRCh38
rs779053098 Chr.11: 47179484-47179484 on Build GRCh38
rs572682863 Chr.11: 47179581-47179581 on Build GRCh38
rs757921678 Chr.11: 47179472-47179472 on Build GRCh38
rs778543860 Chr.11: 47179320-47179320 on Build GRCh38
rs773246741 Chr.11: 47179247-47179247 on Build GRCh38
rs751126553 Chr.11: 47179517-47179517 on Build GRCh38
rs759031295 Chr.11: 47179611-47179611 on Build GRCh38
rs867450385 Chr.11: 47179376-47179376 on Build GRCh38
rs752865412 Chr.11: 47179351-47179351 on Build GRCh38
rs886934494 Chr.11: 47179365-47179365 on Build GRCh38
rs530734643 Chr.11: 47179535-47179535 on Build GRCh38
rs761275779 Chr.11: 47179426-47179426 on Build GRCh38
rs375065601 Chr.11: 47179299-47179299 on Build GRCh38
rs772101272 Chr.11: 47179488-47179488 on Build GRCh38
rs761341362 Chr.11: 47179456-47179456 on Build GRCh38
rs555969464 Chr.11: 47179574-47179574 on Build GRCh38
rs770687710 Chr.11: 47179610-47179610 on Build GRCh38
rs772870340 Chr.11: 47179227-47179227 on Build GRCh38
rs569802805 Chr.11: 47179546-47179546 on Build GRCh38
rs745750554 Chr.11: 47179222-47179222 on Build GRCh38
rs753542803 Chr.11: 47179206-47179206 on Build GRCh38
rs199902250 Chr.11: 47179490-47179490 on Build GRCh38
rs762723225 Chr.11: 47179503-47179503 on Build GRCh38
rs749939339 Chr.11: 47179468-47179468 on Build GRCh38
rs746844467 Chr.11: 47179244-47179244 on Build GRCh38
rs563675998 Chr.11: 47179228-47179228 on Build GRCh38
rs371464465 Chr.11: 47179578-47179578 on Build GRCh38
rs762324925 Chr.11: 47179493-47179493 on Build GRCh38
rs924893885 Chr.11: 47179538-47179538 on Build GRCh38
rs147000809 Chr.11: 47179415-47179415 on Build GRCh38
rs765708998 Chr.11: 47179495-47179495 on Build GRCh38
rs755842192 Chr.11: 47179547-47179547 on Build GRCh38
rs777576673 Chr.11: 47179553-47179553 on Build GRCh38
rs371598454 Chr.11: 47179391-47179391 on Build GRCh38
rs746042413 Chr.11: 47179563-47179563 on Build GRCh38
rs987663238 Chr.11: 47179552-47179552 on Build GRCh38
rs758404736 Chr.11: 47179328-47179328 on Build GRCh38
rs770041815 Chr.11: 47179246-47179246 on Build GRCh38
rs975298471 Chr.11: 47179354-47179354 on Build GRCh38
rs754961841 Chr.11: 47179479-47179479 on Build GRCh38
rs780823150 Chr.11: 47179481-47179481 on Build GRCh38
rs200875557 Chr.11: 47179487-47179487 on Build GRCh38
rs768266845 Chr.11: 47179381-47179381 on Build GRCh38
rs759674365 Chr.11: 47179414-47179414 on Build GRCh38
rs781069008 Chr.11: 47179390-47179390 on Build GRCh38
rs767850572 Chr.11: 47179179-47179179 on Build GRCh38
rs753048704 Chr.11: 47179273-47179273 on Build GRCh38
rs141460458 Chr.11: 47179267-47179267 on Build GRCh38
rs753103854 Chr.11: 47179185-47179185 on Build GRCh38
rs765753216 Chr.11: 47179511-47179511 on Build GRCh38
rs772929606 Chr.11: 47179500-47179500 on Build GRCh38
rs777102189 Chr.11: 47179446-47179446 on Build GRCh38
rs150148866 Chr.11: 47179215-47179215 on Build GRCh38
rs762669881 Chr.11: 47179594-47179594 on Build GRCh38
rs548013601 Chr.11: 47179334-47179334 on Build GRCh38
rs149470424 Chr.11: 47179564-47179564 on Build GRCh38
rs140983686 Chr.11: 47179194-47179194 on Build GRCh38
rs747180932 Chr.11: 47179566-47179566 on Build GRCh38
rs199638473 Chr.11: 47179363-47179363 on Build GRCh38
rs368057358 Chr.11: 47179569-47179569 on Build GRCh38
rs138948043 Chr.11: 47179440-47179440 on Build GRCh38

View Full Product Details