Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47573327-47573839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KBTBD4
Entrez Gene ID
55709
Gene Name
kelch repeat and BTB domain containing 4
Gene Aliases
BKLHD4, HSPC252
UniGene
Hs.718483
Gene Chromosome Location
Chr.11: 47572197-47579015 on Build GRCh38
Transcripts
NM_001318716.1 NM_001318717.1 NM_001318718.1 NM_001318719.1 NM_001318720.1 NM_001318721.1 NM_001318722.1 NM_001318723.1 NM_001318724.1 NM_001318725.1 NM_016506.6 NM_018095.5 XM_017018009.1 NM_001143984.1 NM_175732.2

Gene Symbol
PTPMT1
Entrez Gene ID
114971
Gene Name
protein tyrosine phosphatase, mitochondrial 1
Gene Aliases
DUSP23, MOSP, PLIP, PNAS-129
UniGene
Hs.656205
Gene Chromosome Location
Chr.11: 47565430-47573461 on Build GRCh38
Transcripts
NM_001318716.1 NM_001318717.1 NM_001318718.1 NM_001318719.1 NM_001318720.1 NM_001318721.1 NM_001318722.1 NM_001318723.1 NM_001318724.1 NM_001318725.1 NM_016506.6 NM_018095.5 XM_017018009.1 NM_001143984.1 NM_175732.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4868156 c.1171A>T p.N391Y Chr.11: 47573364-47573364 on Build GRCh38
5999490 c.855C>T p.G285G Chr.11: 47573680-47573680 on Build GRCh38
4821334 c.856G>C p.D286H Chr.11: 47573631-47573631 on Build GRCh38
7416886 c.930G>A p.W310* Chr.11: 47573557-47573557 on Build GRCh38
4574404 c.981C>T p.P327P Chr.11: 47573506-47573506 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768581356 Chr.11: 47573620-47573620 on Build GRCh38
rs909392785 Chr.11: 47573456-47573456 on Build GRCh38
rs145366063 Chr.11: 47573706-47573706 on Build GRCh38
rs967832403 Chr.11: 47573356-47573356 on Build GRCh38
rs753654999 Chr.11: 47573476-47573476 on Build GRCh38
rs773599306 Chr.11: 47573429-47573429 on Build GRCh38
rs963573962 Chr.11: 47573813-47573813 on Build GRCh38
rs760765614 Chr.11: 47573786-47573786 on Build GRCh38
rs778273465 Chr.11: 47573697-47573697 on Build GRCh38
rs923366379 Chr.11: 47573369-47573369 on Build GRCh38
rs779031903 Chr.11: 47573379-47573380 on Build GRCh38
rs759804817 Chr.11: 47573651-47573651 on Build GRCh38
rs755548747 Chr.11: 47573400-47573400 on Build GRCh38
rs752807696 Chr.11: 47573686-47573686 on Build GRCh38
rs769674732 Chr.11: 47573752-47573752 on Build GRCh38
rs780056745 Chr.11: 47573387-47573387 on Build GRCh38
rs367925558 Chr.11: 47573632-47573632 on Build GRCh38
rs903787949 Chr.11: 47573769-47573769 on Build GRCh38
rs867676589 Chr.11: 47573460-47573460 on Build GRCh38
rs944140893 Chr.11: 47573662-47573662 on Build GRCh38
rs750660482 Chr.11: 47573366-47573366 on Build GRCh38
rs745403900 Chr.11: 47573582-47573582 on Build GRCh38
rs754182984 Chr.11: 47573700-47573700 on Build GRCh38
rs758977537 Chr.11: 47573434-47573434 on Build GRCh38
rs754415608 Chr.11: 47573790-47573790 on Build GRCh38
rs137865982 Chr.11: 47573357-47573357 on Build GRCh38
rs756681498 Chr.11: 47573533-47573533 on Build GRCh38
rs931437850 Chr.11: 47573541-47573541 on Build GRCh38
rs770492766 Chr.11: 47573572-47573572 on Build GRCh38
rs11039302 Chr.11: 47573499-47573499 on Build GRCh38
rs770334695 Chr.11: 47573417-47573417 on Build GRCh38
rs767471949 Chr.11: 47573447-47573447 on Build GRCh38
rs147179417 Chr.11: 47573461-47573461 on Build GRCh38
rs779554052 Chr.11: 47573725-47573725 on Build GRCh38
rs748140900 Chr.11: 47573615-47573615 on Build GRCh38
rs61751915 Chr.11: 47573770-47573770 on Build GRCh38
rs765630705 Chr.11: 47573485-47573485 on Build GRCh38
rs746451387 Chr.11: 47573732-47573732 on Build GRCh38
rs139512073 Chr.11: 47573506-47573506 on Build GRCh38
rs776295137 Chr.11: 47573625-47573625 on Build GRCh38
rs756698270 Chr.11: 47573688-47573688 on Build GRCh38
rs976089966 Chr.11: 47573726-47573726 on Build GRCh38
rs775712548 Chr.11: 47573785-47573785 on Build GRCh38
rs781751790 Chr.11: 47573405-47573405 on Build GRCh38
rs911308940 Chr.11: 47573660-47573660 on Build GRCh38
rs146789563 Chr.11: 47573761-47573761 on Build GRCh38
rs764529147 Chr.11: 47573788-47573788 on Build GRCh38
rs781403257 Chr.11: 47573528-47573528 on Build GRCh38
rs745558885 Chr.11: 47573715-47573715 on Build GRCh38
rs1036617814 Chr.11: 47573505-47573505 on Build GRCh38
rs200513185 Chr.11: 47573373-47573373 on Build GRCh38
rs372051017 Chr.11: 47573650-47573650 on Build GRCh38
rs775355395 Chr.11: 47573449-47573449 on Build GRCh38
rs35920762 Chr.11: 47573707-47573707 on Build GRCh38
rs757673059 Chr.11: 47573793-47573793 on Build GRCh38
rs758741599 Chr.11: 47573537-47573537 on Build GRCh38
rs754116406 Chr.11: 47573448-47573448 on Build GRCh38
rs143529938 Chr.11: 47573431-47573431 on Build GRCh38
rs965703669 Chr.11: 47573717-47573717 on Build GRCh38
rs375208548 Chr.11: 47573509-47573509 on Build GRCh38
rs748955906 Chr.11: 47573430-47573430 on Build GRCh38
rs754462828 Chr.11: 47573794-47573794 on Build GRCh38
rs149622767 Chr.11: 47573530-47573530 on Build GRCh38
rs757591605 Chr.11: 47573702-47573702 on Build GRCh38
rs771700327 Chr.11: 47573722-47573722 on Build GRCh38
rs767787104 Chr.11: 47573781-47573781 on Build GRCh38
rs748549438 Chr.11: 47573411-47573411 on Build GRCh38
rs774949831 Chr.11: 47573597-47573597 on Build GRCh38
rs372227345 Chr.11: 47573365-47573365 on Build GRCh38
rs144325348 Chr.11: 47573579-47573579 on Build GRCh38
rs1030355641 Chr.11: 47573731-47573731 on Build GRCh38
rs767776935 Chr.11: 47573652-47573652 on Build GRCh38
rs749606273 Chr.11: 47573540-47573540 on Build GRCh38
rs780457703 Chr.11: 47573741-47573741 on Build GRCh38
rs747054418 Chr.11: 47573391-47573391 on Build GRCh38
rs142438274 Chr.11: 47573746-47573746 on Build GRCh38
rs766534364 Chr.11: 47573641-47573641 on Build GRCh38
rs755063316 Chr.11: 47573511-47573511 on Build GRCh38
rs758643174 Chr.11: 47573385-47573385 on Build GRCh38
rs760663862 Chr.11: 47573598-47573598 on Build GRCh38
rs755861672 Chr.11: 47573355-47573355 on Build GRCh38
rs1046517505 Chr.11: 47573571-47573571 on Build GRCh38
rs760584018 Chr.11: 47573459-47573459 on Build GRCh38
rs773744022 Chr.11: 47573432-47573432 on Build GRCh38
rs768677773 Chr.11: 47573379-47573402 on Build GRCh38

View Full Product Details