Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 49153754-49154261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOLH1
Entrez Gene ID
2346
Gene Name
folate hydrolase (prostate-specific membrane antigen) 1
Gene Aliases
FGCP, FOLH, GCP2, GCPII, NAALAD1, NAALAdase, PSM, PSMA, mGCP
UniGene
Hs.645352
Gene Chromosome Location
Chr.11: 49145094-49208706 on Build GRCh38
Transcripts
NM_001014986.1 NM_001193471.1 NM_001193472.1 NM_001193473.1 NM_004476.1 XM_017017432.1 XM_017017433.1 XM_017017434.1 XM_017017435.1 XM_017017436.1 XM_017017437.1 XM_017017438.1 XM_017017439.1 XM_017017440.1 XM_017017441.1 XM_017017442.1 XM_017017443.1 XM_017017444.1 XM_017017445.1 XM_017017446.1 XM_017017447.1 XM_017017448.1 XM_017017449.1 XM_017017450.1 XM_017017451.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4532174 c.1861G>A p.E621K Chr.11: 49154255-49154255 on Build GRCh38
6132578 c.1910A>G p.K637R Chr.11: 49153906-49153906 on Build GRCh38
429108 c.1884A>G p.S628S Chr.11: 49154232-49154232 on Build GRCh38
6578562 c.1958T>A p.F653Y Chr.11: 49153858-49153858 on Build GRCh38
320479 c.1927G>T p.A643S Chr.11: 49153889-49153889 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780989203 Chr.11: 49154181-49154181 on Build GRCh38
rs370043154 Chr.11: 49153825-49153825 on Build GRCh38
rs759180548 Chr.11: 49154214-49154214 on Build GRCh38
rs1052108857 Chr.11: 49154117-49154117 on Build GRCh38
rs757724515 Chr.11: 49153905-49153905 on Build GRCh38
rs375319677 Chr.11: 49153800-49153800 on Build GRCh38
rs566020349 Chr.11: 49153889-49153889 on Build GRCh38
rs551721616 Chr.11: 49154202-49154202 on Build GRCh38
rs779001124 Chr.11: 49153808-49153808 on Build GRCh38
rs61886483 Chr.11: 49153999-49153999 on Build GRCh38
rs772351958 Chr.11: 49153814-49153814 on Build GRCh38
rs567082334 Chr.11: 49153804-49153804 on Build GRCh38
rs531040952 Chr.11: 49153934-49153934 on Build GRCh38
rs747902803 Chr.11: 49154183-49154183 on Build GRCh38
rs754177008 Chr.11: 49153904-49153904 on Build GRCh38
rs137991256 Chr.11: 49153923-49153923 on Build GRCh38
rs993482340 Chr.11: 49153954-49153954 on Build GRCh38
rs1030292837 Chr.11: 49153776-49153776 on Build GRCh38
rs757078050 Chr.11: 49153950-49153950 on Build GRCh38
rs1017631643 Chr.11: 49153792-49153793 on Build GRCh38
rs762079463 Chr.11: 49154233-49154233 on Build GRCh38
rs546821955 Chr.11: 49153885-49153885 on Build GRCh38
rs747198634 Chr.11: 49153977-49153977 on Build GRCh38
rs2988342 Chr.11: 49154237-49154237 on Build GRCh38
rs769699407 Chr.11: 49154191-49154191 on Build GRCh38
rs773124927 Chr.11: 49154196-49154196 on Build GRCh38
rs890453344 Chr.11: 49153778-49153782 on Build GRCh38
rs780327979 Chr.11: 49153965-49153965 on Build GRCh38
rs747290318 Chr.11: 49153824-49153824 on Build GRCh38
rs746442269 Chr.11: 49153940-49153940 on Build GRCh38
rs768788468 Chr.11: 49154180-49154180 on Build GRCh38
rs529840097 Chr.11: 49153806-49153806 on Build GRCh38
rs199517010 Chr.11: 49153875-49153875 on Build GRCh38
rs761766231 Chr.11: 49153836-49153836 on Build GRCh38
rs767089077 Chr.11: 49154229-49154229 on Build GRCh38
rs764479615 Chr.11: 49153895-49153895 on Build GRCh38
rs765103565 Chr.11: 49153854-49153854 on Build GRCh38
rs535155505 Chr.11: 49153902-49153902 on Build GRCh38
rs370390473 Chr.11: 49154209-49154209 on Build GRCh38
rs890402284 Chr.11: 49154120-49154120 on Build GRCh38
rs190062771 Chr.11: 49154068-49154068 on Build GRCh38
rs762928518 Chr.11: 49154197-49154197 on Build GRCh38
rs144512496 Chr.11: 49153857-49153857 on Build GRCh38
rs745957524 Chr.11: 49153809-49153809 on Build GRCh38
rs980455677 Chr.11: 49154043-49154043 on Build GRCh38
rs945268688 Chr.11: 49154112-49154112 on Build GRCh38
rs657335 Chr.11: 49153796-49153796 on Build GRCh38
rs913730128 Chr.11: 49154109-49154109 on Build GRCh38
rs770223876 Chr.11: 49153820-49153821 on Build GRCh38
rs754163774 Chr.11: 49154232-49154232 on Build GRCh38
rs1007550583 Chr.11: 49153783-49153783 on Build GRCh38
rs776325789 Chr.11: 49153828-49153828 on Build GRCh38
rs574319046 Chr.11: 49154139-49154139 on Build GRCh38
rs762448723 Chr.11: 49153872-49153872 on Build GRCh38
rs970367885 Chr.11: 49154042-49154042 on Build GRCh38
rs557675288 Chr.11: 49154067-49154067 on Build GRCh38
rs770398257 Chr.11: 49154200-49154200 on Build GRCh38
rs775849303 Chr.11: 49153816-49153816 on Build GRCh38
rs143005014 Chr.11: 49154057-49154057 on Build GRCh38

View Full Product Details