Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 55812172-55812677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR5L1
Entrez Gene ID
219437
Gene Name
olfactory receptor family 5 subfamily L member 1 (gene/pseudogene)
Gene Aliases
OR11-151, OST262
UniGene
Hs.553623
Gene Chromosome Location
Chr.11: 55811467-55812402 on Build GRCh38
Transcripts
NM_001004738.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4194826 c.809C>T p.A270V Chr.11: 55812275-55812275 on Build GRCh38
6735013 c.744C>A p.I248I Chr.11: 55812210-55812210 on Build GRCh38
2149159 c.817G>T p.V273L Chr.11: 55812283-55812283 on Build GRCh38
5018514 c.820G>T p.A274S Chr.11: 55812286-55812286 on Build GRCh38
6133090 c.729_730CC>TA p.H244N Chr.11: 55812195-55812196 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777592351 Chr.11: 55812256-55812258 on Build GRCh38
rs373838130 Chr.11: 55812229-55812229 on Build GRCh38
rs748815260 Chr.11: 55812376-55812376 on Build GRCh38
rs914319252 Chr.11: 55812585-55812585 on Build GRCh38
rs758557451 Chr.11: 55812354-55812354 on Build GRCh38
rs574575780 Chr.11: 55812488-55812488 on Build GRCh38
rs556607867 Chr.11: 55812404-55812404 on Build GRCh38
rs772313759 Chr.11: 55812275-55812275 on Build GRCh38
rs761390190 Chr.11: 55812227-55812227 on Build GRCh38
rs778351093 Chr.11: 55812269-55812269 on Build GRCh38
rs769390899 Chr.11: 55812290-55812290 on Build GRCh38
rs765677393 Chr.11: 55812396-55812396 on Build GRCh38
rs754693929 Chr.11: 55812421-55812421 on Build GRCh38
rs199917722 Chr.11: 55812448-55812448 on Build GRCh38
rs1054531283 Chr.11: 55812312-55812312 on Build GRCh38
rs573607602 Chr.11: 55812633-55812633 on Build GRCh38
rs61732350 Chr.11: 55812392-55812392 on Build GRCh38
rs756015487 Chr.11: 55812436-55812436 on Build GRCh38
rs775944699 Chr.11: 55812226-55812226 on Build GRCh38
rs746819194 Chr.11: 55812279-55812279 on Build GRCh38
rs533270077 Chr.11: 55812453-55812453 on Build GRCh38
rs918193257 Chr.11: 55812473-55812473 on Build GRCh38
rs745542153 Chr.11: 55812515-55812515 on Build GRCh38
rs745378452 Chr.11: 55812420-55812421 on Build GRCh38
rs766574619 Chr.11: 55812419-55812419 on Build GRCh38
rs757091033 Chr.11: 55812383-55812383 on Build GRCh38
rs267602978 Chr.11: 55812194-55812194 on Build GRCh38
rs879027365 Chr.11: 55812454-55812454 on Build GRCh38
rs990779268 Chr.11: 55812459-55812459 on Build GRCh38
rs199628629 Chr.11: 55812360-55812360 on Build GRCh38
rs146137791 Chr.11: 55812291-55812291 on Build GRCh38
rs1007222597 Chr.11: 55812349-55812349 on Build GRCh38
rs1038705126 Chr.11: 55812627-55812627 on Build GRCh38
rs368833257 Chr.11: 55812313-55812313 on Build GRCh38
rs139907396 Chr.11: 55812394-55812394 on Build GRCh38
rs540023295 Chr.11: 55812634-55812634 on Build GRCh38
rs1054243135 Chr.11: 55812562-55812562 on Build GRCh38
rs777927405 Chr.11: 55812439-55812439 on Build GRCh38
rs780258092 Chr.11: 55812355-55812355 on Build GRCh38
rs779265776 Chr.11: 55812203-55812203 on Build GRCh38
rs148919465 Chr.11: 55812511-55812511 on Build GRCh38
rs746802821 Chr.11: 55812358-55812358 on Build GRCh38
rs775933074 Chr.11: 55812295-55812295 on Build GRCh38
rs759953541 Chr.11: 55812246-55812246 on Build GRCh38
rs934413639 Chr.11: 55812520-55812520 on Build GRCh38
rs764561184 Chr.11: 55812384-55812384 on Build GRCh38
rs61735738 Chr.11: 55812307-55812307 on Build GRCh38
rs559759119 Chr.11: 55812387-55812387 on Build GRCh38
rs761192799 Chr.11: 55812303-55812303 on Build GRCh38
rs770706404 Chr.11: 55812379-55812379 on Build GRCh38
rs752455991 Chr.11: 55812272-55812272 on Build GRCh38
rs145975508 Chr.11: 55812198-55812198 on Build GRCh38
rs1012855448 Chr.11: 55812458-55812458 on Build GRCh38
rs780783427 Chr.11: 55812514-55812514 on Build GRCh38
rs774209103 Chr.11: 55812380-55812380 on Build GRCh38
rs1005968869 Chr.11: 55812638-55812638 on Build GRCh38
rs12790505 Chr.11: 55812325-55812325 on Build GRCh38
rs901776619 Chr.11: 55812632-55812632 on Build GRCh38
rs763833746 Chr.11: 55812300-55812300 on Build GRCh38
rs954330267 Chr.11: 55812512-55812512 on Build GRCh38
rs201671558 Chr.11: 55812294-55812294 on Build GRCh38
rs267602979 Chr.11: 55812343-55812343 on Build GRCh38
rs764498675 Chr.11: 55812306-55812306 on Build GRCh38
rs753140877 Chr.11: 55812261-55812261 on Build GRCh38
rs768601431 Chr.11: 55812221-55812221 on Build GRCh38
rs533061973 Chr.11: 55812231-55812231 on Build GRCh38
rs886241439 Chr.11: 55812639-55812639 on Build GRCh38
rs571617569 Chr.11: 55812416-55812416 on Build GRCh38
rs772796247 Chr.11: 55812365-55812365 on Build GRCh38
rs1004030869 Chr.11: 55812423-55812423 on Build GRCh38
rs887647681 Chr.11: 55812462-55812462 on Build GRCh38
rs746622169 Chr.11: 55812295-55812295 on Build GRCh38
rs746128736 Chr.11: 55812274-55812274 on Build GRCh38
rs775377558 Chr.11: 55812208-55812208 on Build GRCh38
rs771904689 Chr.11: 55812207-55812207 on Build GRCh38
rs1021469330 Chr.11: 55812461-55812461 on Build GRCh38
rs114117519 Chr.11: 55812420-55812420 on Build GRCh38
rs978682369 Chr.11: 55812499-55812499 on Build GRCh38
rs751055029 Chr.11: 55812508-55812508 on Build GRCh38
rs950884909 Chr.11: 55812430-55812430 on Build GRCh38
rs747914946 Chr.11: 55812433-55812433 on Build GRCh38
rs139839955 Chr.11: 55812253-55812253 on Build GRCh38
rs760567723 Chr.11: 55812215-55812215 on Build GRCh38
rs940254899 Chr.11: 55812560-55812560 on Build GRCh38
rs757254282 Chr.11: 55812311-55812311 on Build GRCh38
rs747594618 Chr.11: 55812288-55812288 on Build GRCh38
rs776697056 Chr.11: 55812285-55812285 on Build GRCh38
rs200650148 Chr.11: 55812264-55812264 on Build GRCh38
rs750144440 Chr.11: 55812243-55812243 on Build GRCh38
rs1006459183 Chr.11: 55812646-55812646 on Build GRCh38
rs778678630 Chr.11: 55812449-55812449 on Build GRCh38
rs769647526 Chr.11: 55812361-55812361 on Build GRCh38
rs768409934 Chr.11: 55812284-55812284 on Build GRCh38
rs750525391 Chr.11: 55812348-55812348 on Build GRCh38
rs750435502 Chr.11: 55812401-55812401 on Build GRCh38
rs539145488 Chr.11: 55812417-55812417 on Build GRCh38
rs770995985 Chr.11: 55812199-55812199 on Build GRCh38
rs540821268 Chr.11: 55812646-55812647 on Build GRCh38
rs943038289 Chr.11: 55812611-55812611 on Build GRCh38
rs868127946 Chr.11: 55812330-55812330 on Build GRCh38
rs112105984 Chr.11: 55812341-55812341 on Build GRCh38
rs780997715 Chr.11: 55812422-55812422 on Build GRCh38
rs757419570 Chr.11: 55812271-55812271 on Build GRCh38
rs776890914 Chr.11: 55812386-55812386 on Build GRCh38
rs141628276 Chr.11: 55812278-55812278 on Build GRCh38
rs745494201 Chr.11: 55812451-55812451 on Build GRCh38
rs775946927 Chr.11: 55812416-55812417 on Build GRCh38
rs562202990 Chr.11: 55812204-55812204 on Build GRCh38
rs770569776 Chr.11: 55812369-55812369 on Build GRCh38
rs541042751 Chr.11: 55812292-55812292 on Build GRCh38

View Full Product Details