Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62129693-62130204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INCENP
Entrez Gene ID
3619
Gene Name
inner centromere protein
Gene Aliases
-
UniGene
Hs.142179
Gene Chromosome Location
Chr.11: 62123973-62155474 on Build GRCh38
Transcripts
NM_001040694.1 NM_020238.2 XM_006718533.2 XM_011544995.2 XM_011544996.2 XM_011544997.2 XM_011544998.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1475643 c.586C>T p.L196L Chr.11: 62130113-62130113 on Build GRCh38
3450832 c.295C>A p.R99R Chr.11: 62129822-62129822 on Build GRCh38
6502346 c.511A>G p.I171V Chr.11: 62130038-62130038 on Build GRCh38
6502347 c.511A>G p.I171V Chr.11: 62130038-62130038 on Build GRCh38
1561721 c.544C>G p.H182D Chr.11: 62130071-62130071 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34210125 Chr.11: 62129996-62129996 on Build GRCh38
rs777641583 Chr.11: 62130054-62130054 on Build GRCh38
rs748644987 Chr.11: 62129751-62129751 on Build GRCh38
rs770021592 Chr.11: 62130005-62130005 on Build GRCh38
rs755496748 Chr.11: 62129760-62129760 on Build GRCh38
rs775661936 Chr.11: 62130010-62130010 on Build GRCh38
rs199974792 Chr.11: 62130158-62130158 on Build GRCh38
rs75566338 Chr.11: 62129734-62129734 on Build GRCh38
rs756338232 Chr.11: 62129813-62129813 on Build GRCh38
rs201192132 Chr.11: 62130143-62130143 on Build GRCh38
rs772094693 Chr.11: 62129904-62129904 on Build GRCh38
rs763857838 Chr.11: 62130149-62130149 on Build GRCh38
rs186971860 Chr.11: 62130094-62130094 on Build GRCh38
rs753340298 Chr.11: 62130035-62130035 on Build GRCh38
rs751480395 Chr.11: 62129790-62129790 on Build GRCh38
rs988825357 Chr.11: 62129830-62129830 on Build GRCh38
rs747969371 Chr.11: 62129901-62129901 on Build GRCh38
rs368225096 Chr.11: 62129744-62129744 on Build GRCh38
rs535923655 Chr.11: 62129957-62129957 on Build GRCh38
rs1020354846 Chr.11: 62129976-62129976 on Build GRCh38
rs924358602 Chr.11: 62129889-62129889 on Build GRCh38
rs773367492 Chr.11: 62129866-62129866 on Build GRCh38
rs958526448 Chr.11: 62129998-62129998 on Build GRCh38
rs868550478 Chr.11: 62130018-62130018 on Build GRCh38
rs751356472 Chr.11: 62130150-62130150 on Build GRCh38
rs764020996 Chr.11: 62130126-62130131 on Build GRCh38
rs750872989 Chr.11: 62130121-62130121 on Build GRCh38
rs1012845150 Chr.11: 62130032-62130032 on Build GRCh38
rs533613179 Chr.11: 62130023-62130023 on Build GRCh38
rs768473115 Chr.11: 62129835-62129835 on Build GRCh38
rs779610174 Chr.11: 62129935-62129935 on Build GRCh38
rs1037223426 Chr.11: 62130111-62130111 on Build GRCh38
rs200416661 Chr.11: 62129820-62129820 on Build GRCh38
rs755587112 Chr.11: 62129891-62129891 on Build GRCh38
rs144643595 Chr.11: 62130087-62130087 on Build GRCh38
rs746941030 Chr.11: 62129977-62129977 on Build GRCh38
rs769445442 Chr.11: 62129782-62129782 on Build GRCh38
rs200221164 Chr.11: 62130014-62130014 on Build GRCh38
rs758789600 Chr.11: 62129926-62129926 on Build GRCh38
rs754415697 Chr.11: 62130040-62130040 on Build GRCh38
rs878970798 Chr.11: 62130085-62130085 on Build GRCh38
rs780043314 Chr.11: 62130163-62130163 on Build GRCh38
rs146595346 Chr.11: 62129832-62129832 on Build GRCh38
rs988879334 Chr.11: 62129968-62129968 on Build GRCh38
rs752382818 Chr.11: 62130049-62130049 on Build GRCh38
rs142399783 Chr.11: 62130108-62130108 on Build GRCh38
rs770781175 Chr.11: 62129985-62129985 on Build GRCh38
rs766470825 Chr.11: 62129916-62129916 on Build GRCh38
rs375511546 Chr.11: 62129894-62129894 on Build GRCh38
rs776717731 Chr.11: 62129995-62129995 on Build GRCh38
rs145875137 Chr.11: 62130070-62130070 on Build GRCh38
rs778533228 Chr.11: 62130041-62130041 on Build GRCh38
rs751989353 Chr.11: 62130022-62130022 on Build GRCh38
rs139869566 Chr.11: 62130120-62130120 on Build GRCh38
rs754070097 Chr.11: 62129878-62129878 on Build GRCh38
rs768996538 Chr.11: 62129750-62129750 on Build GRCh38
rs771971811 Chr.11: 62129964-62129964 on Build GRCh38
rs763895032 Chr.11: 62129789-62129789 on Build GRCh38
rs143235997 Chr.11: 62130146-62130146 on Build GRCh38
rs761804843 Chr.11: 62129844-62129844 on Build GRCh38
rs940260171 Chr.11: 62130117-62130117 on Build GRCh38
rs973238727 Chr.11: 62129836-62129836 on Build GRCh38
rs147033848 Chr.11: 62129924-62129924 on Build GRCh38
rs139054193 Chr.11: 62129801-62129801 on Build GRCh38
rs150267576 Chr.11: 62129869-62129869 on Build GRCh38
rs139863209 Chr.11: 62129825-62129825 on Build GRCh38
rs1027885646 Chr.11: 62129854-62129854 on Build GRCh38
rs894324824 Chr.11: 62130180-62130180 on Build GRCh38
rs145922935 Chr.11: 62130118-62130118 on Build GRCh38
rs562866408 Chr.11: 62129821-62129821 on Build GRCh38
rs774070841 Chr.11: 62129843-62129843 on Build GRCh38
rs565090054 Chr.11: 62130162-62130162 on Build GRCh38
rs757817115 Chr.11: 62129933-62129933 on Build GRCh38
rs758063143 Chr.11: 62130148-62130148 on Build GRCh38
rs781093733 Chr.11: 62130155-62130155 on Build GRCh38
rs774588060 Chr.11: 62130179-62130179 on Build GRCh38
rs774127794 Chr.11: 62129897-62129897 on Build GRCh38
rs373398703 Chr.11: 62129814-62129814 on Build GRCh38
rs143979460 Chr.11: 62129809-62129809 on Build GRCh38
rs778452765 Chr.11: 62129955-62129955 on Build GRCh38
rs140463949 Chr.11: 62129923-62129923 on Build GRCh38
rs756974750 Chr.11: 62130152-62130152 on Build GRCh38
rs112218336 Chr.11: 62129932-62129932 on Build GRCh38
rs754113313 Chr.11: 62129817-62129817 on Build GRCh38
rs1675133 Chr.11: 62129887-62129887 on Build GRCh38
rs888200527 Chr.11: 62129727-62129727 on Build GRCh38
rs759740436 Chr.11: 62129919-62129919 on Build GRCh38
rs369742634 Chr.11: 62129802-62129802 on Build GRCh38
rs768826171 Chr.11: 62129987-62129989 on Build GRCh38
rs146465225 Chr.11: 62129823-62129823 on Build GRCh38
rs764478695 Chr.11: 62129931-62129931 on Build GRCh38
rs201925711 Chr.11: 62130178-62130178 on Build GRCh38
rs34441559 Chr.11: 62129937-62129937 on Build GRCh38
rs909572833 Chr.11: 62130102-62130102 on Build GRCh38
rs1792944 Chr.11: 62129725-62129725 on Build GRCh38
rs767923801 Chr.11: 62130025-62130025 on Build GRCh38
rs949817471 Chr.11: 62129738-62129738 on Build GRCh38
rs775236514 Chr.11: 62129831-62129831 on Build GRCh38
rs151321132 Chr.11: 62129892-62129892 on Build GRCh38
rs554064911 Chr.11: 62129784-62129784 on Build GRCh38
rs773254306 Chr.11: 62129910-62129910 on Build GRCh38
rs776644769 Chr.11: 62129917-62129917 on Build GRCh38
rs753012492 Chr.11: 62129884-62129884 on Build GRCh38
rs539945924 Chr.11: 62129881-62129881 on Build GRCh38
rs746810359 Chr.11: 62130066-62130066 on Build GRCh38
rs768258892 Chr.11: 62129895-62129895 on Build GRCh38
rs775511796 Chr.11: 62129772-62129772 on Build GRCh38
rs367972890 Chr.11: 62129794-62129794 on Build GRCh38
rs770454641 Chr.11: 62129759-62129759 on Build GRCh38
rs766606953 Chr.11: 62129870-62129870 on Build GRCh38
rs758189633 Chr.11: 62130053-62130053 on Build GRCh38
rs765632070 Chr.11: 62129883-62129883 on Build GRCh38
rs202093810 Chr.11: 62130095-62130095 on Build GRCh38
rs749385174 Chr.11: 62130176-62130176 on Build GRCh38
rs760744776 Chr.11: 62129911-62129911 on Build GRCh38
rs113310761 Chr.11: 62129885-62129885 on Build GRCh38
rs1051299573 Chr.11: 62130109-62130109 on Build GRCh38
rs550573247 Chr.11: 62129767-62129767 on Build GRCh38
rs936152629 Chr.11: 62129779-62129779 on Build GRCh38
rs557768254 Chr.11: 62129737-62129737 on Build GRCh38
rs148478726 Chr.11: 62130107-62130107 on Build GRCh38
rs376029375 Chr.11: 62130137-62130137 on Build GRCh38
rs937590975 Chr.11: 62130064-62130064 on Build GRCh38
rs199515936 Chr.11: 62129846-62129846 on Build GRCh38
rs754600824 Chr.11: 62129949-62129949 on Build GRCh38
rs763214600 Chr.11: 62130012-62130012 on Build GRCh38

View Full Product Details