Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62140115-62140609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INCENP
Entrez Gene ID
3619
Gene Name
inner centromere protein
Gene Aliases
-
UniGene
Hs.142179
Gene Chromosome Location
Chr.11: 62123973-62155474 on Build GRCh38
Transcripts
NM_001040694.1 NM_020238.2 XM_006718533.2 XM_011544995.2 XM_011544996.2 XM_011544997.2 XM_011544998.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
348495 c.1303G>T p.D435Y Chr.11: 62140245-62140245 on Build GRCh38
6445673 c.1294G>A p.A432T Chr.11: 62140236-62140236 on Build GRCh38
6445674 c.1294G>A p.A432T Chr.11: 62140236-62140236 on Build GRCh38
5095657 c.1332G>A p.P444P Chr.11: 62140274-62140274 on Build GRCh38
5095658 c.1332G>A p.P444P Chr.11: 62140274-62140274 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs926846185 Chr.11: 62140178-62140178 on Build GRCh38
rs781470391 Chr.11: 62140315-62140315 on Build GRCh38
rs756586827 Chr.11: 62140337-62140337 on Build GRCh38
rs908695372 Chr.11: 62140441-62140441 on Build GRCh38
rs933493157 Chr.11: 62140535-62140535 on Build GRCh38
rs139833093 Chr.11: 62140235-62140235 on Build GRCh38
rs1009520322 Chr.11: 62140498-62140498 on Build GRCh38
rs781757062 Chr.11: 62140223-62140223 on Build GRCh38
rs144117865 Chr.11: 62140244-62140244 on Build GRCh38
rs1053589640 Chr.11: 62140267-62140267 on Build GRCh38
rs371320244 Chr.11: 62140253-62140253 on Build GRCh38
rs776616611 Chr.11: 62140270-62140270 on Build GRCh38
rs751902383 Chr.11: 62140218-62140218 on Build GRCh38
rs199897189 Chr.11: 62140243-62140243 on Build GRCh38
rs997796309 Chr.11: 62140173-62140173 on Build GRCh38
rs977397157 Chr.11: 62140458-62140458 on Build GRCh38
rs757649081 Chr.11: 62140191-62140191 on Build GRCh38
rs888551080 Chr.11: 62140565-62140565 on Build GRCh38
rs138752017 Chr.11: 62140274-62140274 on Build GRCh38
rs748925496 Chr.11: 62140495-62140495 on Build GRCh38
rs542989642 Chr.11: 62140208-62140208 on Build GRCh38
rs368242270 Chr.11: 62140295-62140295 on Build GRCh38
rs1016641345 Chr.11: 62140215-62140215 on Build GRCh38
rs1039241460 Chr.11: 62140162-62140162 on Build GRCh38
rs747770792 Chr.11: 62140237-62140237 on Build GRCh38
rs373715279 Chr.11: 62140190-62140190 on Build GRCh38
rs933117445 Chr.11: 62140225-62140225 on Build GRCh38
rs768203238 Chr.11: 62140227-62140227 on Build GRCh38
rs900660488 Chr.11: 62140169-62140169 on Build GRCh38
rs1025372396 Chr.11: 62140140-62140140 on Build GRCh38
rs372144906 Chr.11: 62140296-62140296 on Build GRCh38
rs76278113 Chr.11: 62140493-62140493 on Build GRCh38
rs12795611 Chr.11: 62140497-62140497 on Build GRCh38
rs922913034 Chr.11: 62140192-62140192 on Build GRCh38
rs61893679 Chr.11: 62140349-62140349 on Build GRCh38
rs761435367 Chr.11: 62140184-62140184 on Build GRCh38
rs753758134 Chr.11: 62140193-62140193 on Build GRCh38
rs200556008 Chr.11: 62140197-62140197 on Build GRCh38
rs560383309 Chr.11: 62140388-62140388 on Build GRCh38
rs758694120 Chr.11: 62140198-62140198 on Build GRCh38
rs961631013 Chr.11: 62140457-62140457 on Build GRCh38
rs750225003 Chr.11: 62140189-62140189 on Build GRCh38
rs376101208 Chr.11: 62140319-62140319 on Build GRCh38
rs757676613 Chr.11: 62140306-62140306 on Build GRCh38
rs201894726 Chr.11: 62140242-62140242 on Build GRCh38
rs767300024 Chr.11: 62140188-62140188 on Build GRCh38
rs771725238 Chr.11: 62140238-62140238 on Build GRCh38
rs1675124 Chr.11: 62140437-62140437 on Build GRCh38
rs758875598 Chr.11: 62140187-62140211 on Build GRCh38
rs376450952 Chr.11: 62140207-62140207 on Build GRCh38
rs548960989 Chr.11: 62140479-62140479 on Build GRCh38
rs761201421 Chr.11: 62140477-62140477 on Build GRCh38
rs1046742591 Chr.11: 62140501-62140501 on Build GRCh38
rs141138727 Chr.11: 62140236-62140236 on Build GRCh38
rs917847615 Chr.11: 62140358-62140358 on Build GRCh38
rs571516011 Chr.11: 62140151-62140151 on Build GRCh38
rs1002624669 Chr.11: 62140548-62140548 on Build GRCh38
rs952880027 Chr.11: 62140180-62140180 on Build GRCh38
rs187738543 Chr.11: 62140273-62140273 on Build GRCh38
rs61893680 Chr.11: 62140550-62140550 on Build GRCh38
rs746413273 Chr.11: 62140224-62140224 on Build GRCh38
rs922999424 Chr.11: 62140476-62140476 on Build GRCh38
rs3741246 Chr.11: 62140159-62140159 on Build GRCh38
rs534187060 Chr.11: 62140155-62140155 on Build GRCh38
rs952764503 Chr.11: 62140402-62140402 on Build GRCh38
rs540464199 Chr.11: 62140366-62140366 on Build GRCh38
rs778220996 Chr.11: 62140203-62140203 on Build GRCh38
rs560539126 Chr.11: 62140431-62140431 on Build GRCh38
rs778164005 Chr.11: 62140206-62140206 on Build GRCh38
rs751867700 Chr.11: 62140302-62140302 on Build GRCh38
rs906412556 Chr.11: 62140526-62140526 on Build GRCh38
rs76160431 Chr.11: 62140482-62140482 on Build GRCh38
rs551076181 Chr.11: 62140494-62140494 on Build GRCh38
rs765291422 Chr.11: 62140195-62140195 on Build GRCh38

View Full Product Details