Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62752745-62753253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB3
Entrez Gene ID
79842
Gene Name
zinc finger and BTB domain containing 3
Gene Aliases
-
UniGene
Hs.147554
Gene Chromosome Location
Chr.11: 62750963-62754184 on Build GRCh38
Transcripts
NM_024784.3 XM_011545256.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4742121 c.986C>T p.T329M Chr.11: 62752829-62752829 on Build GRCh38
4855544 c.701C>T p.P234L Chr.11: 62753114-62753114 on Build GRCh38
6231417 c.989C>T p.S330L Chr.11: 62752826-62752826 on Build GRCh38
4971794 c.839C>T p.P280L Chr.11: 62752976-62752976 on Build GRCh38
2039610 c.771G>A p.L257L Chr.11: 62753044-62753044 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765978918 Chr.11: 62752776-62752776 on Build GRCh38
rs201503930 Chr.11: 62753066-62753066 on Build GRCh38
rs749778744 Chr.11: 62753040-62753040 on Build GRCh38
rs770425058 Chr.11: 62753055-62753055 on Build GRCh38
rs146723418 Chr.11: 62752836-62752836 on Build GRCh38
rs780439565 Chr.11: 62753013-62753013 on Build GRCh38
rs781620902 Chr.11: 62752803-62752803 on Build GRCh38
rs769507305 Chr.11: 62753143-62753143 on Build GRCh38
rs902683697 Chr.11: 62753039-62753039 on Build GRCh38
rs139609670 Chr.11: 62752894-62752894 on Build GRCh38
rs143334355 Chr.11: 62752871-62752871 on Build GRCh38
rs867206156 Chr.11: 62752874-62752874 on Build GRCh38
rs774485308 Chr.11: 62753159-62753159 on Build GRCh38
rs779542676 Chr.11: 62753120-62753120 on Build GRCh38
rs768328499 Chr.11: 62753157-62753157 on Build GRCh38
rs750718381 Chr.11: 62753069-62753069 on Build GRCh38
rs754245746 Chr.11: 62753106-62753106 on Build GRCh38
rs779645199 Chr.11: 62753130-62753131 on Build GRCh38
rs368974485 Chr.11: 62752829-62752829 on Build GRCh38
rs774087313 Chr.11: 62753063-62753063 on Build GRCh38
rs750665396 Chr.11: 62752977-62752977 on Build GRCh38
rs769163765 Chr.11: 62753072-62753072 on Build GRCh38
rs746529059 Chr.11: 62753052-62753052 on Build GRCh38
rs147104405 Chr.11: 62752891-62752891 on Build GRCh38
rs1054430066 Chr.11: 62753172-62753172 on Build GRCh38
rs750216109 Chr.11: 62752813-62752814 on Build GRCh38
rs144306155 Chr.11: 62753042-62753042 on Build GRCh38
rs752626270 Chr.11: 62752935-62752935 on Build GRCh38
rs754004878 Chr.11: 62753212-62753212 on Build GRCh38
rs763897027 Chr.11: 62752955-62752955 on Build GRCh38
rs962794613 Chr.11: 62752823-62752823 on Build GRCh38
rs755232612 Chr.11: 62753107-62753107 on Build GRCh38
rs749785047 Chr.11: 62752862-62752862 on Build GRCh38
rs766466251 Chr.11: 62753099-62753099 on Build GRCh38
rs201044020 Chr.11: 62753196-62753196 on Build GRCh38
rs756440317 Chr.11: 62753093-62753093 on Build GRCh38
rs780648775 Chr.11: 62753135-62753135 on Build GRCh38
rs946462840 Chr.11: 62753031-62753031 on Build GRCh38
rs756600074 Chr.11: 62752994-62752994 on Build GRCh38
rs753657767 Chr.11: 62752783-62752783 on Build GRCh38
rs139216130 Chr.11: 62752848-62752848 on Build GRCh38
rs200424392 Chr.11: 62752818-62752818 on Build GRCh38
rs745340741 Chr.11: 62753136-62753136 on Build GRCh38
rs151167288 Chr.11: 62752789-62752789 on Build GRCh38
rs757352959 Chr.11: 62752967-62752967 on Build GRCh38
rs141062837 Chr.11: 62752835-62752835 on Build GRCh38
rs148738930 Chr.11: 62753056-62753056 on Build GRCh38
rs141773652 Chr.11: 62753125-62753125 on Build GRCh38
rs775935149 Chr.11: 62753012-62753012 on Build GRCh38
rs377221715 Chr.11: 62753214-62753214 on Build GRCh38
rs921269126 Chr.11: 62753188-62753188 on Build GRCh38
rs201620712 Chr.11: 62752912-62752912 on Build GRCh38
rs748894189 Chr.11: 62753176-62753178 on Build GRCh38
rs746083425 Chr.11: 62752811-62752811 on Build GRCh38
rs773451288 Chr.11: 62753197-62753197 on Build GRCh38
rs749111137 Chr.11: 62753152-62753152 on Build GRCh38
rs1033770983 Chr.11: 62753014-62753014 on Build GRCh38
rs761816179 Chr.11: 62753165-62753165 on Build GRCh38
rs891332161 Chr.11: 62752924-62752924 on Build GRCh38
rs200684994 Chr.11: 62752958-62752958 on Build GRCh38
rs146249613 Chr.11: 62753145-62753145 on Build GRCh38
rs531310587 Chr.11: 62753037-62753037 on Build GRCh38
rs772813030 Chr.11: 62752886-62752886 on Build GRCh38
rs558456792 Chr.11: 62752946-62752946 on Build GRCh38
rs749580086 Chr.11: 62753028-62753028 on Build GRCh38
rs766630917 Chr.11: 62753202-62753202 on Build GRCh38
rs755429831 Chr.11: 62753229-62753229 on Build GRCh38
rs1012437341 Chr.11: 62753100-62753100 on Build GRCh38
rs185887296 Chr.11: 62752929-62752929 on Build GRCh38
rs781566683 Chr.11: 62752975-62752975 on Build GRCh38
rs764104414 Chr.11: 62753060-62753060 on Build GRCh38
rs755809742 Chr.11: 62752970-62752970 on Build GRCh38
rs765164533 Chr.11: 62752810-62752810 on Build GRCh38
rs757582679 Chr.11: 62752834-62752834 on Build GRCh38
rs571308707 Chr.11: 62753121-62753121 on Build GRCh38
rs1057028474 Chr.11: 62753091-62753091 on Build GRCh38
rs760781621 Chr.11: 62753198-62753198 on Build GRCh38
rs879236812 Chr.11: 62753035-62753035 on Build GRCh38
rs149562336 Chr.11: 62752938-62752938 on Build GRCh38
rs376259634 Chr.11: 62752931-62752931 on Build GRCh38
rs372985423 Chr.11: 62752830-62752830 on Build GRCh38
rs762606036 Chr.11: 62753057-62753057 on Build GRCh38
rs776200589 Chr.11: 62752920-62752920 on Build GRCh38
rs200019468 Chr.11: 62753067-62753067 on Build GRCh38
rs1042105483 Chr.11: 62753036-62753036 on Build GRCh38

View Full Product Details