Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62753558-62754063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB3
Entrez Gene ID
79842
Gene Name
zinc finger and BTB domain containing 3
Gene Aliases
-
UniGene
Hs.147554
Gene Chromosome Location
Chr.11: 62750963-62754184 on Build GRCh38
Transcripts
NM_024784.3 XM_011545256.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6194311 c.237G>A p.M79I Chr.11: 62753578-62753578 on Build GRCh38
1355459 c.74_76delTTC p.L25delL Chr.11: 62753987-62753989 on Build GRCh38
6194312 c.198G>A p.E66E Chr.11: 62753617-62753617 on Build GRCh38
248409 c.12G>A p.E4E Chr.11: 62754051-62754051 on Build GRCh38
5503749 c.99+1G>T p.? Chr.11: 62753963-62753963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150997515 Chr.11: 62753612-62753612 on Build GRCh38
rs912620498 Chr.11: 62753771-62753771 on Build GRCh38
rs780594017 Chr.11: 62753964-62753964 on Build GRCh38
rs567879465 Chr.11: 62753991-62753991 on Build GRCh38
rs527387724 Chr.11: 62753970-62753970 on Build GRCh38
rs761235515 Chr.11: 62753948-62753948 on Build GRCh38
rs977868059 Chr.11: 62753646-62753647 on Build GRCh38
rs964507037 Chr.11: 62753769-62753769 on Build GRCh38
rs780075545 Chr.11: 62753935-62753935 on Build GRCh38
rs199635331 Chr.11: 62753686-62753686 on Build GRCh38
rs144295270 Chr.11: 62753756-62753757 on Build GRCh38
rs912569772 Chr.11: 62753963-62753963 on Build GRCh38
rs748530624 Chr.11: 62753942-62753942 on Build GRCh38
rs200245732 Chr.11: 62753688-62753688 on Build GRCh38
rs201321255 Chr.11: 62753672-62753672 on Build GRCh38
rs753306811 Chr.11: 62753621-62753621 on Build GRCh38
rs767917747 Chr.11: 62753741-62753741 on Build GRCh38
rs774607363 Chr.11: 62753722-62753722 on Build GRCh38
rs370899793 Chr.11: 62753939-62753939 on Build GRCh38
rs772365078 Chr.11: 62753987-62753987 on Build GRCh38
rs766373985 Chr.11: 62754012-62754012 on Build GRCh38
rs891239476 Chr.11: 62753730-62753730 on Build GRCh38
rs1024611468 Chr.11: 62753907-62753907 on Build GRCh38
rs199665529 Chr.11: 62753966-62753966 on Build GRCh38
rs1022261606 Chr.11: 62753778-62753778 on Build GRCh38
rs751325139 Chr.11: 62753918-62753918 on Build GRCh38
rs763754112 Chr.11: 62753917-62753917 on Build GRCh38
rs372020438 Chr.11: 62754030-62754030 on Build GRCh38
rs781056417 Chr.11: 62753920-62753920 on Build GRCh38
rs778085774 Chr.11: 62754035-62754035 on Build GRCh38
rs894796306 Chr.11: 62753871-62753871 on Build GRCh38
rs774570253 Chr.11: 62753941-62753941 on Build GRCh38
rs371811417 Chr.11: 62753919-62753919 on Build GRCh38
rs773923711 Chr.11: 62754006-62754006 on Build GRCh38
rs902239418 Chr.11: 62753911-62753911 on Build GRCh38
rs375687324 Chr.11: 62753738-62753738 on Build GRCh38
rs923586596 Chr.11: 62753810-62753810 on Build GRCh38
rs775163886 Chr.11: 62753957-62753957 on Build GRCh38
rs1047080294 Chr.11: 62753880-62753880 on Build GRCh38
rs960508657 Chr.11: 62753819-62753819 on Build GRCh38
rs777210321 Chr.11: 62753994-62753994 on Build GRCh38
rs760402045 Chr.11: 62754011-62754011 on Build GRCh38
rs930206589 Chr.11: 62753877-62753877 on Build GRCh38
rs754473707 Chr.11: 62753646-62753646 on Build GRCh38
rs747380700 Chr.11: 62754040-62754040 on Build GRCh38
rs758671866 Chr.11: 62754034-62754034 on Build GRCh38
rs769044932 Chr.11: 62753705-62753705 on Build GRCh38
rs752563220 Chr.11: 62753875-62753875 on Build GRCh38
rs191848051 Chr.11: 62753788-62753788 on Build GRCh38
rs898697786 Chr.11: 62753826-62753826 on Build GRCh38
rs779914302 Chr.11: 62753971-62753971 on Build GRCh38
rs375475738 Chr.11: 62753921-62753921 on Build GRCh38
rs931573912 Chr.11: 62753831-62753831 on Build GRCh38
rs759177624 Chr.11: 62753745-62753745 on Build GRCh38
rs541126059 Chr.11: 62753807-62753807 on Build GRCh38
rs779021393 Chr.11: 62754025-62754025 on Build GRCh38
rs999235274 Chr.11: 62753926-62753926 on Build GRCh38
rs547151576 Chr.11: 62753981-62753981 on Build GRCh38
rs777680291 Chr.11: 62753668-62753668 on Build GRCh38
rs746780003 Chr.11: 62753669-62753669 on Build GRCh38
rs184074297 Chr.11: 62753898-62753898 on Build GRCh38
rs572513 Chr.11: 62753784-62753784 on Build GRCh38
rs747539245 Chr.11: 62753914-62753915 on Build GRCh38
rs948182260 Chr.11: 62753839-62753839 on Build GRCh38
rs145527809 Chr.11: 62754015-62754015 on Build GRCh38
rs957943133 Chr.11: 62753676-62753676 on Build GRCh38
rs149727586 Chr.11: 62753667-62753667 on Build GRCh38
rs573726329 Chr.11: 62753870-62753870 on Build GRCh38
rs553947129 Chr.11: 62753828-62753828 on Build GRCh38
rs975107398 Chr.11: 62753684-62753684 on Build GRCh38
rs372060210 Chr.11: 62753702-62753702 on Build GRCh38
rs762618923 Chr.11: 62754000-62754000 on Build GRCh38
rs373071045 Chr.11: 62753794-62753794 on Build GRCh38
rs752154756 Chr.11: 62753617-62753617 on Build GRCh38
rs72929471 Chr.11: 62754003-62754003 on Build GRCh38
rs368319142 Chr.11: 62753959-62753959 on Build GRCh38
rs796601139 Chr.11: 62753914-62753914 on Build GRCh38
rs374187830 Chr.11: 62753924-62753924 on Build GRCh38
rs72929469 Chr.11: 62753936-62753936 on Build GRCh38
rs992292377 Chr.11: 62753823-62753823 on Build GRCh38
rs752386537 Chr.11: 62753655-62753655 on Build GRCh38
rs1022125825 Chr.11: 62753955-62753955 on Build GRCh38
rs374640664 Chr.11: 62753946-62753946 on Build GRCh38
rs775620877 Chr.11: 62753583-62753583 on Build GRCh38
rs899916826 Chr.11: 62753937-62753937 on Build GRCh38
rs750237316 Chr.11: 62754010-62754010 on Build GRCh38
rs369536933 Chr.11: 62753746-62753746 on Build GRCh38
rs778778670 Chr.11: 62753647-62753647 on Build GRCh38
rs755066689 Chr.11: 62754024-62754024 on Build GRCh38
rs781126365 Chr.11: 62753679-62753679 on Build GRCh38
rs755086140 Chr.11: 62753978-62753978 on Build GRCh38
rs1024704009 Chr.11: 62753608-62753608 on Build GRCh38
rs753923242 Chr.11: 62753974-62753974 on Build GRCh38
rs773767062 Chr.11: 62753743-62753743 on Build GRCh38
rs758156730 Chr.11: 62753764-62753764 on Build GRCh38
rs542970220 Chr.11: 62753874-62753874 on Build GRCh38
rs562754719 Chr.11: 62753889-62753889 on Build GRCh38
rs767035884 Chr.11: 62754008-62754008 on Build GRCh38
rs996098604 Chr.11: 62753772-62753772 on Build GRCh38
rs187328752 Chr.11: 62753993-62753993 on Build GRCh38
rs200771004 Chr.11: 62753739-62753739 on Build GRCh38
rs749713844 Chr.11: 62753965-62753965 on Build GRCh38
rs765267379 Chr.11: 62753846-62753846 on Build GRCh38
rs762191684 Chr.11: 62753619-62753619 on Build GRCh38
rs773641928 Chr.11: 62753947-62753947 on Build GRCh38
rs763373327 Chr.11: 62753604-62753604 on Build GRCh38
rs201229122 Chr.11: 62754003-62754004 on Build GRCh38
rs752439902 Chr.11: 62753754-62753754 on Build GRCh38
rs766953235 Chr.11: 62753954-62753954 on Build GRCh38

View Full Product Details