Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 43596138-43596537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT52
Entrez Gene ID
51098
Gene Name
intraflagellar transport 52
Gene Aliases
C20orf9, CGI-53, NGD2, NGD5
UniGene
Hs.444332
Gene Chromosome Location
Chr.20: 43590613-43647296 on Build GRCh38
Transcripts
NM_001303458.2 NM_001303459.2 NM_001323578.1 NM_001323579.1 NM_001323580.1 NM_001323581.1 NM_016004.4 XM_017027863.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6558786 c.180G>T p.G60G Chr.20: 43596495-43596495 on Build GRCh38
1026989 c.144G>T p.E48D Chr.20: 43596459-43596459 on Build GRCh38
6525836 c.144G>A p.E48E Chr.20: 43596459-43596459 on Build GRCh38
3707780 c.174A>G p.T58T Chr.20: 43596489-43596489 on Build GRCh38
5034470 c.120-4C>A p.? Chr.20: 43596431-43596431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs73907861 Chr.20: 43596255-43596255 on Build GRCh38
rs111861455 Chr.20: 43596358-43596358 on Build GRCh38
rs954469196 Chr.20: 43596227-43596227 on Build GRCh38
rs754067267 Chr.20: 43596450-43596450 on Build GRCh38
rs763277945 Chr.20: 43596247-43596247 on Build GRCh38
rs965554005 Chr.20: 43596213-43596213 on Build GRCh38
rs777934756 Chr.20: 43596447-43596447 on Build GRCh38
rs980184094 Chr.20: 43596379-43596379 on Build GRCh38
rs376715173 Chr.20: 43596503-43596503 on Build GRCh38
rs112680199 Chr.20: 43596364-43596364 on Build GRCh38
rs200481999 Chr.20: 43596425-43596425 on Build GRCh38
rs897739238 Chr.20: 43596423-43596423 on Build GRCh38
rs372543072 Chr.20: 43596369-43596369 on Build GRCh38
rs948783639 Chr.20: 43596374-43596374 on Build GRCh38
rs755929073 Chr.20: 43596436-43596436 on Build GRCh38
rs567767600 Chr.20: 43596245-43596245 on Build GRCh38
rs779105186 Chr.20: 43596470-43596470 on Build GRCh38
rs576440784 Chr.20: 43596300-43596300 on Build GRCh38
rs917072987 Chr.20: 43596294-43596294 on Build GRCh38
rs191549432 Chr.20: 43596309-43596309 on Build GRCh38
rs948558601 Chr.20: 43596311-43596311 on Build GRCh38
rs956146734 Chr.20: 43596290-43596290 on Build GRCh38
rs897521393 Chr.20: 43596188-43596188 on Build GRCh38
rs200191740 Chr.20: 43596269-43596269 on Build GRCh38
rs904712571 Chr.20: 43596504-43596504 on Build GRCh38
rs992817798 Chr.20: 43596360-43596360 on Build GRCh38
rs2664519 Chr.20: 43596474-43596474 on Build GRCh38
rs765928775 Chr.20: 43596400-43596400 on Build GRCh38
rs543756807 Chr.20: 43596304-43596304 on Build GRCh38
rs73272200 Chr.20: 43596189-43596189 on Build GRCh38
rs762328621 Chr.20: 43596390-43596390 on Build GRCh38
rs201225510 Chr.20: 43596435-43596435 on Build GRCh38
rs1048879948 Chr.20: 43596377-43596377 on Build GRCh38
rs992792997 Chr.20: 43596291-43596291 on Build GRCh38
rs1031909396 Chr.20: 43596338-43596338 on Build GRCh38
rs759585842 Chr.20: 43596426-43596426 on Build GRCh38
rs996990832 Chr.20: 43596453-43596453 on Build GRCh38
rs373292042 Chr.20: 43596432-43596432 on Build GRCh38
rs137979762 Chr.20: 43596472-43596472 on Build GRCh38
rs968885488 Chr.20: 43596307-43596307 on Build GRCh38
rs569999809 Chr.20: 43596316-43596316 on Build GRCh38
rs926479809 Chr.20: 43596385-43596385 on Build GRCh38
rs978839498 Chr.20: 43596249-43596249 on Build GRCh38
rs757223938 Chr.20: 43596462-43596462 on Build GRCh38
rs1028054998 Chr.20: 43596246-43596246 on Build GRCh38
rs752501152 Chr.20: 43596302-43596302 on Build GRCh38

View Full Product Details