Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 117203195-117203688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAGLN
Entrez Gene ID
6876
Gene Name
transgelin
Gene Aliases
SM22, SMCC, TAGLN1, WS3-10
UniGene
Hs.410977
Gene Chromosome Location
Chr.11: 117199324-117204792 on Build GRCh38
Transcripts
NM_001001522.1 NM_003186.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
923416 c.203G>T p.S68I Chr.11: 117203329-117203329 on Build GRCh38
6274195 c.307G>T p.D103Y Chr.11: 117203433-117203433 on Build GRCh38
923417 c.252C>T p.P84P Chr.11: 117203378-117203378 on Build GRCh38
6279103 c.248C>A p.P83Q Chr.11: 117203374-117203374 on Build GRCh38
923418 c.322A>G p.K108E Chr.11: 117203448-117203448 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757769043 Chr.11: 117203333-117203333 on Build GRCh38
rs765910054 Chr.11: 117203289-117203289 on Build GRCh38
rs527772623 Chr.11: 117203488-117203488 on Build GRCh38
rs549583527 Chr.11: 117203612-117203612 on Build GRCh38
rs748255025 Chr.11: 117203320-117203322 on Build GRCh38
rs780165958 Chr.11: 117203649-117203649 on Build GRCh38
rs369057227 Chr.11: 117203283-117203283 on Build GRCh38
rs199983006 Chr.11: 117203217-117203217 on Build GRCh38
rs370629366 Chr.11: 117203412-117203412 on Build GRCh38
rs761740387 Chr.11: 117203260-117203260 on Build GRCh38
rs1011566695 Chr.11: 117203262-117203262 on Build GRCh38
rs904446523 Chr.11: 117203538-117203538 on Build GRCh38
rs538574197 Chr.11: 117203624-117203624 on Build GRCh38
rs914658829 Chr.11: 117203599-117203599 on Build GRCh38
rs759010399 Chr.11: 117203367-117203367 on Build GRCh38
rs750902284 Chr.11: 117203509-117203509 on Build GRCh38
rs140957009 Chr.11: 117203629-117203629 on Build GRCh38
rs747051308 Chr.11: 117203234-117203235 on Build GRCh38
rs641620 Chr.11: 117203513-117203513 on Build GRCh38
rs765717235 Chr.11: 117203507-117203507 on Build GRCh38
rs369308744 Chr.11: 117203366-117203366 on Build GRCh38
rs756946116 Chr.11: 117203526-117203526 on Build GRCh38
rs12970 Chr.11: 117203393-117203393 on Build GRCh38
rs781232698 Chr.11: 117203522-117203522 on Build GRCh38
rs752382132 Chr.11: 117203303-117203303 on Build GRCh38
rs918258475 Chr.11: 117203457-117203457 on Build GRCh38
rs762937184 Chr.11: 117203364-117203364 on Build GRCh38
rs529660606 Chr.11: 117203570-117203570 on Build GRCh38
rs747255641 Chr.11: 117203354-117203354 on Build GRCh38
rs775044094 Chr.11: 117203466-117203466 on Build GRCh38
rs755058563 Chr.11: 117203243-117203244 on Build GRCh38
rs770072244 Chr.11: 117203443-117203445 on Build GRCh38
rs139024720 Chr.11: 117203339-117203339 on Build GRCh38
rs370652740 Chr.11: 117203347-117203347 on Build GRCh38
rs559775983 Chr.11: 117203638-117203638 on Build GRCh38
rs772477046 Chr.11: 117203239-117203239 on Build GRCh38
rs780383343 Chr.11: 117203353-117203353 on Build GRCh38
rs903187169 Chr.11: 117203617-117203617 on Build GRCh38
rs373440137 Chr.11: 117203444-117203444 on Build GRCh38
rs947487918 Chr.11: 117203604-117203604 on Build GRCh38
rs180986151 Chr.11: 117203369-117203369 on Build GRCh38
rs773519940 Chr.11: 117203526-117203527 on Build GRCh38
rs762042572 Chr.11: 117203499-117203499 on Build GRCh38
rs751128158 Chr.11: 117203294-117203294 on Build GRCh38
rs11547375 Chr.11: 117203365-117203365 on Build GRCh38
rs746084466 Chr.11: 117203632-117203632 on Build GRCh38
rs779835947 Chr.11: 117203232-117203232 on Build GRCh38
rs1014725928 Chr.11: 117203608-117203608 on Build GRCh38
rs372976677 Chr.11: 117203296-117203296 on Build GRCh38
rs747231223 Chr.11: 117203406-117203406 on Build GRCh38
rs772948498 Chr.11: 117203363-117203363 on Build GRCh38
rs754881284 Chr.11: 117203505-117203507 on Build GRCh38
rs112663706 Chr.11: 117203304-117203304 on Build GRCh38
rs766553946 Chr.11: 117203514-117203514 on Build GRCh38
rs112444683 Chr.11: 117203662-117203662 on Build GRCh38
rs61905529 Chr.11: 117203575-117203575 on Build GRCh38
rs762805297 Chr.11: 117203284-117203284 on Build GRCh38
rs768819001 Chr.11: 117203410-117203410 on Build GRCh38
rs776541730 Chr.11: 117203259-117203259 on Build GRCh38
rs774295976 Chr.11: 117203218-117203218 on Build GRCh38
rs780293624 Chr.11: 117203405-117203405 on Build GRCh38
rs201871529 Chr.11: 117203242-117203242 on Build GRCh38
rs749078222 Chr.11: 117203425-117203425 on Build GRCh38
rs112388114 Chr.11: 117203486-117203486 on Build GRCh38
rs771508133 Chr.11: 117203464-117203464 on Build GRCh38
rs780168789 Chr.11: 117203218-117203225 on Build GRCh38
rs754301137 Chr.11: 117203331-117203331 on Build GRCh38
rs781477268 Chr.11: 117203306-117203314 on Build GRCh38
rs36112166 Chr.11: 117203355-117203355 on Build GRCh38
rs770794053 Chr.11: 117203426-117203426 on Build GRCh38
rs763700396 Chr.11: 117203470-117203470 on Build GRCh38
rs781226472 Chr.11: 117203628-117203628 on Build GRCh38
rs774226652 Chr.11: 117203440-117203440 on Build GRCh38
rs574544173 Chr.11: 117203225-117203225 on Build GRCh38
rs145148872 Chr.11: 117203334-117203334 on Build GRCh38
rs2269397 Chr.11: 117203583-117203583 on Build GRCh38
rs778015450 Chr.11: 117203312-117203312 on Build GRCh38
rs145801025 Chr.11: 117203381-117203381 on Build GRCh38
rs751640529 Chr.11: 117203515-117203515 on Build GRCh38
rs752292029 Chr.11: 117203376-117203376 on Build GRCh38
rs565311785 Chr.11: 117203469-117203469 on Build GRCh38
rs112912944 Chr.11: 117203305-117203305 on Build GRCh38
rs201511147 Chr.11: 117203374-117203374 on Build GRCh38
rs756423771 Chr.11: 117203308-117203308 on Build GRCh38
rs754211410 Chr.11: 117203382-117203382 on Build GRCh38
rs770988312 Chr.11: 117203214-117203214 on Build GRCh38
rs751587553 Chr.11: 117203661-117203661 on Build GRCh38
rs748315637 Chr.11: 117203596-117203596 on Build GRCh38
rs764512264 Chr.11: 117203377-117203377 on Build GRCh38
rs186542186 Chr.11: 117203623-117203623 on Build GRCh38
rs758500974 Chr.11: 117203398-117203398 on Build GRCh38
rs111721331 Chr.11: 117203636-117203636 on Build GRCh38
rs565629794 Chr.11: 117203548-117203548 on Build GRCh38
rs755198671 Chr.11: 117203516-117203516 on Build GRCh38
rs769505828 Chr.11: 117203280-117203280 on Build GRCh38
rs148113093 Chr.11: 117203385-117203385 on Build GRCh38

View Full Product Details