Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118094724-118095123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMPRSS4
Entrez Gene ID
56649
Gene Name
transmembrane protease, serine 4
Gene Aliases
CAPH2, MT-SP2, TMPRSS3
UniGene
Hs.161985
Gene Chromosome Location
Chr.11: 118077012-118125505 on Build GRCh38
Transcripts
NM_001083947.1 NM_001173551.1 NM_001173552.1 NM_001290094.1 NM_001290096.1 NM_019894.3 NR_110734.1 XM_005271613.3 XM_005271614.2 XM_005271615.3 XM_011542901.2 XM_011542902.2 XM_011542903.2 XM_011542904.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3666111 c.8A>G p.Q3R Chr.11: 118094820-118094820 on Build GRCh38
6804641 c.4-2A>G p.? Chr.11: 118094814-118094814 on Build GRCh38
3752161 c.4-1A>G p.? Chr.11: 118094815-118094815 on Build GRCh38
1720721 c.10G>A p.D4N Chr.11: 118094822-118094822 on Build GRCh38
303417 c.43G>T p.D15Y Chr.11: 118094855-118094855 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764731333 Chr.11: 118094993-118094993 on Build GRCh38
rs989760562 Chr.11: 118094851-118094851 on Build GRCh38
rs771586728 Chr.11: 118094913-118094913 on Build GRCh38
rs146209120 Chr.11: 118094991-118094991 on Build GRCh38
rs749219936 Chr.11: 118094905-118094905 on Build GRCh38
rs2276125 Chr.11: 118094931-118094931 on Build GRCh38
rs753768994 Chr.11: 118094973-118094973 on Build GRCh38
rs772733499 Chr.11: 118094826-118094826 on Build GRCh38
rs201884421 Chr.11: 118094843-118094843 on Build GRCh38
rs778042969 Chr.11: 118094797-118094797 on Build GRCh38
rs375515074 Chr.11: 118094810-118094810 on Build GRCh38
rs770576090 Chr.11: 118094830-118094830 on Build GRCh38
rs761279517 Chr.11: 118094859-118094859 on Build GRCh38
rs757564265 Chr.11: 118094808-118094808 on Build GRCh38
rs747847490 Chr.11: 118094884-118094884 on Build GRCh38
rs770419857 Chr.11: 118094840-118094840 on Build GRCh38
rs200645289 Chr.11: 118094822-118094822 on Build GRCh38
rs762393761 Chr.11: 118094829-118094829 on Build GRCh38
rs753916847 Chr.11: 118094803-118094803 on Build GRCh38
rs767843983 Chr.11: 118094783-118094783 on Build GRCh38
rs914910448 Chr.11: 118095021-118095021 on Build GRCh38
rs978302103 Chr.11: 118095069-118095069 on Build GRCh38
rs1021477475 Chr.11: 118094959-118094959 on Build GRCh38
rs989512491 Chr.11: 118095020-118095020 on Build GRCh38
rs550415590 Chr.11: 118095093-118095093 on Build GRCh38
rs997678093 Chr.11: 118094917-118094917 on Build GRCh38
rs753970094 Chr.11: 118094866-118094866 on Build GRCh38
rs759101181 Chr.11: 118094839-118094839 on Build GRCh38
rs371804331 Chr.11: 118094798-118094798 on Build GRCh38
rs752975925 Chr.11: 118094854-118094854 on Build GRCh38
rs2276124 Chr.11: 118094887-118094887 on Build GRCh38
rs2276122 Chr.11: 118094815-118094815 on Build GRCh38
rs773715544 Chr.11: 118094909-118094909 on Build GRCh38
rs767978290 Chr.11: 118094846-118094846 on Build GRCh38
rs764002925 Chr.11: 118094953-118094953 on Build GRCh38
rs964417033 Chr.11: 118095015-118095015 on Build GRCh38
rs745576297 Chr.11: 118094910-118094910 on Build GRCh38
rs968622584 Chr.11: 118094970-118094970 on Build GRCh38
rs939467890 Chr.11: 118094889-118094889 on Build GRCh38
rs544153597 Chr.11: 118095084-118095084 on Build GRCh38
rs368070089 Chr.11: 118094882-118094882 on Build GRCh38
rs55861358 Chr.11: 118094906-118094906 on Build GRCh38
rs1026179109 Chr.11: 118094989-118094989 on Build GRCh38
rs756559920 Chr.11: 118094785-118094785 on Build GRCh38
rs71482127 Chr.11: 118094920-118094920 on Build GRCh38
rs137961034 Chr.11: 118095022-118095022 on Build GRCh38
rs765390822 Chr.11: 118094870-118094870 on Build GRCh38
rs2276126 Chr.11: 118095068-118095068 on Build GRCh38
rs895171178 Chr.11: 118094912-118094912 on Build GRCh38
rs76960081 Chr.11: 118094971-118094971 on Build GRCh38
rs767772092 Chr.11: 118094774-118094774 on Build GRCh38
rs2276123 Chr.11: 118094880-118094880 on Build GRCh38
rs148199783 Chr.11: 118094779-118094779 on Build GRCh38
rs774057352 Chr.11: 118094836-118094836 on Build GRCh38
rs537725599 Chr.11: 118095065-118095065 on Build GRCh38
rs777623336 Chr.11: 118094895-118094895 on Build GRCh38
rs200168548 Chr.11: 118094864-118094864 on Build GRCh38
rs993418667 Chr.11: 118094975-118094975 on Build GRCh38
rs953204169 Chr.11: 118094939-118094939 on Build GRCh38
rs757475601 Chr.11: 118094869-118094869 on Build GRCh38
rs536458128 Chr.11: 118094992-118094992 on Build GRCh38
rs369297379 Chr.11: 118094881-118094881 on Build GRCh38

View Full Product Details