Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118162720-118163229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCN2B
Entrez Gene ID
6327
Gene Name
sodium voltage-gated channel beta subunit 2
Gene Aliases
ATFB14
UniGene
Hs.129783
Gene Chromosome Location
Chr.11: 118162804-118176622 on Build GRCh38
Transcripts
NM_004588.4

SNP Detail

SNP ID Chromosome Location
rs76607319 Chr.11: 118162903-118162903 on Build GRCh38
rs910488809 Chr.11: 118162813-118162813 on Build GRCh38
rs559793938 Chr.11: 118163072-118163072 on Build GRCh38
rs1005833584 Chr.11: 118162952-118162952 on Build GRCh38
rs757138525 Chr.11: 118162884-118162884 on Build GRCh38
rs115357119 Chr.11: 118162845-118162845 on Build GRCh38
rs915655287 Chr.11: 118163004-118163004 on Build GRCh38
rs1007607903 Chr.11: 118163171-118163172 on Build GRCh38
rs922640154 Chr.11: 118162795-118162795 on Build GRCh38
rs625464 Chr.11: 118162830-118162830 on Build GRCh38
rs778826251 Chr.11: 118162929-118162929 on Build GRCh38
rs1051755357 Chr.11: 118163102-118163102 on Build GRCh38
rs191690025 Chr.11: 118163028-118163028 on Build GRCh38
rs1031311694 Chr.11: 118162947-118162947 on Build GRCh38
rs1042125439 Chr.11: 118162891-118162891 on Build GRCh38
rs972862403 Chr.11: 118163027-118163027 on Build GRCh38
rs539755265 Chr.11: 118162831-118162831 on Build GRCh38
rs910078804 Chr.11: 118162748-118162748 on Build GRCh38
rs537466784 Chr.11: 118162968-118162968 on Build GRCh38
rs767021987 Chr.11: 118162772-118162772 on Build GRCh38
rs608176 Chr.11: 118163074-118163074 on Build GRCh38
rs186849266 Chr.11: 118162907-118162907 on Build GRCh38
rs766966254 Chr.11: 118163067-118163067 on Build GRCh38
rs12276389 Chr.11: 118162803-118162803 on Build GRCh38
rs566704524 Chr.11: 118162858-118162858 on Build GRCh38
rs759094726 Chr.11: 118162768-118162768 on Build GRCh38
rs935993176 Chr.11: 118163110-118163110 on Build GRCh38
rs116573405 Chr.11: 118162995-118162995 on Build GRCh38
rs943325441 Chr.11: 118162819-118162819 on Build GRCh38
rs955417262 Chr.11: 118163029-118163029 on Build GRCh38
rs148125741 Chr.11: 118162948-118162948 on Build GRCh38
rs773962306 Chr.11: 118162751-118162751 on Build GRCh38
rs961570727 Chr.11: 118163003-118163003 on Build GRCh38
rs530938217 Chr.11: 118162800-118162800 on Build GRCh38
rs79825510 Chr.11: 118163097-118163097 on Build GRCh38
rs999526868 Chr.11: 118162942-118162942 on Build GRCh38
rs542204653 Chr.11: 118163095-118163096 on Build GRCh38
rs968909122 Chr.11: 118162908-118162908 on Build GRCh38
rs969832844 Chr.11: 118163018-118163018 on Build GRCh38
rs945239224 Chr.11: 118163191-118163191 on Build GRCh38
rs562179349 Chr.11: 118163119-118163119 on Build GRCh38
rs551078829 Chr.11: 118163196-118163196 on Build GRCh38
rs570959443 Chr.11: 118162953-118162953 on Build GRCh38
rs915106775 Chr.11: 118163186-118163186 on Build GRCh38
rs141919315 Chr.11: 118162971-118162971 on Build GRCh38
rs79365909 Chr.11: 118162799-118162799 on Build GRCh38
rs889161484 Chr.11: 118163163-118163163 on Build GRCh38
rs758772885 Chr.11: 118163011-118163011 on Build GRCh38
rs1037371741 Chr.11: 118163089-118163089 on Build GRCh38
rs150248095 Chr.11: 118163169-118163169 on Build GRCh38
rs989451772 Chr.11: 118163179-118163179 on Build GRCh38
rs933311805 Chr.11: 118163099-118163099 on Build GRCh38

View Full Product Details