Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118533779-118534276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM25
Entrez Gene ID
84866
Gene Name
transmembrane protein 25
Gene Aliases
-
UniGene
Hs.564188
Gene Chromosome Location
Chr.11: 118531088-118546670 on Build GRCh38
Transcripts
NM_001144034.1 NM_001144035.1 NM_001144036.1 NM_001144037.1 NM_001144038.1 NM_001318755.1 NM_001318757.1 NM_032780.3 NR_134852.1 XM_005271703.1 XM_006718926.2 XM_006718927.2 XM_011543037.2 XM_011543038.2 XM_017018422.1 XM_017018423.1 XM_017018424.1 XM_017018425.1 XM_017018426.1 XM_017018427.1 XM_017018428.1 XM_017018429.1 XM_017018430.1 XM_017018431.1 XM_017018432.1 XM_017018433.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5837937 c.941T>C p.V314A Chr.11: 118534269-118534269 on Build GRCh38
233604 c.833C>T p.S278L Chr.11: 118533884-118533884 on Build GRCh38
6320040 c.938-1G>T p.? Chr.11: 118534265-118534265 on Build GRCh38
5826905 c.897G>A p.P299P Chr.11: 118534089-118534089 on Build GRCh38
3397452 c.891C>T p.S297S Chr.11: 118534083-118534083 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782264638 Chr.11: 118534216-118534216 on Build GRCh38
rs782567522 Chr.11: 118533980-118533980 on Build GRCh38
rs371287328 Chr.11: 118534008-118534008 on Build GRCh38
rs910949064 Chr.11: 118533868-118533868 on Build GRCh38
rs782030117 Chr.11: 118534236-118534236 on Build GRCh38
rs182084396 Chr.11: 118533810-118533810 on Build GRCh38
rs782035583 Chr.11: 118534020-118534020 on Build GRCh38
rs375011884 Chr.11: 118534082-118534082 on Build GRCh38
rs781790620 Chr.11: 118534042-118534042 on Build GRCh38
rs782356577 Chr.11: 118534099-118534099 on Build GRCh38
rs114968068 Chr.11: 118534195-118534195 on Build GRCh38
rs368081342 Chr.11: 118534132-118534132 on Build GRCh38
rs782795849 Chr.11: 118534030-118534030 on Build GRCh38
rs370883247 Chr.11: 118533827-118533827 on Build GRCh38
rs782553610 Chr.11: 118534056-118534056 on Build GRCh38
rs548876211 Chr.11: 118534158-118534158 on Build GRCh38
rs139492240 Chr.11: 118533879-118533879 on Build GRCh38
rs140792350 Chr.11: 118534070-118534070 on Build GRCh38
rs782483904 Chr.11: 118533809-118533809 on Build GRCh38
rs782223739 Chr.11: 118534172-118534172 on Build GRCh38
rs1045611882 Chr.11: 118533932-118533932 on Build GRCh38
rs782273929 Chr.11: 118534245-118534245 on Build GRCh38
rs140346255 Chr.11: 118533874-118533874 on Build GRCh38
rs137969398 Chr.11: 118534069-118534069 on Build GRCh38
rs782304955 Chr.11: 118533824-118533824 on Build GRCh38
rs782812243 Chr.11: 118534130-118534130 on Build GRCh38
rs781953767 Chr.11: 118534006-118534006 on Build GRCh38
rs955040582 Chr.11: 118534086-118534086 on Build GRCh38
rs367849016 Chr.11: 118534002-118534002 on Build GRCh38
rs782126380 Chr.11: 118534134-118534134 on Build GRCh38
rs782658041 Chr.11: 118534057-118534057 on Build GRCh38
rs781865560 Chr.11: 118534060-118534060 on Build GRCh38
rs945855635 Chr.11: 118534209-118534209 on Build GRCh38
rs376513526 Chr.11: 118534218-118534218 on Build GRCh38
rs782466102 Chr.11: 118534009-118534009 on Build GRCh38
rs539153923 Chr.11: 118534235-118534235 on Build GRCh38
rs782291223 Chr.11: 118533811-118533812 on Build GRCh38
rs368260015 Chr.11: 118533924-118533924 on Build GRCh38
rs782787811 Chr.11: 118533846-118533846 on Build GRCh38
rs782420170 Chr.11: 118534041-118534041 on Build GRCh38
rs781906120 Chr.11: 118533831-118533831 on Build GRCh38
rs781979014 Chr.11: 118533857-118533857 on Build GRCh38
rs782490660 Chr.11: 118534061-118534061 on Build GRCh38
rs370198525 Chr.11: 118533946-118533946 on Build GRCh38
rs371605320 Chr.11: 118534154-118534154 on Build GRCh38
rs782357991 Chr.11: 118533807-118533807 on Build GRCh38
rs782257019 Chr.11: 118534095-118534095 on Build GRCh38
rs201380038 Chr.11: 118533989-118533989 on Build GRCh38
rs972977255 Chr.11: 118533806-118533806 on Build GRCh38
rs782754625 Chr.11: 118534136-118534136 on Build GRCh38
rs782384739 Chr.11: 118534253-118534253 on Build GRCh38
rs535014574 Chr.11: 118534211-118534211 on Build GRCh38
rs781889109 Chr.11: 118533915-118533915 on Build GRCh38
rs6589664 Chr.11: 118534089-118534089 on Build GRCh38
rs199815386 Chr.11: 118533865-118533865 on Build GRCh38
rs192529346 Chr.11: 118533929-118533929 on Build GRCh38
rs782146214 Chr.11: 118534027-118534027 on Build GRCh38
rs781936331 Chr.11: 118534115-118534115 on Build GRCh38
rs782406697 Chr.11: 118534088-118534088 on Build GRCh38
rs782304021 Chr.11: 118534087-118534087 on Build GRCh38
rs961656118 Chr.11: 118533802-118533802 on Build GRCh38
rs781830759 Chr.11: 118534142-118534142 on Build GRCh38
rs782804389 Chr.11: 118533908-118533908 on Build GRCh38
rs369458483 Chr.11: 118534233-118534233 on Build GRCh38
rs782078850 Chr.11: 118534037-118534037 on Build GRCh38
rs957174767 Chr.11: 118534124-118534124 on Build GRCh38
rs145299577 Chr.11: 118534035-118534035 on Build GRCh38
rs782735164 Chr.11: 118533866-118533866 on Build GRCh38
rs782044055 Chr.11: 118534118-118534118 on Build GRCh38
rs782236808 Chr.11: 118534004-118534004 on Build GRCh38
rs76950178 Chr.11: 118534038-118534038 on Build GRCh38
rs186720449 Chr.11: 118533828-118533828 on Build GRCh38
rs782307808 Chr.11: 118534044-118534044 on Build GRCh38
rs372823380 Chr.11: 118533939-118533939 on Build GRCh38
rs374438657 Chr.11: 118533990-118533990 on Build GRCh38
rs782621586 Chr.11: 118534067-118534067 on Build GRCh38
rs561626011 Chr.11: 118533923-118533923 on Build GRCh38
rs563773002 Chr.11: 118534122-118534122 on Build GRCh38
rs782579901 Chr.11: 118534016-118534016 on Build GRCh38

View Full Product Details