Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118534424-118534919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM25
Entrez Gene ID
84866
Gene Name
transmembrane protein 25
Gene Aliases
-
UniGene
Hs.564188
Gene Chromosome Location
Chr.11: 118531088-118546670 on Build GRCh38
Transcripts
NM_001144034.1 NM_001144035.1 NM_001144036.1 NM_001144037.1 NM_001144038.1 NM_001318755.1 NM_001318757.1 NM_032780.3 NR_134852.1 XM_005271703.1 XM_006718926.2 XM_006718927.2 XM_011543037.2 XM_011543038.2 XM_017018422.1 XM_017018423.1 XM_017018424.1 XM_017018425.1 XM_017018426.1 XM_017018427.1 XM_017018428.1 XM_017018429.1 XM_017018430.1 XM_017018431.1 XM_017018432.1 XM_017018433.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1604175 c.1028-4G>T p.? Chr.11: 118534503-118534503 on Build GRCh38
6485866 c.1063C>T p.R355* Chr.11: 118534542-118534542 on Build GRCh38
238321 c.1028-4G>A p.? Chr.11: 118534503-118534503 on Build GRCh38
4457092 c.1032C>T p.F344F Chr.11: 118534511-118534511 on Build GRCh38
7238457 c.1050G>T p.L350L Chr.11: 118534529-118534529 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782338676 Chr.11: 118534775-118534775 on Build GRCh38
rs892823641 Chr.11: 118534618-118534618 on Build GRCh38
rs782518937 Chr.11: 118534748-118534748 on Build GRCh38
rs146720659 Chr.11: 118534739-118534739 on Build GRCh38
rs782646518 Chr.11: 118534631-118534631 on Build GRCh38
rs512849 Chr.11: 118534628-118534628 on Build GRCh38
rs768273965 Chr.11: 118534531-118534531 on Build GRCh38
rs386757953 Chr.11: 118534722-118534724 on Build GRCh38
rs781975964 Chr.11: 118534581-118534581 on Build GRCh38
rs782345274 Chr.11: 118534563-118534563 on Build GRCh38
rs45522239 Chr.11: 118534723-118534723 on Build GRCh38
rs782792546 Chr.11: 118534471-118534471 on Build GRCh38
rs937314593 Chr.11: 118534479-118534479 on Build GRCh38
rs143320045 Chr.11: 118534584-118534584 on Build GRCh38
rs374676641 Chr.11: 118534450-118534450 on Build GRCh38
rs782573665 Chr.11: 118534484-118534484 on Build GRCh38
rs782047246 Chr.11: 118534589-118534589 on Build GRCh38
rs45474901 Chr.11: 118534738-118534738 on Build GRCh38
rs782303597 Chr.11: 118534485-118534485 on Build GRCh38
rs140014518 Chr.11: 118534582-118534582 on Build GRCh38
rs782341063 Chr.11: 118534583-118534583 on Build GRCh38
rs781838289 Chr.11: 118534529-118534529 on Build GRCh38
rs200372342 Chr.11: 118534486-118534486 on Build GRCh38
rs782666912 Chr.11: 118534487-118534487 on Build GRCh38
rs782023677 Chr.11: 118534597-118534597 on Build GRCh38
rs147480435 Chr.11: 118534546-118534546 on Build GRCh38
rs781926040 Chr.11: 118534470-118534470 on Build GRCh38
rs782126960 Chr.11: 118534517-118534517 on Build GRCh38
rs369490411 Chr.11: 118534571-118534571 on Build GRCh38
rs139331936 Chr.11: 118534773-118534773 on Build GRCh38
rs386757954 Chr.11: 118534738-118534740 on Build GRCh38
rs782540514 Chr.11: 118534629-118534629 on Build GRCh38
rs981208280 Chr.11: 118534444-118534444 on Build GRCh38
rs918580586 Chr.11: 118534801-118534801 on Build GRCh38
rs528361697 Chr.11: 118534850-118534850 on Build GRCh38
rs782012028 Chr.11: 118534516-118534516 on Build GRCh38
rs373128179 Chr.11: 118534503-118534503 on Build GRCh38
rs200093993 Chr.11: 118534515-118534515 on Build GRCh38
rs782513204 Chr.11: 118534543-118534543 on Build GRCh38
rs782136504 Chr.11: 118534601-118534601 on Build GRCh38
rs183299430 Chr.11: 118534502-118534502 on Build GRCh38
rs958771690 Chr.11: 118534691-118534691 on Build GRCh38
rs45548238 Chr.11: 118534722-118534722 on Build GRCh38
rs930081780 Chr.11: 118534811-118534811 on Build GRCh38
rs967134548 Chr.11: 118534654-118534654 on Build GRCh38
rs891309835 Chr.11: 118534827-118534827 on Build GRCh38
rs45485302 Chr.11: 118534662-118534662 on Build GRCh38
rs782274677 Chr.11: 118534565-118534565 on Build GRCh38
rs781847243 Chr.11: 118534614-118534614 on Build GRCh38
rs781869257 Chr.11: 118534458-118534458 on Build GRCh38
rs201031896 Chr.11: 118534525-118534525 on Build GRCh38
rs540591835 Chr.11: 118534679-118534679 on Build GRCh38
rs571575507 Chr.11: 118534557-118534557 on Build GRCh38
rs542400130 Chr.11: 118534787-118534787 on Build GRCh38
rs45505501 Chr.11: 118534590-118534590 on Build GRCh38
rs941342607 Chr.11: 118534776-118534776 on Build GRCh38
rs373031545 Chr.11: 118534602-118534602 on Build GRCh38
rs546940760 Chr.11: 118534554-118534554 on Build GRCh38
rs782557057 Chr.11: 118534535-118534535 on Build GRCh38
rs1035551968 Chr.11: 118534671-118534671 on Build GRCh38
rs782227074 Chr.11: 118534760-118534760 on Build GRCh38
rs782668188 Chr.11: 118534536-118534536 on Build GRCh38
rs193199803 Chr.11: 118534830-118534830 on Build GRCh38
rs762759732 Chr.11: 118534849-118534849 on Build GRCh38
rs782762698 Chr.11: 118534605-118534605 on Build GRCh38
rs782624272 Chr.11: 118534759-118534759 on Build GRCh38
rs782505053 Chr.11: 118534469-118534469 on Build GRCh38
rs546583320 Chr.11: 118534882-118534882 on Build GRCh38
rs199605481 Chr.11: 118534622-118534622 on Build GRCh38
rs1051170138 Chr.11: 118534819-118534819 on Build GRCh38
rs139193877 Chr.11: 118534540-118534540 on Build GRCh38
rs143259331 Chr.11: 118534542-118534542 on Build GRCh38

View Full Product Details