Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119185833-119186333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDZD3
Entrez Gene ID
79849
Gene Name
PDZ domain containing 3
Gene Aliases
IKEPP, NHERF4, PDZK2
UniGene
Hs.374726
Gene Chromosome Location
Chr.11: 119185457-119190223 on Build GRCh38
Transcripts
NM_001168468.1 NM_024791.3 NR_033122.1 XM_011543000.2 XM_011543001.2 XM_011543002.2 XM_011543005.2 XM_017018334.1 XM_017018335.1 XM_017018336.1 XM_017018337.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5932889 c.92C>T p.S31F Chr.11: 119186139-119186139 on Build GRCh38
7329791 c.47C>A p.A16E Chr.11: 119186209-119186209 on Build GRCh38
7268394 c.69C>A p.S23S Chr.11: 119186231-119186231 on Build GRCh38
6045702 c.53G>A p.R18Q Chr.11: 119186215-119186215 on Build GRCh38
4713453 c.16delC p.P7fs*29 Chr.11: 119186178-119186178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184359193 Chr.11: 119185963-119185963 on Build GRCh38
rs138112455 Chr.11: 119185936-119185936 on Build GRCh38
rs932617341 Chr.11: 119186152-119186152 on Build GRCh38
rs770637850 Chr.11: 119186159-119186159 on Build GRCh38
rs920315487 Chr.11: 119185973-119185973 on Build GRCh38
rs374000997 Chr.11: 119186155-119186155 on Build GRCh38
rs373685021 Chr.11: 119186057-119186057 on Build GRCh38
rs535876847 Chr.11: 119186080-119186080 on Build GRCh38
rs772694760 Chr.11: 119185944-119185944 on Build GRCh38
rs763105997 Chr.11: 119186287-119186287 on Build GRCh38
rs145857998 Chr.11: 119186156-119186156 on Build GRCh38
rs778275101 Chr.11: 119186163-119186163 on Build GRCh38
rs528710397 Chr.11: 119186242-119186248 on Build GRCh38
rs925923321 Chr.11: 119186257-119186257 on Build GRCh38
rs749439728 Chr.11: 119185906-119185906 on Build GRCh38
rs776104211 Chr.11: 119185867-119185867 on Build GRCh38
rs761145003 Chr.11: 119186119-119186119 on Build GRCh38
rs746860572 Chr.11: 119186064-119186064 on Build GRCh38
rs369620980 Chr.11: 119186146-119186146 on Build GRCh38
rs759297320 Chr.11: 119186235-119186235 on Build GRCh38
rs1031492888 Chr.11: 119185907-119185907 on Build GRCh38
rs779332496 Chr.11: 119186238-119186239 on Build GRCh38
rs200108938 Chr.11: 119186189-119186189 on Build GRCh38
rs762315159 Chr.11: 119186269-119186269 on Build GRCh38
rs754857242 Chr.11: 119186151-119186151 on Build GRCh38
rs950415421 Chr.11: 119186247-119186247 on Build GRCh38
rs202218681 Chr.11: 119186200-119186200 on Build GRCh38
rs768327702 Chr.11: 119185861-119185861 on Build GRCh38
rs1016486657 Chr.11: 119186193-119186193 on Build GRCh38
rs574754585 Chr.11: 119186036-119186036 on Build GRCh38
rs748833685 Chr.11: 119185952-119185953 on Build GRCh38
rs752989485 Chr.11: 119186176-119186177 on Build GRCh38
rs766400897 Chr.11: 119186214-119186214 on Build GRCh38
rs375849599 Chr.11: 119185995-119185995 on Build GRCh38
rs761290881 Chr.11: 119186179-119186179 on Build GRCh38
rs754318192 Chr.11: 119186190-119186193 on Build GRCh38
rs776181588 Chr.11: 119185893-119185895 on Build GRCh38
rs1026272064 Chr.11: 119186226-119186226 on Build GRCh38
rs754945375 Chr.11: 119186071-119186071 on Build GRCh38
rs752806323 Chr.11: 119185902-119185902 on Build GRCh38
rs769554452 Chr.11: 119185943-119185943 on Build GRCh38
rs745542687 Chr.11: 119186242-119186242 on Build GRCh38
rs542292854 Chr.11: 119186244-119186244 on Build GRCh38
rs556458439 Chr.11: 119186007-119186007 on Build GRCh38
rs759574044 Chr.11: 119186129-119186129 on Build GRCh38
rs983153195 Chr.11: 119186250-119186250 on Build GRCh38
rs769414569 Chr.11: 119186077-119186077 on Build GRCh38
rs377671878 Chr.11: 119186126-119186126 on Build GRCh38
rs776495808 Chr.11: 119185929-119185929 on Build GRCh38
rs773918256 Chr.11: 119186083-119186083 on Build GRCh38
rs757622655 Chr.11: 119186224-119186225 on Build GRCh38
rs200186795 Chr.11: 119185964-119185964 on Build GRCh38
rs757009602 Chr.11: 119186305-119186305 on Build GRCh38
rs757562208 Chr.11: 119186142-119186142 on Build GRCh38
rs762732068 Chr.11: 119185988-119185991 on Build GRCh38
rs773443795 Chr.11: 119186285-119186285 on Build GRCh38
rs558328629 Chr.11: 119186173-119186173 on Build GRCh38
rs201135995 Chr.11: 119186208-119186208 on Build GRCh38
rs754060799 Chr.11: 119186139-119186139 on Build GRCh38
rs1025143691 Chr.11: 119185921-119185921 on Build GRCh38
rs761409369 Chr.11: 119185909-119185911 on Build GRCh38
rs772891034 Chr.11: 119186078-119186078 on Build GRCh38
rs772430194 Chr.11: 119186165-119186165 on Build GRCh38
rs983187807 Chr.11: 119186010-119186010 on Build GRCh38
rs766286306 Chr.11: 119186093-119186098 on Build GRCh38
rs201438643 Chr.11: 119186298-119186298 on Build GRCh38
rs972115253 Chr.11: 119186225-119186225 on Build GRCh38
rs557483583 Chr.11: 119185882-119185883 on Build GRCh38
rs777136462 Chr.11: 119186190-119186190 on Build GRCh38
rs1002887014 Chr.11: 119186192-119186192 on Build GRCh38
rs369415415 Chr.11: 119186055-119186055 on Build GRCh38
rs753174596 Chr.11: 119185992-119185992 on Build GRCh38
rs781237348 Chr.11: 119186153-119186153 on Build GRCh38
rs748032812 Chr.11: 119186075-119186075 on Build GRCh38
rs764967969 Chr.11: 119186307-119186307 on Build GRCh38
rs749936614 Chr.11: 119185875-119185875 on Build GRCh38
rs760594369 Chr.11: 119186299-119186299 on Build GRCh38
rs976953649 Chr.11: 119186286-119186286 on Build GRCh38
rs769591612 Chr.11: 119185953-119185954 on Build GRCh38
rs762498139 Chr.11: 119185960-119185960 on Build GRCh38
rs994438361 Chr.11: 119186233-119186233 on Build GRCh38
rs751490670 Chr.11: 119186099-119186100 on Build GRCh38
rs769350909 Chr.11: 119186265-119186265 on Build GRCh38
rs371843707 Chr.11: 119185892-119185892 on Build GRCh38
rs773700945 Chr.11: 119186162-119186162 on Build GRCh38
rs764069301 Chr.11: 119186300-119186300 on Build GRCh38
rs757375681 Chr.11: 119186002-119186002 on Build GRCh38
rs202222447 Chr.11: 119186164-119186164 on Build GRCh38
rs554526048 Chr.11: 119186081-119186081 on Build GRCh38
rs972571194 Chr.11: 119186005-119186005 on Build GRCh38
rs772926771 Chr.11: 119185978-119185978 on Build GRCh38
rs45612531 Chr.11: 119185972-119185972 on Build GRCh38
rs780184915 Chr.11: 119185909-119185909 on Build GRCh38
rs761545329 Chr.11: 119185868-119185868 on Build GRCh38
rs778282407 Chr.11: 119186187-119186188 on Build GRCh38
rs781637056 Chr.11: 119185898-119185898 on Build GRCh38
rs753942497 Chr.11: 119186001-119186001 on Build GRCh38
rs748873672 Chr.11: 119186158-119186158 on Build GRCh38
rs756344540 Chr.11: 119186186-119186186 on Build GRCh38
rs756051469 Chr.11: 119186230-119186230 on Build GRCh38
rs759958037 Chr.11: 119186085-119186085 on Build GRCh38
rs543000579 Chr.11: 119186224-119186224 on Build GRCh38
rs191882344 Chr.11: 119185912-119185912 on Build GRCh38
rs763444293 Chr.11: 119185886-119185886 on Build GRCh38
rs141007283 Chr.11: 119186232-119186232 on Build GRCh38
rs562744717 Chr.11: 119185903-119185903 on Build GRCh38
rs1042942657 Chr.11: 119186137-119186137 on Build GRCh38
rs747450109 Chr.11: 119186256-119186256 on Build GRCh38
rs777541744 Chr.11: 119186231-119186231 on Build GRCh38
rs768129879 Chr.11: 119186097-119186097 on Build GRCh38
rs1014092922 Chr.11: 119186217-119186217 on Build GRCh38
rs1014003858 Chr.11: 119186160-119186160 on Build GRCh38
rs550902994 Chr.11: 119185871-119185871 on Build GRCh38
rs78436260 Chr.11: 119186255-119186255 on Build GRCh38
rs746995004 Chr.11: 119185910-119185910 on Build GRCh38
rs769874710 Chr.11: 119186278-119186278 on Build GRCh38
rs138704363 Chr.11: 119186157-119186157 on Build GRCh38
rs766416021 Chr.11: 119186288-119186288 on Build GRCh38
rs527643764 Chr.11: 119186304-119186304 on Build GRCh38
rs751899069 Chr.11: 119185888-119185888 on Build GRCh38
rs552538682 Chr.11: 119185969-119185969 on Build GRCh38
rs777120192 Chr.11: 119186267-119186267 on Build GRCh38
rs750580661 Chr.11: 119186147-119186147 on Build GRCh38
rs970090320 Chr.11: 119185966-119185966 on Build GRCh38
rs768076940 Chr.11: 119186247-119186250 on Build GRCh38
rs764462623 Chr.11: 119185996-119185996 on Build GRCh38
rs775697643 Chr.11: 119186098-119186098 on Build GRCh38
rs370592843 Chr.11: 119186074-119186074 on Build GRCh38

View Full Product Details