Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 31791032-31791576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
H3F3C
Entrez Gene ID
440093
Gene Name
H3 histone, family 3C
Gene Aliases
H3.5
UniGene
Hs.448697
Gene Chromosome Location
Chr.12: 31791185-31792241 on Build GRCh38
Transcripts
NM_001013699.2

SNP Detail

SNP ID Chromosome Location
rs959531675 Chr.12: 31791193-31791193 on Build GRCh38
rs142261223 Chr.12: 31791497-31791497 on Build GRCh38
rs970552516 Chr.12: 31791064-31791064 on Build GRCh38
rs564070029 Chr.12: 31791283-31791283 on Build GRCh38
rs765812183 Chr.12: 31791537-31791537 on Build GRCh38
rs34845138 Chr.12: 31791284-31791284 on Build GRCh38
rs780612913 Chr.12: 31791067-31791067 on Build GRCh38
rs566775619 Chr.12: 31791391-31791391 on Build GRCh38
rs878931549 Chr.12: 31791262-31791262 on Build GRCh38
rs944512888 Chr.12: 31791295-31791295 on Build GRCh38
rs398076550 Chr.12: 31791294-31791294 on Build GRCh38
rs774128699 Chr.12: 31791494-31791494 on Build GRCh38
rs189860374 Chr.12: 31791387-31791387 on Build GRCh38
rs747642627 Chr.12: 31791110-31791110 on Build GRCh38
rs772853625 Chr.12: 31791257-31791257 on Build GRCh38
rs770770505 Chr.12: 31791449-31791449 on Build GRCh38
rs550362047 Chr.12: 31791236-31791236 on Build GRCh38
rs1024398917 Chr.12: 31791058-31791058 on Build GRCh38
rs920230531 Chr.12: 31791215-31791215 on Build GRCh38
rs1039594749 Chr.12: 31791351-31791351 on Build GRCh38
rs561783829 Chr.12: 31791079-31791079 on Build GRCh38
rs1054731197 Chr.12: 31791403-31791403 on Build GRCh38
rs867509832 Chr.12: 31791232-31791232 on Build GRCh38
rs1001648883 Chr.12: 31791538-31791538 on Build GRCh38
rs530659713 Chr.12: 31791177-31791177 on Build GRCh38
rs950403798 Chr.12: 31791220-31791220 on Build GRCh38
rs1045763627 Chr.12: 31791521-31791521 on Build GRCh38
rs377724975 Chr.12: 31791170-31791170 on Build GRCh38
rs533042890 Chr.12: 31791326-31791326 on Build GRCh38
rs36058687 Chr.12: 31791398-31791398 on Build GRCh38
rs973321439 Chr.12: 31791213-31791213 on Build GRCh38
rs769360662 Chr.12: 31791149-31791149 on Build GRCh38
rs397851048 Chr.12: 31791295-31791295 on Build GRCh38
rs573769421 Chr.12: 31791396-31791403 on Build GRCh38
rs549218527 Chr.12: 31791469-31791469 on Build GRCh38
rs112907660 Chr.12: 31791542-31791542 on Build GRCh38
rs907277126 Chr.12: 31791534-31791534 on Build GRCh38
rs11051595 Chr.12: 31791541-31791541 on Build GRCh38

View Full Product Details