Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 32790528-32791067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PKP2
Entrez Gene ID
5318
Gene Name
plakophilin 2
Gene Aliases
ARVD9
UniGene
Hs.164384
Gene Chromosome Location
Chr.12: 32790746-32896846 on Build GRCh38
Transcripts
NM_001005242.2 NM_004572.3

SNP Detail

SNP ID Chromosome Location
rs1053179209 Chr.12: 32790773-32790773 on Build GRCh38
rs967952649 Chr.12: 32790982-32790982 on Build GRCh38
rs7312621 Chr.12: 32790595-32790595 on Build GRCh38
rs866441639 Chr.12: 32790939-32790939 on Build GRCh38
rs558152120 Chr.12: 32790829-32790829 on Build GRCh38
rs911120316 Chr.12: 32790716-32790716 on Build GRCh38
rs191176283 Chr.12: 32790671-32790671 on Build GRCh38
rs879877211 Chr.12: 32790612-32790612 on Build GRCh38
rs934630363 Chr.12: 32791019-32791019 on Build GRCh38
rs552084860 Chr.12: 32790899-32790899 on Build GRCh38
rs961071334 Chr.12: 32790613-32790613 on Build GRCh38
rs772250499 Chr.12: 32790676-32790676 on Build GRCh38
rs1018852485 Chr.12: 32790565-32790565 on Build GRCh38
rs934765727 Chr.12: 32790767-32790767 on Build GRCh38
rs926030767 Chr.12: 32790761-32790761 on Build GRCh38
rs955404839 Chr.12: 32790923-32790923 on Build GRCh38
rs974510944 Chr.12: 32790583-32790583 on Build GRCh38
rs970146142 Chr.12: 32790564-32790564 on Build GRCh38
rs563181178 Chr.12: 32790715-32790715 on Build GRCh38
rs760973876 Chr.12: 32790822-32790822 on Build GRCh38
rs1046150 Chr.12: 32790993-32790993 on Build GRCh38
rs895996553 Chr.12: 32790782-32790782 on Build GRCh38
rs925970516 Chr.12: 32791012-32791012 on Build GRCh38
rs961022087 Chr.12: 32790854-32790854 on Build GRCh38
rs11539836 Chr.12: 32790930-32790930 on Build GRCh38
rs911137911 Chr.12: 32790944-32790944 on Build GRCh38
rs972452445 Chr.12: 32790638-32790638 on Build GRCh38
rs10771995 Chr.12: 32790683-32790683 on Build GRCh38
rs547378328 Chr.12: 32790942-32790942 on Build GRCh38
rs114800490 Chr.12: 32790569-32790569 on Build GRCh38
rs1005176715 Chr.12: 32790838-32790838 on Build GRCh38
rs1044930221 Chr.12: 32790724-32790724 on Build GRCh38
rs947431140 Chr.12: 32790652-32790652 on Build GRCh38
rs1016592111 Chr.12: 32790852-32790852 on Build GRCh38
rs556485698 Chr.12: 32791038-32791038 on Build GRCh38
rs112547924 Chr.12: 32791030-32791030 on Build GRCh38
rs571538640 Chr.12: 32790802-32790802 on Build GRCh38
rs551630511 Chr.12: 32790707-32790707 on Build GRCh38
rs554108262 Chr.12: 32790883-32790884 on Build GRCh38
rs1057158875 Chr.12: 32790685-32790685 on Build GRCh38
rs987348070 Chr.12: 32790551-32790551 on Build GRCh38
rs1043045304 Chr.12: 32790747-32790747 on Build GRCh38
rs1014401016 Chr.12: 32790799-32790799 on Build GRCh38
rs112891106 Chr.12: 32790591-32790591 on Build GRCh38
rs1046154 Chr.12: 32790861-32790861 on Build GRCh38

View Full Product Details