Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 42317700-42318200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZCRB1
Entrez Gene ID
85437
Gene Name
zinc finger CCHC-type and RNA binding motif containing 1
Gene Aliases
MADP-1, MADP1, RBM36, SNRNP31, ZCCHC19
UniGene
Hs.496279
Gene Chromosome Location
Chr.12: 42312086-42326130 on Build GRCh38
Transcripts
NM_033114.3 XM_017020124.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
468336 c.204C>A p.T68T Chr.12: 42317808-42317808 on Build GRCh38
5622224 c.160G>A p.A54T Chr.12: 42317852-42317852 on Build GRCh38
1188470 c.209C>A p.A70E Chr.12: 42317803-42317803 on Build GRCh38
3968201 c.219C>A p.N73K Chr.12: 42317793-42317793 on Build GRCh38
939243 c.116T>A p.V39D Chr.12: 42317896-42317896 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs997870737 Chr.12: 42318122-42318122 on Build GRCh38
rs766385830 Chr.12: 42317914-42317914 on Build GRCh38
rs770310277 Chr.12: 42317849-42317849 on Build GRCh38
rs960594611 Chr.12: 42318130-42318130 on Build GRCh38
rs375892287 Chr.12: 42317826-42317826 on Build GRCh38
rs750269763 Chr.12: 42317747-42317747 on Build GRCh38
rs765039710 Chr.12: 42317928-42317928 on Build GRCh38
rs781682347 Chr.12: 42317815-42317815 on Build GRCh38
rs537122885 Chr.12: 42318087-42318087 on Build GRCh38
rs915964372 Chr.12: 42317898-42317898 on Build GRCh38
rs899903737 Chr.12: 42317958-42317958 on Build GRCh38
rs576162100 Chr.12: 42318057-42318057 on Build GRCh38
rs557919180 Chr.12: 42318012-42318012 on Build GRCh38
rs768551251 Chr.12: 42317872-42317872 on Build GRCh38
rs1026105639 Chr.12: 42317858-42317858 on Build GRCh38
rs146723341 Chr.12: 42317753-42317753 on Build GRCh38
rs530087726 Chr.12: 42317945-42317946 on Build GRCh38
rs759288360 Chr.12: 42317927-42317927 on Build GRCh38
rs771538069 Chr.12: 42317811-42317811 on Build GRCh38
rs113176268 Chr.12: 42317889-42317889 on Build GRCh38
rs951545698 Chr.12: 42318048-42318048 on Build GRCh38
rs887955877 Chr.12: 42317988-42317988 on Build GRCh38
rs138912646 Chr.12: 42317804-42317804 on Build GRCh38
rs775802409 Chr.12: 42317853-42317853 on Build GRCh38
rs758078383 Chr.12: 42317943-42317943 on Build GRCh38
rs375869339 Chr.12: 42317773-42317773 on Build GRCh38
rs763278060 Chr.12: 42317868-42317868 on Build GRCh38
rs777148079 Chr.12: 42317950-42317950 on Build GRCh38
rs957407438 Chr.12: 42317846-42317846 on Build GRCh38
rs777361182 Chr.12: 42317801-42317801 on Build GRCh38
rs1018362780 Chr.12: 42317752-42317752 on Build GRCh38
rs935163224 Chr.12: 42317933-42317933 on Build GRCh38
rs977727528 Chr.12: 42317822-42317822 on Build GRCh38
rs775283823 Chr.12: 42317999-42317999 on Build GRCh38
rs143792467 Chr.12: 42317916-42317916 on Build GRCh38
rs767826976 Chr.12: 42317913-42317914 on Build GRCh38
rs774787885 Chr.12: 42317875-42317877 on Build GRCh38
rs774555988 Chr.12: 42317880-42317880 on Build GRCh38
rs371619207 Chr.12: 42317793-42317793 on Build GRCh38
rs566318216 Chr.12: 42317931-42317931 on Build GRCh38
rs752379848 Chr.12: 42317779-42317779 on Build GRCh38
rs1035946183 Chr.12: 42318127-42318127 on Build GRCh38
rs368470866 Chr.12: 42317745-42317745 on Build GRCh38
rs774750463 Chr.12: 42318171-42318171 on Build GRCh38
rs778280219 Chr.12: 42317929-42317929 on Build GRCh38
rs113734418 Chr.12: 42317952-42317952 on Build GRCh38
rs759988364 Chr.12: 42317880-42317880 on Build GRCh38
rs1029311434 Chr.12: 42318152-42318152 on Build GRCh38
rs113567879 Chr.12: 42317765-42317765 on Build GRCh38
rs771593784 Chr.12: 42317868-42317868 on Build GRCh38
rs951642891 Chr.12: 42317790-42317790 on Build GRCh38
rs928891336 Chr.12: 42317823-42317824 on Build GRCh38
rs778578854 Chr.12: 42317775-42317775 on Build GRCh38
rs752289957 Chr.12: 42317942-42317942 on Build GRCh38
rs368015638 Chr.12: 42317769-42317769 on Build GRCh38
rs779658020 Chr.12: 42317749-42317749 on Build GRCh38

View Full Product Details