Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 43797524-43798070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM117
Entrez Gene ID
84216
Gene Name
transmembrane protein 117
Gene Aliases
-
UniGene
Hs.444668
Gene Chromosome Location
Chr.12: 43795806-44389762 on Build GRCh38
Transcripts
XM_011538831.2 NM_001242397.1 NM_002822.4 NR_073472.1 NR_073473.1

Gene Symbol
TWF1
Entrez Gene ID
5756
Gene Name
twinfilin actin binding protein 1
Gene Aliases
A6, PTK9
UniGene
Hs.189075
Gene Chromosome Location
Chr.12: 43793723-43806375 on Build GRCh38
Transcripts
XM_011538831.2 NM_001242397.1 NM_002822.4 NR_073472.1 NR_073473.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4907429 c.598T>A p.L200M Chr.12: 43797740-43797740 on Build GRCh38
6304657 c.565A>G p.I189V Chr.12: 43797773-43797773 on Build GRCh38
4907409 c.627G>A p.Q209Q Chr.12: 43797711-43797711 on Build GRCh38
6054156 c.615C>A p.L205L Chr.12: 43797723-43797723 on Build GRCh38
7247711 c.572G>C p.R191P Chr.12: 43797766-43797766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs996390768 Chr.12: 43797929-43797929 on Build GRCh38
rs976662003 Chr.12: 43797982-43797982 on Build GRCh38
rs989587609 Chr.12: 43797649-43797649 on Build GRCh38
rs774469240 Chr.12: 43797885-43797885 on Build GRCh38
rs957716891 Chr.12: 43797617-43797617 on Build GRCh38
rs560109504 Chr.12: 43797838-43797838 on Build GRCh38
rs745493693 Chr.12: 43797817-43797817 on Build GRCh38
rs767247411 Chr.12: 43797694-43797694 on Build GRCh38
rs771059417 Chr.12: 43797833-43797833 on Build GRCh38
rs386762596 Chr.12: 43797990-43797992 on Build GRCh38
rs200546520 Chr.12: 43797992-43797992 on Build GRCh38
rs768597831 Chr.12: 43797857-43797857 on Build GRCh38
rs989262380 Chr.12: 43797832-43797832 on Build GRCh38
rs551046347 Chr.12: 43797912-43797912 on Build GRCh38
rs559992269 Chr.12: 43797670-43797670 on Build GRCh38
rs746551688 Chr.12: 43797985-43797985 on Build GRCh38
rs1024536429 Chr.12: 43797701-43797701 on Build GRCh38
rs572194838 Chr.12: 43797578-43797579 on Build GRCh38
rs1038145286 Chr.12: 43797910-43797910 on Build GRCh38
rs534411387 Chr.12: 43798023-43798023 on Build GRCh38
rs779362774 Chr.12: 43797933-43797933 on Build GRCh38
rs778809199 Chr.12: 43797763-43797763 on Build GRCh38
rs55979505 Chr.12: 43797826-43797826 on Build GRCh38
rs762880207 Chr.12: 43797659-43797659 on Build GRCh38
rs541755170 Chr.12: 43797571-43797571 on Build GRCh38
rs144360384 Chr.12: 43797924-43797924 on Build GRCh38
rs147256169 Chr.12: 43797580-43797581 on Build GRCh38
rs770233154 Chr.12: 43798004-43798004 on Build GRCh38
rs144307978 Chr.12: 43797987-43797987 on Build GRCh38
rs567817752 Chr.12: 43797911-43797911 on Build GRCh38
rs1051849084 Chr.12: 43797973-43797973 on Build GRCh38
rs981908155 Chr.12: 43797723-43797723 on Build GRCh38
rs760088863 Chr.12: 43797760-43797760 on Build GRCh38
rs17093732 Chr.12: 43797672-43797672 on Build GRCh38
rs947956198 Chr.12: 43797712-43797712 on Build GRCh38
rs373979274 Chr.12: 43797840-43797840 on Build GRCh38
rs765850455 Chr.12: 43797761-43797761 on Build GRCh38
rs558835219 Chr.12: 43798044-43798044 on Build GRCh38
rs761730564 Chr.12: 43797686-43797686 on Build GRCh38
rs1001689121 Chr.12: 43797808-43797808 on Build GRCh38
rs764473250 Chr.12: 43797726-43797729 on Build GRCh38
rs753641006 Chr.12: 43797591-43797591 on Build GRCh38
rs777169099 Chr.12: 43797731-43797733 on Build GRCh38
rs762627379 Chr.12: 43797838-43797839 on Build GRCh38
rs913239779 Chr.12: 43797859-43797859 on Build GRCh38
rs777961269 Chr.12: 43797821-43797821 on Build GRCh38
rs372296866 Chr.12: 43797766-43797766 on Build GRCh38
rs115243912 Chr.12: 43797992-43797992 on Build GRCh38
rs12829856 Chr.12: 43797822-43797822 on Build GRCh38
rs565383143 Chr.12: 43797795-43797795 on Build GRCh38
rs1036706083 Chr.12: 43797654-43797654 on Build GRCh38
rs755464501 Chr.12: 43797775-43797775 on Build GRCh38
rs536627879 Chr.12: 43797927-43797927 on Build GRCh38
rs745952843 Chr.12: 43797853-43797853 on Build GRCh38
rs1005670687 Chr.12: 43797648-43797649 on Build GRCh38
rs548923462 Chr.12: 43797990-43797990 on Build GRCh38
rs766099263 Chr.12: 43797828-43797828 on Build GRCh38
rs753032364 Chr.12: 43797812-43797812 on Build GRCh38
rs750148003 Chr.12: 43797732-43797732 on Build GRCh38
rs890639761 Chr.12: 43797974-43797974 on Build GRCh38

View Full Product Details