Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 80445352-80445849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRQ
Entrez Gene ID
374462
Gene Name
protein tyrosine phosphatase, receptor type Q
Gene Aliases
DFNB84, DFNB84A, PTPGMC1, R-PTP-Q
UniGene
Hs.539284
Gene Chromosome Location
Chr.12: 80424261-80680273 on Build GRCh38
Transcripts
NM_001145026.1 XM_017019273.1 XM_017019274.1 XM_017019275.1

SNP Detail

SNP ID Chromosome Location
rs749326611 Chr.12: 80445714-80445714 on Build GRCh38
rs184871373 Chr.12: 80445582-80445582 on Build GRCh38
rs531266049 Chr.12: 80445647-80445647 on Build GRCh38
rs761005570 Chr.12: 80445803-80445803 on Build GRCh38
rs148527546 Chr.12: 80445613-80445613 on Build GRCh38
rs945515857 Chr.12: 80445744-80445744 on Build GRCh38
rs866508489 Chr.12: 80445470-80445470 on Build GRCh38
rs866361668 Chr.12: 80445577-80445577 on Build GRCh38
rs766087465 Chr.12: 80445538-80445538 on Build GRCh38
rs61729303 Chr.12: 80445529-80445529 on Build GRCh38
rs532097338 Chr.12: 80445769-80445769 on Build GRCh38
rs763472295 Chr.12: 80445583-80445583 on Build GRCh38
rs953285669 Chr.12: 80445475-80445475 on Build GRCh38
rs919038441 Chr.12: 80445746-80445746 on Build GRCh38
rs888732444 Chr.12: 80445581-80445581 on Build GRCh38
rs1042566009 Chr.12: 80445750-80445750 on Build GRCh38
rs1018785665 Chr.12: 80445742-80445742 on Build GRCh38
rs1003677730 Chr.12: 80445619-80445619 on Build GRCh38
rs564939738 Chr.12: 80445748-80445748 on Build GRCh38
rs1024136876 Chr.12: 80445608-80445608 on Build GRCh38
rs60509052 Chr.12: 80445440-80445440 on Build GRCh38
rs957098555 Chr.12: 80445664-80445664 on Build GRCh38
rs867256932 Chr.12: 80445645-80445645 on Build GRCh38
rs1010928776 Chr.12: 80445678-80445678 on Build GRCh38
rs190463905 Chr.12: 80445735-80445735 on Build GRCh38
rs915178529 Chr.12: 80445446-80445446 on Build GRCh38
rs911725794 Chr.12: 80445515-80445515 on Build GRCh38
rs1053953555 Chr.12: 80445487-80445487 on Build GRCh38
rs768055115 Chr.12: 80445510-80445510 on Build GRCh38
rs889815704 Chr.12: 80445792-80445792 on Build GRCh38
rs769654922 Chr.12: 80445570-80445570 on Build GRCh38
rs1036121588 Chr.12: 80445524-80445524 on Build GRCh38
rs979728496 Chr.12: 80445635-80445635 on Build GRCh38
rs187435968 Chr.12: 80445528-80445528 on Build GRCh38
rs558947480 Chr.12: 80445516-80445516 on Build GRCh38
rs755453927 Chr.12: 80445592-80445592 on Build GRCh38
rs956737760 Chr.12: 80445436-80445436 on Build GRCh38
rs4268567 Chr.12: 80445417-80445417 on Build GRCh38
rs151025918 Chr.12: 80445755-80445755 on Build GRCh38
rs577247628 Chr.12: 80445522-80445522 on Build GRCh38
rs1006090903 Chr.12: 80445562-80445562 on Build GRCh38
rs1043006583 Chr.12: 80445517-80445517 on Build GRCh38
rs961472539 Chr.12: 80445584-80445584 on Build GRCh38
rs764016474 Chr.12: 80445753-80445753 on Build GRCh38
rs990337020 Chr.12: 80445694-80445694 on Build GRCh38
rs761805006 Chr.12: 80445523-80445523 on Build GRCh38
rs972284793 Chr.12: 80445745-80445745 on Build GRCh38
rs994033861 Chr.12: 80445605-80445605 on Build GRCh38
rs773511370 Chr.12: 80445515-80445515 on Build GRCh38
rs756626854 Chr.12: 80445656-80445656 on Build GRCh38
rs1038835687 Chr.12: 80445814-80445814 on Build GRCh38
rs985962173 Chr.12: 80445482-80445482 on Build GRCh38
rs61729302 Chr.12: 80445533-80445533 on Build GRCh38
rs767425598 Chr.12: 80445552-80445552 on Build GRCh38
rs953336283 Chr.12: 80445698-80445698 on Build GRCh38
rs748647118 Chr.12: 80445621-80445621 on Build GRCh38

View Full Product Details