Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120303599-120304088 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIRT4
Entrez Gene ID
23409
Gene Name
sirtuin 4
Gene Aliases
SIR2L4
UniGene
Hs.50861
Gene Chromosome Location
Chr.12: 120292199-120313242 on Build GRCh38
Transcripts
NM_012240.2 XM_005253865.4 XM_006719308.2 XM_006719309.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6218074 c.483C>T p.H161H Chr.12: 120304044-120304044 on Build GRCh38
6071232 c.488G>T p.C163F Chr.12: 120304049-120304049 on Build GRCh38
4039394 c.361C>T p.P121S Chr.12: 120303922-120303922 on Build GRCh38
6318676 c.370G>T p.A124S Chr.12: 120303931-120303931 on Build GRCh38
4510858 c.84C>T p.S28S Chr.12: 120303645-120303645 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369009892 Chr.12: 120303812-120303812 on Build GRCh38
rs746601591 Chr.12: 120304044-120304044 on Build GRCh38
rs770666458 Chr.12: 120303871-120303871 on Build GRCh38
rs758753317 Chr.12: 120303750-120303750 on Build GRCh38
rs745477180 Chr.12: 120303694-120303694 on Build GRCh38
rs144924773 Chr.12: 120303621-120303621 on Build GRCh38
rs768961942 Chr.12: 120303909-120303928 on Build GRCh38
rs763432378 Chr.12: 120303901-120303901 on Build GRCh38
rs903042992 Chr.12: 120303705-120303705 on Build GRCh38
rs371691236 Chr.12: 120303862-120303862 on Build GRCh38
rs368638386 Chr.12: 120303681-120303681 on Build GRCh38
rs368193292 Chr.12: 120303824-120303824 on Build GRCh38
rs762269633 Chr.12: 120304008-120304009 on Build GRCh38
rs757053577 Chr.12: 120303934-120303934 on Build GRCh38
rs1022881826 Chr.12: 120304022-120304022 on Build GRCh38
rs751124265 Chr.12: 120303703-120303703 on Build GRCh38
rs201019576 Chr.12: 120303677-120303677 on Build GRCh38
rs771865381 Chr.12: 120303701-120303702 on Build GRCh38
rs201393186 Chr.12: 120303872-120303872 on Build GRCh38
rs763487544 Chr.12: 120303975-120303975 on Build GRCh38
rs771611985 Chr.12: 120304057-120304057 on Build GRCh38
rs151314467 Chr.12: 120303670-120303670 on Build GRCh38
rs779080507 Chr.12: 120303728-120303728 on Build GRCh38
rs767763097 Chr.12: 120303908-120303908 on Build GRCh38
rs201166468 Chr.12: 120303850-120303850 on Build GRCh38
rs751811664 Chr.12: 120303905-120303905 on Build GRCh38
rs200817330 Chr.12: 120303889-120303889 on Build GRCh38
rs375210930 Chr.12: 120303948-120303948 on Build GRCh38
rs774712434 Chr.12: 120303909-120303911 on Build GRCh38
rs200695003 Chr.12: 120303629-120303629 on Build GRCh38
rs749067576 Chr.12: 120303778-120303778 on Build GRCh38
rs760212493 Chr.12: 120303884-120303884 on Build GRCh38
rs776399992 Chr.12: 120303661-120303661 on Build GRCh38
rs146483726 Chr.12: 120303955-120303955 on Build GRCh38
rs200022151 Chr.12: 120303976-120303976 on Build GRCh38
rs766884688 Chr.12: 120303814-120303814 on Build GRCh38
rs752917367 Chr.12: 120304017-120304017 on Build GRCh38
rs751683420 Chr.12: 120303841-120303841 on Build GRCh38
rs753915067 Chr.12: 120304027-120304027 on Build GRCh38
rs779594236 Chr.12: 120304042-120304042 on Build GRCh38
rs774632340 Chr.12: 120303990-120303990 on Build GRCh38
rs778125945 Chr.12: 120303623-120303623 on Build GRCh38
rs761698760 Chr.12: 120303671-120303671 on Build GRCh38
rs551853830 Chr.12: 120303763-120303763 on Build GRCh38
rs200041014 Chr.12: 120303636-120303636 on Build GRCh38
rs770616938 Chr.12: 120303797-120303797 on Build GRCh38
rs762295806 Chr.12: 120303821-120303821 on Build GRCh38
rs201402427 Chr.12: 120303892-120303892 on Build GRCh38
rs754272406 Chr.12: 120303624-120303624 on Build GRCh38
rs199844619 Chr.12: 120303882-120303882 on Build GRCh38
rs202219623 Chr.12: 120304025-120304025 on Build GRCh38
rs756153620 Chr.12: 120303912-120303912 on Build GRCh38
rs766421957 Chr.12: 120303861-120303861 on Build GRCh38
rs766790909 Chr.12: 120303904-120303904 on Build GRCh38
rs200624221 Chr.12: 120304011-120304011 on Build GRCh38
rs201686223 Chr.12: 120303630-120303630 on Build GRCh38
rs372046001 Chr.12: 120303711-120303711 on Build GRCh38
rs765009393 Chr.12: 120303699-120303699 on Build GRCh38
rs752765477 Chr.12: 120303911-120303911 on Build GRCh38
rs140277065 Chr.12: 120303877-120303877 on Build GRCh38
rs202016280 Chr.12: 120303876-120303876 on Build GRCh38
rs145385123 Chr.12: 120303881-120303881 on Build GRCh38
rs778888736 Chr.12: 120304029-120304029 on Build GRCh38
rs375186455 Chr.12: 120303773-120303773 on Build GRCh38
rs771556756 Chr.12: 120303977-120303977 on Build GRCh38
rs758806432 Chr.12: 120303833-120303833 on Build GRCh38
rs755283087 Chr.12: 120303669-120303671 on Build GRCh38
rs769632132 Chr.12: 120303768-120303768 on Build GRCh38
rs778750745 Chr.12: 120303945-120303945 on Build GRCh38
rs765814554 Chr.12: 120303619-120303619 on Build GRCh38
rs768286255 Chr.12: 120304051-120304051 on Build GRCh38
rs774887268 Chr.12: 120304061-120304061 on Build GRCh38
rs768689629 Chr.12: 120303647-120303647 on Build GRCh38
rs780402014 Chr.12: 120303752-120303752 on Build GRCh38
rs957287189 Chr.12: 120303792-120303792 on Build GRCh38
rs565558341 Chr.12: 120303733-120303733 on Build GRCh38
rs201091007 Chr.12: 120304028-120304028 on Build GRCh38
rs12307919 Chr.12: 120303865-120303865 on Build GRCh38
rs768231075 Chr.12: 120303970-120303970 on Build GRCh38
rs779222815 Chr.12: 120303631-120303631 on Build GRCh38
rs769880558 Chr.12: 120304065-120304066 on Build GRCh38
rs764695227 Chr.12: 120303707-120303707 on Build GRCh38
rs755218104 Chr.12: 120303906-120303906 on Build GRCh38
rs375219114 Chr.12: 120303891-120303891 on Build GRCh38
rs199996507 Chr.12: 120303811-120303811 on Build GRCh38
rs145472722 Chr.12: 120303866-120303866 on Build GRCh38
rs755098471 Chr.12: 120303842-120303842 on Build GRCh38
rs750869437 Chr.12: 120303831-120303831 on Build GRCh38
rs971421473 Chr.12: 120303817-120303817 on Build GRCh38
rs780500047 Chr.12: 120303737-120303738 on Build GRCh38
rs750226767 Chr.12: 120304030-120304030 on Build GRCh38
rs545864291 Chr.12: 120303641-120303641 on Build GRCh38
rs201713338 Chr.12: 120303706-120303706 on Build GRCh38
rs762926007 Chr.12: 120303837-120303837 on Build GRCh38
rs766009438 Chr.12: 120303685-120303685 on Build GRCh38
rs143132490 Chr.12: 120304052-120304052 on Build GRCh38
rs916643694 Chr.12: 120304045-120304045 on Build GRCh38
rs199700265 Chr.12: 120303909-120303909 on Build GRCh38
rs773868069 Chr.12: 120303972-120303972 on Build GRCh38
rs777204231 Chr.12: 120303820-120303820 on Build GRCh38
rs747374479 Chr.12: 120303745-120303745 on Build GRCh38
rs200142301 Chr.12: 120304016-120304016 on Build GRCh38
rs757657278 Chr.12: 120303726-120303726 on Build GRCh38
rs139416968 Chr.12: 120303849-120303849 on Build GRCh38
rs772860627 Chr.12: 120303679-120303679 on Build GRCh38
rs778326065 Chr.12: 120303684-120303684 on Build GRCh38
rs749168988 Chr.12: 120303928-120303928 on Build GRCh38
rs777713269 Chr.12: 120303918-120303918 on Build GRCh38
rs140954170 Chr.12: 120303620-120303620 on Build GRCh38
rs201604619 Chr.12: 120304014-120304014 on Build GRCh38
rs745662184 Chr.12: 120303953-120303953 on Build GRCh38
rs200820893 Chr.12: 120303963-120303963 on Build GRCh38
rs772284370 Chr.12: 120303635-120303635 on Build GRCh38
rs528028271 Chr.12: 120303762-120303762 on Build GRCh38
rs746032708 Chr.12: 120303633-120303633 on Build GRCh38
rs113815121 Chr.12: 120303903-120303903 on Build GRCh38
rs772353626 Chr.12: 120303952-120303953 on Build GRCh38
rs182780868 Chr.12: 120303767-120303767 on Build GRCh38
rs759847738 Chr.12: 120304063-120304063 on Build GRCh38
rs141059071 Chr.12: 120304032-120304032 on Build GRCh38
rs760760134 Chr.12: 120304019-120304019 on Build GRCh38
rs201382659 Chr.12: 120303964-120303964 on Build GRCh38

View Full Product Details