Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120700120-120700618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MLEC
Entrez Gene ID
9761
Gene Name
malectin
Gene Aliases
KIAA0152
UniGene
Hs.744910
Gene Chromosome Location
Chr.12: 120687124-120701864 on Build GRCh38
Transcripts
NM_001303627.1 NM_001303628.1 NM_014730.3 XM_011539031.1 XM_011539032.1

SNP Detail

SNP ID Chromosome Location
rs557747789 Chr.12: 120700163-120700163 on Build GRCh38
rs73225033 Chr.12: 120700590-120700590 on Build GRCh38
rs778357910 Chr.12: 120700559-120700559 on Build GRCh38
rs935461222 Chr.12: 120700388-120700388 on Build GRCh38
rs905501040 Chr.12: 120700506-120700506 on Build GRCh38
rs3751166 Chr.12: 120700474-120700474 on Build GRCh38
rs1038052162 Chr.12: 120700279-120700279 on Build GRCh38
rs140187895 Chr.12: 120700302-120700302 on Build GRCh38
rs145559337 Chr.12: 120700580-120700580 on Build GRCh38
rs952814132 Chr.12: 120700550-120700550 on Build GRCh38
rs572243451 Chr.12: 120700449-120700449 on Build GRCh38
rs979350554 Chr.12: 120700449-120700452 on Build GRCh38
rs1052839285 Chr.12: 120700405-120700405 on Build GRCh38
rs559839026 Chr.12: 120700327-120700327 on Build GRCh38
rs895656106 Chr.12: 120700535-120700535 on Build GRCh38
rs898198980 Chr.12: 120700288-120700288 on Build GRCh38
rs758035580 Chr.12: 120700521-120700521 on Build GRCh38
rs969280289 Chr.12: 120700448-120700449 on Build GRCh38
rs913288507 Chr.12: 120700170-120700170 on Build GRCh38
rs942840570 Chr.12: 120700552-120700552 on Build GRCh38
rs760880699 Chr.12: 120700362-120700362 on Build GRCh38
rs560736 Chr.12: 120700547-120700547 on Build GRCh38
rs1035539601 Chr.12: 120700166-120700166 on Build GRCh38
rs551060495 Chr.12: 120700536-120700536 on Build GRCh38
rs942947227 Chr.12: 120700298-120700298 on Build GRCh38
rs186688524 Chr.12: 120700144-120700144 on Build GRCh38
rs905763869 Chr.12: 120700156-120700156 on Build GRCh38
rs952864213 Chr.12: 120700262-120700262 on Build GRCh38
rs959830638 Chr.12: 120700169-120700169 on Build GRCh38
rs776119576 Chr.12: 120700268-120700268 on Build GRCh38
rs972822052 Chr.12: 120700252-120700252 on Build GRCh38
rs1023866930 Chr.12: 120700443-120700443 on Build GRCh38
rs993834039 Chr.12: 120700335-120700335 on Build GRCh38
rs547194 Chr.12: 120700495-120700495 on Build GRCh38
rs1013433418 Chr.12: 120700410-120700410 on Build GRCh38
rs191100831 Chr.12: 120700339-120700339 on Build GRCh38
rs1035574084 Chr.12: 120700522-120700522 on Build GRCh38
rs532100170 Chr.12: 120700505-120700505 on Build GRCh38
rs1025697097 Chr.12: 120700254-120700254 on Build GRCh38
rs1803510 Chr.12: 120700195-120700195 on Build GRCh38
rs768045388 Chr.12: 120700544-120700544 on Build GRCh38
rs934042933 Chr.12: 120700541-120700541 on Build GRCh38
rs1032217684 Chr.12: 120700464-120700464 on Build GRCh38
rs367799664 Chr.12: 120700363-120700363 on Build GRCh38
rs890060946 Chr.12: 120700256-120700256 on Build GRCh38
rs79711373 Chr.12: 120700558-120700558 on Build GRCh38
rs147547010 Chr.12: 120700271-120700271 on Build GRCh38

View Full Product Details