Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 20367417-20367931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEP1
Entrez Gene ID
7011
Gene Name
telomerase associated protein 1
Gene Aliases
TLP1, TP1, TROVE1, VAULT2, p240
UniGene
Hs.508835
Gene Chromosome Location
Chr.14: 20365667-20413540 on Build GRCh38
Transcripts
NM_001319035.1 NM_007110.4 XM_005268027.4 XM_011537110.2

SNP Detail

SNP ID Chromosome Location
rs1051114331 Chr.14: 20367565-20367565 on Build GRCh38
rs566818873 Chr.14: 20367703-20367703 on Build GRCh38
rs907384022 Chr.14: 20367643-20367643 on Build GRCh38
rs534325400 Chr.14: 20367848-20367848 on Build GRCh38
rs537395690 Chr.14: 20367448-20367448 on Build GRCh38
rs996624979 Chr.14: 20367784-20367785 on Build GRCh38
rs567699830 Chr.14: 20367906-20367906 on Build GRCh38
rs140444610 Chr.14: 20367650-20367650 on Build GRCh38
rs1034658346 Chr.14: 20367472-20367472 on Build GRCh38
rs115962044 Chr.14: 20367526-20367526 on Build GRCh38
rs868132838 Chr.14: 20367860-20367860 on Build GRCh38
rs765044001 Chr.14: 20367733-20367733 on Build GRCh38
rs763846172 Chr.14: 20367825-20367825 on Build GRCh38
rs542208396 Chr.14: 20367737-20367737 on Build GRCh38
rs999169671 Chr.14: 20367904-20367904 on Build GRCh38
rs76335370 Chr.14: 20367489-20367489 on Build GRCh38
rs1056010773 Chr.14: 20367695-20367695 on Build GRCh38
rs549394001 Chr.14: 20367853-20367853 on Build GRCh38
rs988123790 Chr.14: 20367820-20367820 on Build GRCh38
rs191507131 Chr.14: 20367758-20367758 on Build GRCh38
rs757662927 Chr.14: 20367560-20367562 on Build GRCh38
rs555429425 Chr.14: 20367898-20367898 on Build GRCh38
rs911327795 Chr.14: 20367567-20367567 on Build GRCh38
rs959151244 Chr.14: 20367671-20367671 on Build GRCh38
rs942691335 Chr.14: 20367573-20367573 on Build GRCh38
rs934048096 Chr.14: 20367538-20367538 on Build GRCh38
rs919957787 Chr.14: 20367761-20367761 on Build GRCh38
rs146424991 Chr.14: 20367905-20367905 on Build GRCh38
rs968344143 Chr.14: 20367684-20367684 on Build GRCh38
rs534946326 Chr.14: 20367529-20367529 on Build GRCh38
rs369100214 Chr.14: 20367615-20367615 on Build GRCh38
rs752510796 Chr.14: 20367760-20367760 on Build GRCh38
rs1713442 Chr.14: 20367632-20367632 on Build GRCh38
rs188222816 Chr.14: 20367710-20367710 on Build GRCh38
rs1022347985 Chr.14: 20367889-20367889 on Build GRCh38
rs923885730 Chr.14: 20367709-20367709 on Build GRCh38
rs987766632 Chr.14: 20367630-20367630 on Build GRCh38
rs915141265 Chr.14: 20367522-20367522 on Build GRCh38
rs548102163 Chr.14: 20367800-20367800 on Build GRCh38
rs917067262 Chr.14: 20367899-20367899 on Build GRCh38
rs923793947 Chr.14: 20367534-20367534 on Build GRCh38
rs762785016 Chr.14: 20367811-20367811 on Build GRCh38
rs1003071975 Chr.14: 20367646-20367646 on Build GRCh38
rs570123995 Chr.14: 20367844-20367844 on Build GRCh38
rs920613363 Chr.14: 20367862-20367862 on Build GRCh38
rs955324951 Chr.14: 20367907-20367907 on Build GRCh38
rs559808274 Chr.14: 20367801-20367801 on Build GRCh38
rs551838042 Chr.14: 20367677-20367677 on Build GRCh38
rs755533212 Chr.14: 20367891-20367891 on Build GRCh38
rs930191976 Chr.14: 20367772-20367772 on Build GRCh38
rs868284752 Chr.14: 20367702-20367702 on Build GRCh38
rs1029565448 Chr.14: 20367802-20367802 on Build GRCh38
rs560597767 Chr.14: 20367757-20367757 on Build GRCh38
rs879743976 Chr.14: 20367616-20367616 on Build GRCh38
rs1038404591 Chr.14: 20367574-20367574 on Build GRCh38
rs994434033 Chr.14: 20367606-20367606 on Build GRCh38
rs967060423 Chr.14: 20367488-20367488 on Build GRCh38
rs898707669 Chr.14: 20367587-20367587 on Build GRCh38
rs1020357737 Chr.14: 20367474-20367474 on Build GRCh38
rs902843554 Chr.14: 20367753-20367753 on Build GRCh38
rs990524206 Chr.14: 20367506-20367506 on Build GRCh38
rs544897623 Chr.14: 20367696-20367696 on Build GRCh38
rs1047239784 Chr.14: 20367608-20367608 on Build GRCh38
rs750706683 Chr.14: 20367830-20367830 on Build GRCh38
rs143988875 Chr.14: 20367832-20367832 on Build GRCh38
rs1021970161 Chr.14: 20367678-20367678 on Build GRCh38
rs541631809 Chr.14: 20367814-20367814 on Build GRCh38

View Full Product Details