Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15617223-15617761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDI2
Entrez Gene ID
84301
Gene Name
DNA damage inducible 1 homolog 2
Gene Aliases
-
UniGene
Hs.718857
Gene Chromosome Location
Chr.1: 15617458-15661057 on Build GRCh38
Transcripts
NM_032341.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7429210 c.12C>G p.T4T Chr.1: 15617682-15617682 on Build GRCh38
5529625 c.40G>T p.E14* Chr.1: 15617710-15617710 on Build GRCh38
302034 c.76G>C p.E26Q Chr.1: 15617746-15617746 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs200371661 Chr.1: 15617271-15617272 on Build GRCh38
rs985090323 Chr.1: 15617357-15617357 on Build GRCh38
rs534720894 Chr.1: 15617360-15617360 on Build GRCh38
rs751857898 Chr.1: 15617648-15617648 on Build GRCh38
rs572126856 Chr.1: 15617353-15617353 on Build GRCh38
rs554792571 Chr.1: 15617397-15617397 on Build GRCh38
rs924343852 Chr.1: 15617465-15617465 on Build GRCh38
rs545357150 Chr.1: 15617474-15617474 on Build GRCh38
rs760434142 Chr.1: 15617630-15617630 on Build GRCh38
rs1013361625 Chr.1: 15617637-15617637 on Build GRCh38
rs1008814748 Chr.1: 15617514-15617514 on Build GRCh38
rs991431078 Chr.1: 15617432-15617432 on Build GRCh38
rs998975864 Chr.1: 15617430-15617430 on Build GRCh38
rs897902017 Chr.1: 15617577-15617577 on Build GRCh38
rs756354196 Chr.1: 15617657-15617657 on Build GRCh38
rs35499183 Chr.1: 15617400-15617401 on Build GRCh38
rs1030698820 Chr.1: 15617338-15617338 on Build GRCh38
rs774009806 Chr.1: 15617621-15617621 on Build GRCh38
rs765937495 Chr.1: 15617640-15617640 on Build GRCh38
rs574729955 Chr.1: 15617401-15617401 on Build GRCh38
rs1027937986 Chr.1: 15617423-15617423 on Build GRCh38
rs962442589 Chr.1: 15617419-15617419 on Build GRCh38
rs932233936 Chr.1: 15617487-15617487 on Build GRCh38
rs935786520 Chr.1: 15617469-15617469 on Build GRCh38
rs969344797 Chr.1: 15617340-15617340 on Build GRCh38
rs971020937 Chr.1: 15617440-15617440 on Build GRCh38
rs916950080 Chr.1: 15617276-15617276 on Build GRCh38
rs1005888655 Chr.1: 15617662-15617662 on Build GRCh38
rs757689930 Chr.1: 15617652-15617652 on Build GRCh38
rs947365605 Chr.1: 15617593-15617593 on Build GRCh38
rs543327560 Chr.1: 15617409-15617409 on Build GRCh38
rs764672662 Chr.1: 15617637-15617641 on Build GRCh38
rs527770876 Chr.1: 15617636-15617636 on Build GRCh38
rs371567113 Chr.1: 15617733-15617733 on Build GRCh38
rs761723037 Chr.1: 15617623-15617623 on Build GRCh38
rs538596464 Chr.1: 15617323-15617323 on Build GRCh38
rs753410323 Chr.1: 15617641-15617641 on Build GRCh38
rs1014928916 Chr.1: 15617413-15617413 on Build GRCh38
rs758883268 Chr.1: 15617636-15617637 on Build GRCh38
rs1028936734 Chr.1: 15617396-15617396 on Build GRCh38
rs1021764615 Chr.1: 15617263-15617263 on Build GRCh38
rs933436453 Chr.1: 15617489-15617489 on Build GRCh38
rs977090468 Chr.1: 15617453-15617453 on Build GRCh38
rs779166213 Chr.1: 15617655-15617656 on Build GRCh38
rs933364117 Chr.1: 15617393-15617394 on Build GRCh38
rs902946390 Chr.1: 15617306-15617306 on Build GRCh38
rs1036361798 Chr.1: 15617550-15617550 on Build GRCh38
rs765859109 Chr.1: 15617631-15617635 on Build GRCh38
rs138288509 Chr.1: 15617703-15617703 on Build GRCh38
rs757645610 Chr.1: 15617653-15617654 on Build GRCh38
rs772953101 Chr.1: 15617627-15617627 on Build GRCh38
rs55736480 Chr.1: 15617510-15617510 on Build GRCh38
rs781303453 Chr.1: 15617320-15617320 on Build GRCh38
rs781244808 Chr.1: 15617654-15617654 on Build GRCh38
rs772904667 Chr.1: 15617740-15617740 on Build GRCh38
rs1014494552 Chr.1: 15617392-15617411 on Build GRCh38
rs1050613051 Chr.1: 15617493-15617493 on Build GRCh38
rs557256023 Chr.1: 15617436-15617436 on Build GRCh38
rs1050458974 Chr.1: 15617408-15617408 on Build GRCh38
rs903447907 Chr.1: 15617596-15617596 on Build GRCh38
rs986600253 Chr.1: 15617504-15617504 on Build GRCh38
rs774392792 Chr.1: 15617704-15617704 on Build GRCh38
rs758906942 Chr.1: 15617642-15617642 on Build GRCh38
rs991165578 Chr.1: 15617257-15617257 on Build GRCh38
rs996865376 Chr.1: 15617346-15617346 on Build GRCh38
rs973259959 Chr.1: 15617422-15617422 on Build GRCh38
rs958763110 Chr.1: 15617634-15617634 on Build GRCh38
rs899471705 Chr.1: 15617586-15617586 on Build GRCh38
rs764730178 Chr.1: 15617647-15617647 on Build GRCh38
rs1054363983 Chr.1: 15617335-15617335 on Build GRCh38
rs747862615 Chr.1: 15617724-15617724 on Build GRCh38
rs564938807 Chr.1: 15617480-15617480 on Build GRCh38
rs541175373 Chr.1: 15617651-15617651 on Build GRCh38
rs768597124 Chr.1: 15617620-15617620 on Build GRCh38
rs753360708 Chr.1: 15617631-15617632 on Build GRCh38
rs1043328256 Chr.1: 15617594-15617594 on Build GRCh38
rs1053949964 Chr.1: 15617476-15617476 on Build GRCh38
rs200110210 Chr.1: 15617331-15617331 on Build GRCh38
rs958638304 Chr.1: 15617424-15617424 on Build GRCh38
rs780053363 Chr.1: 15617658-15617658 on Build GRCh38
rs746054311 Chr.1: 15617655-15617655 on Build GRCh38
rs376419303 Chr.1: 15617339-15617339 on Build GRCh38
rs577078237 Chr.1: 15617452-15617452 on Build GRCh38
rs1041465979 Chr.1: 15617289-15617289 on Build GRCh38
rs949766066 Chr.1: 15617282-15617282 on Build GRCh38
rs972178393 Chr.1: 15617391-15617391 on Build GRCh38
rs890716823 Chr.1: 15617512-15617512 on Build GRCh38
rs940494163 Chr.1: 15617378-15617378 on Build GRCh38
rs915422718 Chr.1: 15617478-15617478 on Build GRCh38
rs561125006 Chr.1: 15617653-15617653 on Build GRCh38
rs749434244 Chr.1: 15617665-15617665 on Build GRCh38

View Full Product Details