Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 84816599-84817109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALPK3
Entrez Gene ID
57538
Gene Name
alpha kinase 3
Gene Aliases
MAK, MIDORI
UniGene
Hs.459183
Gene Chromosome Location
Chr.15: 84816631-84873482 on Build GRCh38
Transcripts
NM_020778.4 XM_017022444.1 XM_017022445.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7216737 c.192C>A p.C64* Chr.15: 84817038-84817038 on Build GRCh38
1678642 c.97C>A p.L33I Chr.15: 84816943-84816943 on Build GRCh38
7207907 c.78G>T p.V26V Chr.15: 84816924-84816924 on Build GRCh38
7321458 c.218G>A p.R73Q Chr.15: 84817064-84817064 on Build GRCh38
5971558 c.242C>T p.T81M Chr.15: 84817088-84817088 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774883028 Chr.15: 84817043-84817043 on Build GRCh38
rs775720199 Chr.15: 84817030-84817030 on Build GRCh38
rs756797001 Chr.15: 84817022-84817022 on Build GRCh38
rs200966727 Chr.15: 84817088-84817088 on Build GRCh38
rs980013521 Chr.15: 84816990-84816990 on Build GRCh38
rs760305811 Chr.15: 84816871-84816871 on Build GRCh38
rs759153626 Chr.15: 84816949-84816949 on Build GRCh38
rs376798667 Chr.15: 84817044-84817044 on Build GRCh38
rs777873908 Chr.15: 84816810-84816810 on Build GRCh38
rs200319924 Chr.15: 84816827-84816827 on Build GRCh38
rs780059969 Chr.15: 84816831-84816831 on Build GRCh38
rs966924018 Chr.15: 84816784-84816784 on Build GRCh38
rs752536784 Chr.15: 84817009-84817009 on Build GRCh38
rs776990543 Chr.15: 84816883-84816883 on Build GRCh38
rs11857356 Chr.15: 84816870-84816870 on Build GRCh38
rs573092257 Chr.15: 84816771-84816771 on Build GRCh38
rs1055086679 Chr.15: 84816717-84816717 on Build GRCh38
rs761936977 Chr.15: 84816973-84816973 on Build GRCh38
rs957404836 Chr.15: 84816825-84816825 on Build GRCh38
rs753180388 Chr.15: 84816896-84816896 on Build GRCh38
rs28431354 Chr.15: 84817040-84817040 on Build GRCh38
rs758170095 Chr.15: 84817072-84817072 on Build GRCh38
rs771523068 Chr.15: 84816862-84816862 on Build GRCh38
rs770590590 Chr.15: 84816931-84816931 on Build GRCh38
rs934975014 Chr.15: 84816695-84816695 on Build GRCh38
rs139583474 Chr.15: 84816925-84816925 on Build GRCh38
rs764593970 Chr.15: 84816972-84816972 on Build GRCh38
rs779182898 Chr.15: 84816654-84816654 on Build GRCh38
rs745577314 Chr.15: 84817024-84817024 on Build GRCh38
rs368149305 Chr.15: 84816840-84816840 on Build GRCh38
rs1056573513 Chr.15: 84816703-84816703 on Build GRCh38
rs55736017 Chr.15: 84816773-84816773 on Build GRCh38
rs770568285 Chr.15: 84816882-84816882 on Build GRCh38
rs773855739 Chr.15: 84816824-84816824 on Build GRCh38
rs778946543 Chr.15: 84816851-84816851 on Build GRCh38
rs370679146 Chr.15: 84816817-84816817 on Build GRCh38
rs138870728 Chr.15: 84816894-84816894 on Build GRCh38
rs766559319 Chr.15: 84817064-84817064 on Build GRCh38
rs746463337 Chr.15: 84816928-84816928 on Build GRCh38
rs115479471 Chr.15: 84817038-84817038 on Build GRCh38
rs768729678 Chr.15: 84816841-84816841 on Build GRCh38
rs28408840 Chr.15: 84816855-84816855 on Build GRCh38
rs773416498 Chr.15: 84817053-84817053 on Build GRCh38
rs763763411 Chr.15: 84817076-84817076 on Build GRCh38
rs776222983 Chr.15: 84816946-84816946 on Build GRCh38
rs902271450 Chr.15: 84816664-84816664 on Build GRCh38
rs149320215 Chr.15: 84816951-84816951 on Build GRCh38
rs748089220 Chr.15: 84816924-84816924 on Build GRCh38
rs898171110 Chr.15: 84816708-84816708 on Build GRCh38
rs887586280 Chr.15: 84816791-84816791 on Build GRCh38
rs769431148 Chr.15: 84817029-84817029 on Build GRCh38
rs148968322 Chr.15: 84816882-84816883 on Build GRCh38
rs149818266 Chr.15: 84817069-84817069 on Build GRCh38
rs751652599 Chr.15: 84816907-84816907 on Build GRCh38
rs146005696 Chr.15: 84816921-84816921 on Build GRCh38
rs566770275 Chr.15: 84816720-84816720 on Build GRCh38
rs373770472 Chr.15: 84816917-84816917 on Build GRCh38
rs566259735 Chr.15: 84817001-84817001 on Build GRCh38
rs886880346 Chr.15: 84816644-84816644 on Build GRCh38
rs371628844 Chr.15: 84816992-84816992 on Build GRCh38
rs1001102638 Chr.15: 84817054-84817054 on Build GRCh38
rs532756173 Chr.15: 84816964-84816964 on Build GRCh38
rs750716031 Chr.15: 84816997-84816997 on Build GRCh38
rs866050590 Chr.15: 84817012-84817012 on Build GRCh38
rs775909774 Chr.15: 84816953-84816953 on Build GRCh38
rs868789582 Chr.15: 84816748-84816748 on Build GRCh38
rs753709710 Chr.15: 84816918-84816918 on Build GRCh38
rs28416295 Chr.15: 84817011-84817011 on Build GRCh38
rs758275836 Chr.15: 84817010-84817010 on Build GRCh38
rs778737671 Chr.15: 84816923-84816923 on Build GRCh38
rs761047932 Chr.15: 84817059-84817059 on Build GRCh38
rs528811019 Chr.15: 84817017-84817017 on Build GRCh38
rs896397393 Chr.15: 84816716-84816716 on Build GRCh38
rs115389253 Chr.15: 84817021-84817021 on Build GRCh38
rs1012200949 Chr.15: 84816737-84816737 on Build GRCh38
rs576650922 Chr.15: 84816643-84816643 on Build GRCh38
rs373853510 Chr.15: 84817000-84817000 on Build GRCh38
rs780654617 Chr.15: 84817023-84817023 on Build GRCh38
rs764136870 Chr.15: 84816904-84816904 on Build GRCh38
rs998964895 Chr.15: 84816792-84816792 on Build GRCh38
rs767754656 Chr.15: 84817045-84817045 on Build GRCh38
rs887695831 Chr.15: 84817083-84817083 on Build GRCh38
rs1056121343 Chr.15: 84817018-84817018 on Build GRCh38
rs545549693 Chr.15: 84816694-84816694 on Build GRCh38
rs748498340 Chr.15: 84816671-84816671 on Build GRCh38

View Full Product Details