Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 18804772-18805270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMG1
Entrez Gene ID
23049
Gene Name
SMG1, nonsense mediated mRNA decay associated PI3K related kinase
Gene Aliases
61E3.4, ATX, LIP
UniGene
Hs.460179
Gene Chromosome Location
Chr.16: 18804853-18926417 on Build GRCh38
Transcripts
NM_015092.4 XM_005255182.3 XM_005255183.3 XM_005255184.3 XM_011545769.2 XM_011545770.1 XM_017023066.1 XM_017023067.1

SNP Detail

SNP ID Chromosome Location
rs746468325 Chr.16: 18805043-18805043 on Build GRCh38
rs1011408005 Chr.16: 18804978-18804978 on Build GRCh38
rs1012376727 Chr.16: 18804851-18804851 on Build GRCh38
rs962252151 Chr.16: 18805186-18805186 on Build GRCh38
rs999803198 Chr.16: 18804921-18804921 on Build GRCh38
rs866568913 Chr.16: 18805165-18805165 on Build GRCh38
rs920252755 Chr.16: 18805227-18805227 on Build GRCh38
rs758745744 Chr.16: 18805056-18805056 on Build GRCh38
rs1045720459 Chr.16: 18804906-18804906 on Build GRCh38
rs987316369 Chr.16: 18805096-18805096 on Build GRCh38
rs779450948 Chr.16: 18805053-18805053 on Build GRCh38
rs949858147 Chr.16: 18804891-18804893 on Build GRCh38
rs1028069156 Chr.16: 18805157-18805160 on Build GRCh38
rs892779153 Chr.16: 18804951-18804951 on Build GRCh38
rs1055507612 Chr.16: 18804797-18804797 on Build GRCh38
rs568916885 Chr.16: 18805210-18805210 on Build GRCh38
rs983977998 Chr.16: 18805222-18805222 on Build GRCh38
rs781541797 Chr.16: 18805056-18805057 on Build GRCh38
rs757909658 Chr.16: 18805054-18805056 on Build GRCh38
rs1019182705 Chr.16: 18805071-18805071 on Build GRCh38
rs370362108 Chr.16: 18805034-18805034 on Build GRCh38
rs536134713 Chr.16: 18805176-18805176 on Build GRCh38
rs906795816 Chr.16: 18804919-18804919 on Build GRCh38
rs927178146 Chr.16: 18804918-18804918 on Build GRCh38
rs895278687 Chr.16: 18804804-18804804 on Build GRCh38
rs777290313 Chr.16: 18804932-18804932 on Build GRCh38
rs1010409543 Chr.16: 18804991-18804991 on Build GRCh38
rs1029086893 Chr.16: 18804979-18804979 on Build GRCh38
rs754230509 Chr.16: 18804826-18804826 on Build GRCh38
rs958100574 Chr.16: 18804949-18804949 on Build GRCh38
rs996359503 Chr.16: 18805123-18805123 on Build GRCh38
rs746401023 Chr.16: 18805055-18805055 on Build GRCh38
rs1046602301 Chr.16: 18804890-18804890 on Build GRCh38

View Full Product Details