Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 18806659-18807166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMG1
Entrez Gene ID
23049
Gene Name
SMG1, nonsense mediated mRNA decay associated PI3K related kinase
Gene Aliases
61E3.4, ATX, LIP
UniGene
Hs.460179
Gene Chromosome Location
Chr.16: 18804853-18926417 on Build GRCh38
Transcripts
NM_015092.4 XM_005255182.3 XM_005255183.3 XM_005255184.3 XM_011545769.2 XM_011545770.1 XM_017023066.1 XM_017023067.1

SNP Detail

SNP ID Chromosome Location
rs768631791 Chr.16: 18806827-18806827 on Build GRCh38
rs888159118 Chr.16: 18806909-18806909 on Build GRCh38
rs991778356 Chr.16: 18807031-18807031 on Build GRCh38
rs558292753 Chr.16: 18806769-18806769 on Build GRCh38
rs539874251 Chr.16: 18806763-18806763 on Build GRCh38
rs1011670959 Chr.16: 18807067-18807067 on Build GRCh38
rs979185962 Chr.16: 18807132-18807132 on Build GRCh38
rs934750467 Chr.16: 18807142-18807142 on Build GRCh38
rs938605016 Chr.16: 18806984-18806984 on Build GRCh38
rs907075180 Chr.16: 18806978-18806978 on Build GRCh38
rs2606559 Chr.16: 18806955-18806955 on Build GRCh38
rs374811698 Chr.16: 18807092-18807092 on Build GRCh38
rs1044192647 Chr.16: 18806968-18806968 on Build GRCh38
rs910747247 Chr.16: 18806783-18806783 on Build GRCh38
rs972237356 Chr.16: 18806686-18806686 on Build GRCh38
rs900974794 Chr.16: 18806884-18806884 on Build GRCh38
rs528387764 Chr.16: 18807011-18807011 on Build GRCh38
rs575645794 Chr.16: 18806704-18806704 on Build GRCh38
rs968179423 Chr.16: 18806751-18806751 on Build GRCh38
rs1021388260 Chr.16: 18806731-18806731 on Build GRCh38
rs1056021859 Chr.16: 18806762-18806762 on Build GRCh38
rs976772897 Chr.16: 18806802-18806802 on Build GRCh38
rs1028651267 Chr.16: 18806898-18806898 on Build GRCh38
rs891422835 Chr.16: 18807014-18807014 on Build GRCh38
rs981817946 Chr.16: 18806964-18806964 on Build GRCh38
rs190646438 Chr.16: 18807000-18807000 on Build GRCh38
rs916129222 Chr.16: 18807036-18807036 on Build GRCh38
rs1035866567 Chr.16: 18806996-18806996 on Build GRCh38
rs929299540 Chr.16: 18806856-18806856 on Build GRCh38
rs573439991 Chr.16: 18806826-18806826 on Build GRCh38
rs919259483 Chr.16: 18806847-18806847 on Build GRCh38
rs777924199 Chr.16: 18806885-18806885 on Build GRCh38
rs1011324110 Chr.16: 18806729-18806729 on Build GRCh38
rs778704126 Chr.16: 18806916-18806916 on Build GRCh38
rs573992424 Chr.16: 18806770-18806773 on Build GRCh38
rs1015230574 Chr.16: 18806935-18806935 on Build GRCh38
rs1043173273 Chr.16: 18806808-18806809 on Build GRCh38
rs889521179 Chr.16: 18807130-18807130 on Build GRCh38
rs960185182 Chr.16: 18807001-18807001 on Build GRCh38
rs1005616618 Chr.16: 18806931-18806931 on Build GRCh38
rs749049382 Chr.16: 18806732-18806732 on Build GRCh38
rs1040877147 Chr.16: 18806863-18806864 on Build GRCh38
rs966306013 Chr.16: 18806797-18806797 on Build GRCh38
rs540849219 Chr.16: 18806848-18806848 on Build GRCh38
rs924613438 Chr.16: 18807135-18807135 on Build GRCh38
rs1021360418 Chr.16: 18807079-18807079 on Build GRCh38

View Full Product Details