Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20781626-20782128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM3
Entrez Gene ID
6296
Gene Name
acyl-CoA synthetase medium-chain family member 3
Gene Aliases
SA, SAH
UniGene
Hs.706754
Gene Chromosome Location
Chr.16: 20763990-20797157 on Build GRCh38
Transcripts
NM_005622.3 NM_202000.2 XM_011545911.1 XM_017023523.1 XM_017023524.1 NM_080663.2

Gene Symbol
ERI2
Entrez Gene ID
112479
Gene Name
ERI1 exoribonuclease family member 2
Gene Aliases
EXOD1, ZGRF5
UniGene
Hs.248437
Gene Chromosome Location
Chr.16: 20780193-20806473 on Build GRCh38
Transcripts
NM_005622.3 NM_202000.2 XM_011545911.1 XM_017023523.1 XM_017023524.1 NM_080663.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1580412 c.994A>G p.M332V Chr.16: 20781762-20781762 on Build GRCh38
3507064 c.1005G>A p.Q335Q Chr.16: 20781773-20781773 on Build GRCh38
238668 c.991C>T p.R331* Chr.16: 20781759-20781759 on Build GRCh38
4513933 c.956C>T p.P319L Chr.16: 20781724-20781724 on Build GRCh38
3507063 c.1005G>A p.Q335Q Chr.16: 20781773-20781773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142925231 Chr.16: 20781656-20781656 on Build GRCh38
rs781750943 Chr.16: 20781780-20781780 on Build GRCh38
rs1026728403 Chr.16: 20781882-20781882 on Build GRCh38
rs759079778 Chr.16: 20781803-20781804 on Build GRCh38
rs758277289 Chr.16: 20781723-20781723 on Build GRCh38
rs977966298 Chr.16: 20781713-20781713 on Build GRCh38
rs896807425 Chr.16: 20781808-20781808 on Build GRCh38
rs970880577 Chr.16: 20781904-20781904 on Build GRCh38
rs769972281 Chr.16: 20781730-20781730 on Build GRCh38
rs777832332 Chr.16: 20781732-20781732 on Build GRCh38
rs915325777 Chr.16: 20782085-20782085 on Build GRCh38
rs138146688 Chr.16: 20781995-20781995 on Build GRCh38
rs13306604 Chr.16: 20781807-20781807 on Build GRCh38
rs749561403 Chr.16: 20781743-20781743 on Build GRCh38
rs773894016 Chr.16: 20781756-20781773 on Build GRCh38
rs989472175 Chr.16: 20782038-20782038 on Build GRCh38
rs750243791 Chr.16: 20781722-20781722 on Build GRCh38
rs760264524 Chr.16: 20781672-20781672 on Build GRCh38
rs762728134 Chr.16: 20781659-20781660 on Build GRCh38
rs765787575 Chr.16: 20781661-20781662 on Build GRCh38
rs189867767 Chr.16: 20781758-20781758 on Build GRCh38
rs952610592 Chr.16: 20781886-20781886 on Build GRCh38
rs547166655 Chr.16: 20781969-20781969 on Build GRCh38
rs756528643 Chr.16: 20781901-20781901 on Build GRCh38
rs199802576 Chr.16: 20781784-20781784 on Build GRCh38
rs1012002805 Chr.16: 20781895-20781895 on Build GRCh38
rs779973608 Chr.16: 20781724-20781724 on Build GRCh38
rs944527565 Chr.16: 20781753-20781753 on Build GRCh38
rs1034884592 Chr.16: 20782004-20782004 on Build GRCh38
rs915339165 Chr.16: 20781671-20781671 on Build GRCh38
rs200477998 Chr.16: 20781759-20781759 on Build GRCh38
rs147005302 Chr.16: 20781913-20781913 on Build GRCh38
rs938088696 Chr.16: 20781878-20781878 on Build GRCh38
rs778729502 Chr.16: 20781719-20781719 on Build GRCh38
rs192560104 Chr.16: 20781762-20781762 on Build GRCh38
rs756970657 Chr.16: 20781697-20781697 on Build GRCh38
rs924216989 Chr.16: 20781933-20781933 on Build GRCh38
rs763700746 Chr.16: 20781810-20781810 on Build GRCh38
rs748225553 Chr.16: 20781729-20781729 on Build GRCh38
rs753321212 Chr.16: 20781824-20781824 on Build GRCh38
rs377065310 Chr.16: 20781786-20781786 on Build GRCh38
rs911747422 Chr.16: 20781727-20781727 on Build GRCh38
rs185451727 Chr.16: 20781982-20781982 on Build GRCh38
rs560395088 Chr.16: 20781663-20781663 on Build GRCh38
rs753578255 Chr.16: 20781692-20781692 on Build GRCh38
rs267604443 Chr.16: 20781760-20781760 on Build GRCh38
rs763641191 Chr.16: 20781682-20781682 on Build GRCh38
rs879013534 Chr.16: 20781919-20781919 on Build GRCh38

View Full Product Details