Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67885619-67886109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NRN1L
Entrez Gene ID
123904
Gene Name
neuritin 1 like
Gene Aliases
UNQ2446
UniGene
Hs.435464
Gene Chromosome Location
Chr.16: 67884878-67886368 on Build GRCh38
Transcripts
NM_198443.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
343236 c.121C>T p.R41* Chr.16: 67885763-67885763 on Build GRCh38
7348600 c.264G>A p.P88P Chr.16: 67886025-67886025 on Build GRCh38
2995837 c.215C>A p.S72Y Chr.16: 67885976-67885976 on Build GRCh38
972613 c.193G>A p.E65K Chr.16: 67885835-67885835 on Build GRCh38
3818303 c.340C>T p.L114L Chr.16: 67886101-67886101 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755560871 Chr.16: 67885826-67885826 on Build GRCh38
rs754327699 Chr.16: 67885821-67885821 on Build GRCh38
rs368368599 Chr.16: 67885752-67885752 on Build GRCh38
rs760313001 Chr.16: 67885973-67885973 on Build GRCh38
rs925048438 Chr.16: 67886031-67886031 on Build GRCh38
rs781276209 Chr.16: 67885899-67885899 on Build GRCh38
rs984636293 Chr.16: 67885951-67885951 on Build GRCh38
rs115666575 Chr.16: 67885753-67885753 on Build GRCh38
rs374302021 Chr.16: 67886083-67886083 on Build GRCh38
rs371372021 Chr.16: 67885946-67885946 on Build GRCh38
rs766524013 Chr.16: 67885818-67885826 on Build GRCh38
rs755480178 Chr.16: 67885868-67885868 on Build GRCh38
rs748027241 Chr.16: 67886042-67886042 on Build GRCh38
rs759005964 Chr.16: 67885987-67885987 on Build GRCh38
rs369746345 Chr.16: 67885867-67885867 on Build GRCh38
rs763715572 Chr.16: 67885717-67885717 on Build GRCh38
rs539741822 Chr.16: 67885661-67885661 on Build GRCh38
rs759179404 Chr.16: 67885978-67885978 on Build GRCh38
rs956554635 Chr.16: 67885997-67885997 on Build GRCh38
rs774883844 Chr.16: 67885764-67885764 on Build GRCh38
rs373144527 Chr.16: 67885894-67885894 on Build GRCh38
rs114883420 Chr.16: 67886084-67886084 on Build GRCh38
rs761950965 Chr.16: 67885704-67885704 on Build GRCh38
rs760274823 Chr.16: 67885865-67885865 on Build GRCh38
rs765861674 Chr.16: 67885866-67885866 on Build GRCh38
rs770608050 Chr.16: 67886071-67886071 on Build GRCh38
rs773363288 Chr.16: 67885780-67885780 on Build GRCh38
rs775650340 Chr.16: 67885681-67885681 on Build GRCh38
rs777828496 Chr.16: 67885884-67885884 on Build GRCh38
rs762900458 Chr.16: 67885831-67885831 on Build GRCh38
rs922233989 Chr.16: 67886054-67886054 on Build GRCh38
rs572731480 Chr.16: 67885640-67885640 on Build GRCh38
rs771913331 Chr.16: 67886047-67886047 on Build GRCh38
rs139776238 Chr.16: 67885793-67885793 on Build GRCh38
rs550319897 Chr.16: 67885919-67885919 on Build GRCh38
rs187638337 Chr.16: 67886085-67886085 on Build GRCh38
rs753300370 Chr.16: 67885986-67885986 on Build GRCh38
rs150898109 Chr.16: 67885792-67885792 on Build GRCh38
rs1005764997 Chr.16: 67885648-67885648 on Build GRCh38
rs907740601 Chr.16: 67885965-67885965 on Build GRCh38
rs201603662 Chr.16: 67885776-67885776 on Build GRCh38
rs774761274 Chr.16: 67885703-67885703 on Build GRCh38
rs370856682 Chr.16: 67885763-67885763 on Build GRCh38
rs764473223 Chr.16: 67886001-67886001 on Build GRCh38
rs969330688 Chr.16: 67885909-67885909 on Build GRCh38
rs141270296 Chr.16: 67886003-67886003 on Build GRCh38
rs144361985 Chr.16: 67885827-67885827 on Build GRCh38
rs770346589 Chr.16: 67885674-67885674 on Build GRCh38
rs761548781 Chr.16: 67885714-67885732 on Build GRCh38
rs752434126 Chr.16: 67885714-67885714 on Build GRCh38
rs998910850 Chr.16: 67885942-67885942 on Build GRCh38
rs776378679 Chr.16: 67885709-67885709 on Build GRCh38
rs751379417 Chr.16: 67885801-67885801 on Build GRCh38
rs936446161 Chr.16: 67886063-67886063 on Build GRCh38
rs142084682 Chr.16: 67885790-67885790 on Build GRCh38
rs779634477 Chr.16: 67885879-67885879 on Build GRCh38
rs768584826 Chr.16: 67885834-67885834 on Build GRCh38
rs183334989 Chr.16: 67885828-67885828 on Build GRCh38
rs761653018 Chr.16: 67885859-67885859 on Build GRCh38
rs764514724 Chr.16: 67886082-67886082 on Build GRCh38
rs200752015 Chr.16: 67886080-67886080 on Build GRCh38
rs772903199 Chr.16: 67885962-67885962 on Build GRCh38
rs771811015 Chr.16: 67885953-67885953 on Build GRCh38
rs765819536 Chr.16: 67885979-67885979 on Build GRCh38
rs774568142 Chr.16: 67885847-67885847 on Build GRCh38
rs759602582 Chr.16: 67886075-67886075 on Build GRCh38
rs756936016 Chr.16: 67885750-67885750 on Build GRCh38
rs201170067 Chr.16: 67885863-67885863 on Build GRCh38
rs763496028 Chr.16: 67886081-67886081 on Build GRCh38
rs751253028 Chr.16: 67885747-67885747 on Build GRCh38
rs753101913 Chr.16: 67885880-67885880 on Build GRCh38
rs773444821 Chr.16: 67885708-67885708 on Build GRCh38
rs757495434 Chr.16: 67886010-67886010 on Build GRCh38
rs1013193053 Chr.16: 67885915-67885915 on Build GRCh38
rs760878086 Chr.16: 67885789-67885789 on Build GRCh38
rs370251298 Chr.16: 67885941-67885941 on Build GRCh38
rs920016807 Chr.16: 67885667-67885667 on Build GRCh38
rs146612935 Chr.16: 67885830-67885830 on Build GRCh38
rs558526728 Chr.16: 67885705-67885705 on Build GRCh38
rs376322030 Chr.16: 67885896-67885896 on Build GRCh38
rs746850390 Chr.16: 67886061-67886061 on Build GRCh38
rs35880060 Chr.16: 67885687-67885688 on Build GRCh38
rs564512461 Chr.16: 67885851-67885851 on Build GRCh38
rs767624331 Chr.16: 67886074-67886075 on Build GRCh38
rs562219500 Chr.16: 67885762-67885762 on Build GRCh38
rs975974316 Chr.16: 67885998-67885998 on Build GRCh38
rs766267987 Chr.16: 67885949-67885949 on Build GRCh38
rs371968237 Chr.16: 67885805-67885805 on Build GRCh38
rs749727808 Chr.16: 67885697-67885697 on Build GRCh38
rs780136416 Chr.16: 67886037-67886037 on Build GRCh38
rs747040669 Chr.16: 67885829-67885829 on Build GRCh38
rs781327123 Chr.16: 67886024-67886024 on Build GRCh38
rs1013814767 Chr.16: 67885893-67885893 on Build GRCh38
rs370621251 Chr.16: 67885709-67885709 on Build GRCh38
rs768745437 Chr.16: 67885698-67885698 on Build GRCh38
rs767224452 Chr.16: 67885806-67885806 on Build GRCh38
rs372327387 Chr.16: 67885860-67885860 on Build GRCh38
rs780391880 Chr.16: 67885813-67885813 on Build GRCh38
rs768488408 Chr.16: 67885703-67885703 on Build GRCh38
rs149800685 Chr.16: 67886025-67886025 on Build GRCh38
rs765948716 Chr.16: 67885974-67885974 on Build GRCh38
rs73593844 Chr.16: 67886027-67886027 on Build GRCh38
rs369400959 Chr.16: 67885931-67885931 on Build GRCh38
rs778399173 Chr.16: 67886053-67886053 on Build GRCh38
rs748328780 Chr.16: 67885766-67885766 on Build GRCh38
rs201561757 Chr.16: 67886078-67886078 on Build GRCh38
rs758274434 Chr.16: 67885716-67885716 on Build GRCh38

View Full Product Details