Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7854512-7855003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM6B
Entrez Gene ID
23135
Gene Name
lysine demethylase 6B
Gene Aliases
JMJD3
UniGene
Hs.223678
Gene Chromosome Location
Chr.17: 7834210-7854800 on Build GRCh38
Transcripts
NM_001080424.1 XM_005256549.3 XM_005256550.4 XM_005256551.3 XM_005256552.4 XM_006721483.3 XM_011523750.2 XM_011523752.2 XM_017024381.1 XM_005256856.3

Gene Symbol
TMEM88
Entrez Gene ID
92162
Gene Name
transmembrane protein 88
Gene Aliases
-
UniGene
Hs.389669
Gene Chromosome Location
Chr.17: 7854886-7856099 on Build GRCh38
Transcripts
NM_001080424.1 XM_005256549.3 XM_005256550.4 XM_005256551.3 XM_005256552.4 XM_006721483.3 XM_011523750.2 XM_011523752.2 XM_017024381.1 XM_005256856.3

SNP Detail

SNP ID Chromosome Location
rs4791855 Chr.17: 7854550-7854550 on Build GRCh38
rs981273114 Chr.17: 7854630-7854630 on Build GRCh38
rs898658883 Chr.17: 7854566-7854566 on Build GRCh38
rs550093679 Chr.17: 7854769-7854769 on Build GRCh38
rs573936345 Chr.17: 7854796-7854796 on Build GRCh38
rs190030774 Chr.17: 7854811-7854811 on Build GRCh38
rs969890135 Chr.17: 7854587-7854587 on Build GRCh38
rs1049315571 Chr.17: 7854698-7854698 on Build GRCh38
rs1012038050 Chr.17: 7854641-7854641 on Build GRCh38
rs930420967 Chr.17: 7854567-7854567 on Build GRCh38
rs939752765 Chr.17: 7854652-7854652 on Build GRCh38
rs961498650 Chr.17: 7854939-7854939 on Build GRCh38
rs919631733 Chr.17: 7854690-7854690 on Build GRCh38
rs1012152890 Chr.17: 7854912-7854912 on Build GRCh38
rs1040511706 Chr.17: 7854741-7854741 on Build GRCh38
rs896373344 Chr.17: 7854635-7854635 on Build GRCh38
rs578223263 Chr.17: 7854860-7854860 on Build GRCh38
rs967899844 Chr.17: 7854659-7854659 on Build GRCh38
rs943670616 Chr.17: 7854973-7854973 on Build GRCh38
rs371945892 Chr.17: 7854627-7854627 on Build GRCh38
rs751631671 Chr.17: 7854712-7854712 on Build GRCh38
rs1049346213 Chr.17: 7854949-7854949 on Build GRCh38
rs1011553040 Chr.17: 7854789-7854789 on Build GRCh38
rs896153051 Chr.17: 7854890-7854890 on Build GRCh38
rs972531771 Chr.17: 7854940-7854940 on Build GRCh38
rs556685397 Chr.17: 7854836-7854836 on Build GRCh38
rs957388248 Chr.17: 7854788-7854789 on Build GRCh38
rs1022904510 Chr.17: 7854798-7854798 on Build GRCh38
rs754964115 Chr.17: 7854756-7854756 on Build GRCh38
rs919649605 Chr.17: 7854945-7854945 on Build GRCh38
rs574707413 Chr.17: 7854844-7854870 on Build GRCh38
rs766375767 Chr.17: 7854676-7854676 on Build GRCh38
rs886159642 Chr.17: 7854875-7854875 on Build GRCh38
rs1008144152 Chr.17: 7854739-7854739 on Build GRCh38
rs542932896 Chr.17: 7854976-7854976 on Build GRCh38
rs1037180680 Chr.17: 7854665-7854665 on Build GRCh38
rs975652037 Chr.17: 7854814-7854814 on Build GRCh38
rs986772231 Chr.17: 7854727-7854727 on Build GRCh38
rs1063746 Chr.17: 7854701-7854701 on Build GRCh38
rs928520959 Chr.17: 7854917-7854917 on Build GRCh38
rs902003256 Chr.17: 7854744-7854744 on Build GRCh38
rs998991081 Chr.17: 7854772-7854772 on Build GRCh38
rs1039864045 Chr.17: 7854557-7854557 on Build GRCh38
rs565101556 Chr.17: 7854951-7854951 on Build GRCh38
rs886126069 Chr.17: 7854583-7854583 on Build GRCh38
rs1048788671 Chr.17: 7854573-7854573 on Build GRCh38
rs781084062 Chr.17: 7854938-7854938 on Build GRCh38
rs915815816 Chr.17: 7854571-7854571 on Build GRCh38
rs566713372 Chr.17: 7854640-7854640 on Build GRCh38
rs942861900 Chr.17: 7854551-7854551 on Build GRCh38
rs920115178 Chr.17: 7854822-7854822 on Build GRCh38
rs940369052 Chr.17: 7854883-7854883 on Build GRCh38
rs545570766 Chr.17: 7854889-7854889 on Build GRCh38
rs931000434 Chr.17: 7854947-7854947 on Build GRCh38
rs912828422 Chr.17: 7854729-7854729 on Build GRCh38
rs1031744504 Chr.17: 7854784-7854784 on Build GRCh38
rs1023929110 Chr.17: 7854653-7854653 on Build GRCh38
rs931479072 Chr.17: 7854834-7854834 on Build GRCh38
rs572662562 Chr.17: 7854972-7854972 on Build GRCh38
rs1031067435 Chr.17: 7854677-7854677 on Build GRCh38
rs1034911754 Chr.17: 7854634-7854634 on Build GRCh38
rs533962815 Chr.17: 7854695-7854695 on Build GRCh38

View Full Product Details