Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7854959-7855452 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM88
Entrez Gene ID
92162
Gene Name
transmembrane protein 88
Gene Aliases
-
UniGene
Hs.389669
Gene Chromosome Location
Chr.17: 7854886-7856099 on Build GRCh38
Transcripts
NM_001319941.1 NM_203411.1 XM_005256856.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6253362 c.218G>T p.R73L Chr.17: 7855452-7855452 on Build GRCh38
985390 c.6G>A p.A2A Chr.17: 7855080-7855080 on Build GRCh38
6350800 c.130C>T p.L44F Chr.17: 7855204-7855204 on Build GRCh38
3890606 c.22C>G p.Q8E Chr.17: 7855096-7855096 on Build GRCh38
6021522 c.58G>A p.D20N Chr.17: 7855132-7855132 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771583051 Chr.17: 7855129-7855129 on Build GRCh38
rs763924463 Chr.17: 7855088-7855088 on Build GRCh38
rs758637840 Chr.17: 7855414-7855414 on Build GRCh38
rs369352680 Chr.17: 7855030-7855030 on Build GRCh38
rs866071380 Chr.17: 7855243-7855243 on Build GRCh38
rs376644922 Chr.17: 7855042-7855042 on Build GRCh38
rs755893018 Chr.17: 7855028-7855028 on Build GRCh38
rs760853465 Chr.17: 7855330-7855330 on Build GRCh38
rs373749404 Chr.17: 7855275-7855275 on Build GRCh38
rs1041026857 Chr.17: 7854984-7854984 on Build GRCh38
rs200343296 Chr.17: 7855257-7855257 on Build GRCh38
rs760359891 Chr.17: 7855132-7855132 on Build GRCh38
rs778195848 Chr.17: 7855108-7855108 on Build GRCh38
rs757978270 Chr.17: 7855303-7855308 on Build GRCh38
rs765288258 Chr.17: 7855158-7855158 on Build GRCh38
rs752835650 Chr.17: 7855242-7855242 on Build GRCh38
rs960184890 Chr.17: 7855347-7855347 on Build GRCh38
rs749589269 Chr.17: 7855116-7855116 on Build GRCh38
rs751978653 Chr.17: 7855418-7855418 on Build GRCh38
rs757670105 Chr.17: 7855263-7855263 on Build GRCh38
rs199705559 Chr.17: 7855113-7855113 on Build GRCh38
rs374435489 Chr.17: 7855394-7855394 on Build GRCh38
rs947707303 Chr.17: 7855197-7855197 on Build GRCh38
rs750809308 Chr.17: 7855406-7855406 on Build GRCh38
rs940483683 Chr.17: 7855137-7855137 on Build GRCh38
rs972517684 Chr.17: 7855051-7855051 on Build GRCh38
rs777518917 Chr.17: 7855430-7855430 on Build GRCh38
rs780845598 Chr.17: 7855425-7855425 on Build GRCh38
rs770494733 Chr.17: 7855069-7855069 on Build GRCh38
rs369865793 Chr.17: 7855072-7855072 on Build GRCh38
rs893230877 Chr.17: 7855194-7855194 on Build GRCh38
rs765374753 Chr.17: 7855402-7855402 on Build GRCh38
rs774794839 Chr.17: 7855125-7855125 on Build GRCh38
rs752930970 Chr.17: 7855176-7855176 on Build GRCh38
rs768364742 Chr.17: 7855225-7855225 on Build GRCh38
rs201354502 Chr.17: 7855214-7855214 on Build GRCh38
rs572778104 Chr.17: 7855360-7855362 on Build GRCh38
rs905913110 Chr.17: 7855359-7855359 on Build GRCh38
rs757251579 Chr.17: 7855092-7855092 on Build GRCh38
rs1019690473 Chr.17: 7855287-7855287 on Build GRCh38
rs893201081 Chr.17: 7854995-7854995 on Build GRCh38
rs758597691 Chr.17: 7855032-7855032 on Build GRCh38
rs117866774 Chr.17: 7855040-7855040 on Build GRCh38
rs368549452 Chr.17: 7855333-7855333 on Build GRCh38
rs769339123 Chr.17: 7855309-7855314 on Build GRCh38
rs188022759 Chr.17: 7855365-7855365 on Build GRCh38
rs1008342892 Chr.17: 7855276-7855276 on Build GRCh38
rs766163605 Chr.17: 7855238-7855238 on Build GRCh38
rs780051854 Chr.17: 7855207-7855208 on Build GRCh38
rs1022935796 Chr.17: 7855021-7855021 on Build GRCh38
rs1053205637 Chr.17: 7854991-7854991 on Build GRCh38
rs770154202 Chr.17: 7855050-7855050 on Build GRCh38
rs367604233 Chr.17: 7855320-7855320 on Build GRCh38
rs754582188 Chr.17: 7855423-7855423 on Build GRCh38
rs773342716 Chr.17: 7855231-7855231 on Build GRCh38
rs991894200 Chr.17: 7855360-7855360 on Build GRCh38
rs139302449 Chr.17: 7855087-7855087 on Build GRCh38
rs139379009 Chr.17: 7855165-7855165 on Build GRCh38
rs1011584591 Chr.17: 7854997-7854997 on Build GRCh38
rs375966206 Chr.17: 7855295-7855295 on Build GRCh38
rs976392530 Chr.17: 7855110-7855110 on Build GRCh38
rs116077752 Chr.17: 7855131-7855131 on Build GRCh38
rs1048284078 Chr.17: 7855398-7855398 on Build GRCh38
rs865982510 Chr.17: 7855109-7855109 on Build GRCh38
rs748481708 Chr.17: 7855048-7855048 on Build GRCh38
rs776568442 Chr.17: 7855136-7855136 on Build GRCh38
rs767622312 Chr.17: 7855175-7855175 on Build GRCh38
rs139351118 Chr.17: 7855180-7855180 on Build GRCh38
rs1040957207 Chr.17: 7855049-7855063 on Build GRCh38
rs878898998 Chr.17: 7855173-7855173 on Build GRCh38
rs1034266427 Chr.17: 7855016-7855016 on Build GRCh38
rs754876624 Chr.17: 7855300-7855301 on Build GRCh38
rs1019657990 Chr.17: 7855079-7855079 on Build GRCh38
rs756496488 Chr.17: 7855106-7855106 on Build GRCh38
rs148728644 Chr.17: 7855270-7855270 on Build GRCh38
rs76623612 Chr.17: 7855314-7855314 on Build GRCh38
rs771445035 Chr.17: 7855117-7855117 on Build GRCh38
rs752452519 Chr.17: 7855024-7855024 on Build GRCh38
rs539248262 Chr.17: 7855220-7855220 on Build GRCh38
rs548985960 Chr.17: 7855152-7855152 on Build GRCh38
rs765507835 Chr.17: 7855045-7855046 on Build GRCh38
rs556737650 Chr.17: 7855315-7855316 on Build GRCh38
rs750724261 Chr.17: 7855095-7855096 on Build GRCh38
rs146914626 Chr.17: 7855086-7855086 on Build GRCh38
rs758263256 Chr.17: 7855204-7855206 on Build GRCh38
rs746550999 Chr.17: 7855283-7855283 on Build GRCh38
rs144286158 Chr.17: 7855246-7855246 on Build GRCh38
rs556697420 Chr.17: 7855212-7855212 on Build GRCh38
rs763858146 Chr.17: 7855135-7855135 on Build GRCh38
rs532965366 Chr.17: 7855055-7855055 on Build GRCh38
rs755845109 Chr.17: 7855429-7855429 on Build GRCh38
rs543355484 Chr.17: 7855264-7855264 on Build GRCh38
rs762945512 Chr.17: 7855237-7855237 on Build GRCh38
rs746355319 Chr.17: 7855215-7855215 on Build GRCh38
rs983435750 Chr.17: 7855329-7855329 on Build GRCh38
rs532365962 Chr.17: 7855093-7855093 on Build GRCh38
rs761730196 Chr.17: 7855228-7855228 on Build GRCh38
rs764335434 Chr.17: 7855251-7855251 on Build GRCh38
rs772815673 Chr.17: 7855311-7855314 on Build GRCh38
rs543268223 Chr.17: 7854990-7854990 on Build GRCh38
rs2270518 Chr.17: 7855204-7855204 on Build GRCh38
rs908748733 Chr.17: 7855345-7855345 on Build GRCh38
rs747298623 Chr.17: 7855039-7855039 on Build GRCh38
rs767101731 Chr.17: 7855074-7855074 on Build GRCh38
rs370531527 Chr.17: 7855308-7855308 on Build GRCh38
rs757582087 Chr.17: 7855206-7855206 on Build GRCh38
rs1063651 Chr.17: 7855141-7855141 on Build GRCh38
rs761642438 Chr.17: 7855146-7855146 on Build GRCh38
rs753775517 Chr.17: 7855031-7855031 on Build GRCh38
rs746791446 Chr.17: 7855300-7855300 on Build GRCh38
rs769680290 Chr.17: 7855229-7855229 on Build GRCh38
rs560324633 Chr.17: 7855393-7855393 on Build GRCh38
rs771537367 Chr.17: 7855216-7855216 on Build GRCh38
rs755057447 Chr.17: 7855099-7855099 on Build GRCh38
rs768011451 Chr.17: 7855284-7855284 on Build GRCh38
rs8065467 Chr.17: 7855428-7855428 on Build GRCh38
rs747778724 Chr.17: 7855291-7855291 on Build GRCh38
rs745416289 Chr.17: 7855127-7855127 on Build GRCh38
rs746433620 Chr.17: 7855222-7855222 on Build GRCh38
rs35535635 Chr.17: 7855089-7855090 on Build GRCh38
rs766939680 Chr.17: 7855415-7855415 on Build GRCh38
rs750992347 Chr.17: 7855269-7855269 on Build GRCh38
rs781019746 Chr.17: 7855302-7855303 on Build GRCh38
rs770924688 Chr.17: 7855309-7855309 on Build GRCh38
rs781300418 Chr.17: 7855100-7855100 on Build GRCh38
rs781029509 Chr.17: 7855290-7855290 on Build GRCh38
rs373234979 Chr.17: 7855103-7855103 on Build GRCh38
rs775171120 Chr.17: 7855081-7855081 on Build GRCh38
rs550304927 Chr.17: 7855015-7855015 on Build GRCh38
rs773148056 Chr.17: 7855303-7855303 on Build GRCh38
rs780264722 Chr.17: 7855034-7855034 on Build GRCh38
rs947738488 Chr.17: 7855375-7855375 on Build GRCh38

View Full Product Details