Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 33088685-33089086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASIC2
Entrez Gene ID
40
Gene Name
acid sensing ion channel subunit 2
Gene Aliases
ACCN, ACCN1, ASIC2a, BNC1, BNaC1, MDEG, hBNaC1
UniGene
Hs.368417
Gene Chromosome Location
Chr.17: 33013087-34156806 on Build GRCh38
Transcripts
NM_001094.4 NM_183377.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1710133 c.892C>T p.Q298* Chr.17: 33088958-33088958 on Build GRCh38
6016569 c.928G>A p.E310K Chr.17: 33088922-33088922 on Build GRCh38
3516029 c.957C>T p.F319F Chr.17: 33088893-33088893 on Build GRCh38
3516031 c.949C>T p.P317S Chr.17: 33088901-33088901 on Build GRCh38
3516033 c.921C>T p.F307F Chr.17: 33088929-33088929 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1052576544 Chr.17: 33088736-33088736 on Build GRCh38
rs574398818 Chr.17: 33088725-33088725 on Build GRCh38
rs946975718 Chr.17: 33088773-33088773 on Build GRCh38
rs375233765 Chr.17: 33088932-33088932 on Build GRCh38
rs760834305 Chr.17: 33088824-33088827 on Build GRCh38
rs115202531 Chr.17: 33089018-33089018 on Build GRCh38
rs540206519 Chr.17: 33088778-33088778 on Build GRCh38
rs138635070 Chr.17: 33088929-33088929 on Build GRCh38
rs184514372 Chr.17: 33089062-33089062 on Build GRCh38
rs369787032 Chr.17: 33089011-33089011 on Build GRCh38
rs1056636238 Chr.17: 33088828-33088828 on Build GRCh38
rs562190167 Chr.17: 33088931-33088931 on Build GRCh38
rs986889385 Chr.17: 33088717-33088717 on Build GRCh38
rs547734582 Chr.17: 33088999-33088999 on Build GRCh38
rs374050459 Chr.17: 33088823-33088823 on Build GRCh38
rs756497058 Chr.17: 33088839-33088839 on Build GRCh38
rs556982917 Chr.17: 33088983-33088983 on Build GRCh38
rs904865294 Chr.17: 33088803-33088803 on Build GRCh38
rs371953023 Chr.17: 33088726-33088726 on Build GRCh38
rs371416854 Chr.17: 33088836-33088836 on Build GRCh38
rs202059141 Chr.17: 33088817-33088817 on Build GRCh38
rs369996202 Chr.17: 33089030-33089030 on Build GRCh38
rs745617613 Chr.17: 33088897-33088897 on Build GRCh38
rs769714774 Chr.17: 33088916-33088916 on Build GRCh38
rs774372425 Chr.17: 33089041-33089041 on Build GRCh38
rs541999706 Chr.17: 33088917-33088917 on Build GRCh38
rs762853994 Chr.17: 33088925-33088925 on Build GRCh38
rs926872236 Chr.17: 33089043-33089043 on Build GRCh38
rs749680275 Chr.17: 33088888-33088888 on Build GRCh38
rs956803357 Chr.17: 33089002-33089002 on Build GRCh38
rs149305028 Chr.17: 33088971-33088971 on Build GRCh38
rs750110652 Chr.17: 33088967-33088967 on Build GRCh38
rs915398469 Chr.17: 33088767-33088767 on Build GRCh38
rs571182369 Chr.17: 33089015-33089015 on Build GRCh38
rs755029202 Chr.17: 33088868-33088868 on Build GRCh38
rs768520543 Chr.17: 33089036-33089036 on Build GRCh38
rs756923404 Chr.17: 33088824-33088824 on Build GRCh38
rs554638132 Chr.17: 33088711-33088711 on Build GRCh38
rs562297571 Chr.17: 33088821-33088821 on Build GRCh38
rs755504700 Chr.17: 33088986-33088986 on Build GRCh38
rs147403420 Chr.17: 33088984-33088984 on Build GRCh38
rs545616961 Chr.17: 33088818-33088818 on Build GRCh38
rs772463415 Chr.17: 33088887-33088887 on Build GRCh38
rs780501546 Chr.17: 33088841-33088841 on Build GRCh38
rs192283562 Chr.17: 33089019-33089019 on Build GRCh38
rs938589875 Chr.17: 33088944-33088944 on Build GRCh38
rs377264444 Chr.17: 33088833-33088833 on Build GRCh38
rs553818063 Chr.17: 33088793-33088793 on Build GRCh38
rs749766264 Chr.17: 33088864-33088864 on Build GRCh38
rs767112053 Chr.17: 33088938-33088938 on Build GRCh38
rs886272776 Chr.17: 33088982-33088982 on Build GRCh38
rs372633638 Chr.17: 33089040-33089040 on Build GRCh38
rs150592728 Chr.17: 33088878-33088878 on Build GRCh38
rs752669551 Chr.17: 33089014-33089014 on Build GRCh38
rs997198571 Chr.17: 33088908-33088908 on Build GRCh38

View Full Product Details