Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35833818-35834318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAF15
Entrez Gene ID
8148
Gene Name
TATA-box binding protein associated factor 15
Gene Aliases
Npl3, RBP56, TAF2N, TAFII68
UniGene
Hs.402752
Gene Chromosome Location
Chr.17: 35809455-35847242 on Build GRCh38
Transcripts
NM_003487.3 NM_139215.2 XM_011525315.1 XM_017025181.1

SNP Detail

SNP ID Chromosome Location
rs760977388 Chr.17: 35833921-35833921 on Build GRCh38
rs200627084 Chr.17: 35833919-35833919 on Build GRCh38
rs756770834 Chr.17: 35833987-35833987 on Build GRCh38
rs1056497559 Chr.17: 35834158-35834158 on Build GRCh38
rs531653885 Chr.17: 35834281-35834281 on Build GRCh38
rs377714683 Chr.17: 35834002-35834002 on Build GRCh38
rs540846230 Chr.17: 35834079-35834079 on Build GRCh38
rs191187849 Chr.17: 35834142-35834142 on Build GRCh38
rs980403927 Chr.17: 35833917-35833917 on Build GRCh38
rs1050378422 Chr.17: 35834136-35834136 on Build GRCh38
rs996474599 Chr.17: 35834113-35834113 on Build GRCh38
rs762758800 Chr.17: 35833982-35833982 on Build GRCh38
rs773631114 Chr.17: 35833920-35833920 on Build GRCh38
rs572190893 Chr.17: 35834077-35834077 on Build GRCh38
rs763966596 Chr.17: 35833984-35833984 on Build GRCh38
rs946684699 Chr.17: 35834103-35834103 on Build GRCh38
rs144534686 Chr.17: 35834188-35834188 on Build GRCh38
rs775122475 Chr.17: 35833965-35833965 on Build GRCh38
rs748246904 Chr.17: 35833860-35833860 on Build GRCh38
rs896633493 Chr.17: 35834165-35834165 on Build GRCh38
rs1016234206 Chr.17: 35834255-35834255 on Build GRCh38
rs763322941 Chr.17: 35833974-35833975 on Build GRCh38
rs905235625 Chr.17: 35834128-35834128 on Build GRCh38
rs914507823 Chr.17: 35834101-35834101 on Build GRCh38
rs555404761 Chr.17: 35834076-35834076 on Build GRCh38
rs79909998 Chr.17: 35834277-35834277 on Build GRCh38
rs1004395036 Chr.17: 35834211-35834211 on Build GRCh38
rs762670345 Chr.17: 35834094-35834094 on Build GRCh38
rs765116444 Chr.17: 35833961-35833961 on Build GRCh38
rs545748074 Chr.17: 35834234-35834234 on Build GRCh38
rs899022707 Chr.17: 35834102-35834102 on Build GRCh38
rs1053432669 Chr.17: 35834099-35834099 on Build GRCh38
rs542105042 Chr.17: 35834052-35834052 on Build GRCh38
rs940458214 Chr.17: 35834089-35834089 on Build GRCh38
rs373860747 Chr.17: 35833870-35833870 on Build GRCh38
rs934537637 Chr.17: 35834096-35834096 on Build GRCh38
rs889752166 Chr.17: 35834187-35834187 on Build GRCh38
rs560189097 Chr.17: 35834153-35834153 on Build GRCh38
rs144140449 Chr.17: 35834080-35834081 on Build GRCh38
rs1043710535 Chr.17: 35834107-35834107 on Build GRCh38

View Full Product Details