Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 36942673-36943166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LHX1
Entrez Gene ID
3975
Gene Name
LIM homeobox 1
Gene Aliases
LIM-1, LIM1
UniGene
Hs.443727
Gene Chromosome Location
Chr.17: 36937475-36944615 on Build GRCh38
Transcripts
NM_005568.4

SNP Detail

SNP ID Chromosome Location
rs912733730 Chr.17: 36942698-36942698 on Build GRCh38
rs890084232 Chr.17: 36943062-36943062 on Build GRCh38
rs759116419 Chr.17: 36942924-36942924 on Build GRCh38
rs777444107 Chr.17: 36942966-36942966 on Build GRCh38
rs775931616 Chr.17: 36942985-36942985 on Build GRCh38
rs770417851 Chr.17: 36942913-36942913 on Build GRCh38
rs750024218 Chr.17: 36943120-36943120 on Build GRCh38
rs748767388 Chr.17: 36943127-36943127 on Build GRCh38
rs747919660 Chr.17: 36942892-36942892 on Build GRCh38
rs760787749 Chr.17: 36943020-36943020 on Build GRCh38
rs751748886 Chr.17: 36942866-36942866 on Build GRCh38
rs989033854 Chr.17: 36942703-36942703 on Build GRCh38
rs148800710 Chr.17: 36942812-36942812 on Build GRCh38
rs772833109 Chr.17: 36942735-36942735 on Build GRCh38
rs1003452956 Chr.17: 36942944-36942944 on Build GRCh38
rs539331458 Chr.17: 36942702-36942702 on Build GRCh38
rs779058919 Chr.17: 36942883-36942883 on Build GRCh38
rs886236099 Chr.17: 36942823-36942823 on Build GRCh38
rs372137646 Chr.17: 36942774-36942774 on Build GRCh38
rs1016113857 Chr.17: 36942859-36942859 on Build GRCh38
rs376663056 Chr.17: 36942925-36942925 on Build GRCh38
rs540796937 Chr.17: 36942953-36942953 on Build GRCh38
rs778039404 Chr.17: 36942795-36942795 on Build GRCh38
rs377221498 Chr.17: 36942905-36942905 on Build GRCh38
rs151062337 Chr.17: 36942996-36942996 on Build GRCh38
rs546566090 Chr.17: 36942807-36942807 on Build GRCh38
rs776029683 Chr.17: 36942929-36942929 on Build GRCh38
rs765794281 Chr.17: 36942854-36942854 on Build GRCh38
rs376578880 Chr.17: 36943098-36943098 on Build GRCh38
rs182084469 Chr.17: 36943050-36943050 on Build GRCh38
rs147937067 Chr.17: 36942830-36942830 on Build GRCh38
rs755550501 Chr.17: 36943066-36943066 on Build GRCh38
rs757317186 Chr.17: 36942867-36942867 on Build GRCh38
rs763352587 Chr.17: 36942858-36942858 on Build GRCh38
rs568432612 Chr.17: 36943104-36943104 on Build GRCh38
rs538710873 Chr.17: 36943103-36943103 on Build GRCh38
rs747372404 Chr.17: 36942954-36942954 on Build GRCh38
rs556078877 Chr.17: 36942704-36942704 on Build GRCh38
rs749084444 Chr.17: 36942705-36942705 on Build GRCh38
rs368870359 Chr.17: 36943125-36943125 on Build GRCh38
rs767076022 Chr.17: 36942731-36942731 on Build GRCh38
rs772041347 Chr.17: 36942903-36942903 on Build GRCh38
rs752624070 Chr.17: 36943028-36943028 on Build GRCh38
rs749211240 Chr.17: 36942808-36942808 on Build GRCh38
rs141144953 Chr.17: 36943085-36943085 on Build GRCh38
rs769201460 Chr.17: 36943007-36943007 on Build GRCh38
rs371131038 Chr.17: 36943107-36943107 on Build GRCh38
rs746313460 Chr.17: 36942788-36942790 on Build GRCh38
rs987682191 Chr.17: 36943041-36943041 on Build GRCh38
rs760168771 Chr.17: 36942740-36942740 on Build GRCh38
rs371586697 Chr.17: 36943146-36943146 on Build GRCh38
rs764470167 Chr.17: 36942940-36942940 on Build GRCh38
rs772208590 Chr.17: 36942972-36942983 on Build GRCh38
rs768354752 Chr.17: 36942708-36942708 on Build GRCh38
rs1030764730 Chr.17: 36942935-36942935 on Build GRCh38
rs768496918 Chr.17: 36942809-36942809 on Build GRCh38
rs773502109 Chr.17: 36943100-36943100 on Build GRCh38
rs1037485513 Chr.17: 36943023-36943023 on Build GRCh38
rs752860474 Chr.17: 36942779-36942779 on Build GRCh38
rs373405460 Chr.17: 36943059-36943059 on Build GRCh38
rs375453089 Chr.17: 36942801-36942801 on Build GRCh38
rs150197430 Chr.17: 36943021-36943021 on Build GRCh38
rs746646319 Chr.17: 36942906-36942906 on Build GRCh38
rs758607846 Chr.17: 36942781-36942781 on Build GRCh38
rs746882089 Chr.17: 36942967-36942967 on Build GRCh38
rs775423572 Chr.17: 36943097-36943098 on Build GRCh38
rs754566332 Chr.17: 36943141-36943141 on Build GRCh38
rs947853594 Chr.17: 36942916-36942916 on Build GRCh38
rs759976630 Chr.17: 36942852-36942852 on Build GRCh38
rs867895446 Chr.17: 36942901-36942901 on Build GRCh38
rs144896333 Chr.17: 36942819-36942819 on Build GRCh38
rs775993744 Chr.17: 36942918-36942918 on Build GRCh38
rs914771898 Chr.17: 36942977-36942977 on Build GRCh38
rs963867352 Chr.17: 36942717-36942717 on Build GRCh38
rs199559106 Chr.17: 36942875-36942875 on Build GRCh38
rs758201785 Chr.17: 36942963-36942963 on Build GRCh38
rs996138732 Chr.17: 36942899-36942899 on Build GRCh38
rs751734142 Chr.17: 36942945-36942945 on Build GRCh38
rs761952441 Chr.17: 36942949-36942949 on Build GRCh38
rs923937180 Chr.17: 36942822-36942822 on Build GRCh38
rs772508352 Chr.17: 36943094-36943094 on Build GRCh38
rs775026965 Chr.17: 36942703-36942703 on Build GRCh38
rs758398612 Chr.17: 36943040-36943040 on Build GRCh38
rs75973369 Chr.17: 36943008-36943008 on Build GRCh38
rs780940543 Chr.17: 36942982-36942982 on Build GRCh38
rs766878151 Chr.17: 36942741-36942741 on Build GRCh38
rs112081488 Chr.17: 36942855-36942855 on Build GRCh38
rs761558885 Chr.17: 36942719-36942719 on Build GRCh38
rs748438626 Chr.17: 36943086-36943086 on Build GRCh38
rs899033442 Chr.17: 36943035-36943035 on Build GRCh38
rs777130906 Chr.17: 36942845-36942845 on Build GRCh38
rs570337695 Chr.17: 36942699-36942699 on Build GRCh38
rs774562789 Chr.17: 36942997-36942997 on Build GRCh38
rs145694637 Chr.17: 36943072-36943072 on Build GRCh38
rs963847409 Chr.17: 36942993-36942993 on Build GRCh38
rs369853713 Chr.17: 36942879-36942879 on Build GRCh38
rs770380533 Chr.17: 36942840-36942840 on Build GRCh38
rs201825686 Chr.17: 36942930-36942930 on Build GRCh38
rs765591915 Chr.17: 36942777-36942777 on Build GRCh38
rs369179293 Chr.17: 36943049-36943049 on Build GRCh38
rs372094506 Chr.17: 36942891-36942891 on Build GRCh38
rs770488670 Chr.17: 36942981-36942981 on Build GRCh38
rs767344391 Chr.17: 36943108-36943108 on Build GRCh38
rs754324037 Chr.17: 36942743-36942743 on Build GRCh38
rs774104700 Chr.17: 36942714-36942714 on Build GRCh38
rs546519526 Chr.17: 36943009-36943009 on Build GRCh38
rs752713616 Chr.17: 36942961-36942961 on Build GRCh38
rs988960278 Chr.17: 36942975-36942975 on Build GRCh38
rs145489633 Chr.17: 36942984-36942984 on Build GRCh38
rs754950912 Chr.17: 36942880-36942880 on Build GRCh38
rs767835432 Chr.17: 36942952-36942952 on Build GRCh38
rs915270470 Chr.17: 36943102-36943102 on Build GRCh38
rs764282476 Chr.17: 36942863-36942863 on Build GRCh38

View Full Product Details