Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39165633-39166126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL5C
Entrez Gene ID
390790
Gene Name
ADP ribosylation factor like GTPase 5C
Gene Aliases
ARL12
UniGene
Hs.568007
Gene Chromosome Location
Chr.17: 39156894-39166161 on Build GRCh38
Transcripts
NM_001143968.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7215676 c.46+7G>A p.? Chr.17: 39165708-39165708 on Build GRCh38
978538 c.16G>A p.A6T Chr.17: 39165745-39165745 on Build GRCh38
3691522 c.15C>T p.I5I Chr.17: 39165746-39165746 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751260465 Chr.17: 39166087-39166087 on Build GRCh38
rs563041686 Chr.17: 39165841-39165841 on Build GRCh38
rs973759166 Chr.17: 39166105-39166105 on Build GRCh38
rs1048239354 Chr.17: 39165775-39165775 on Build GRCh38
rs593772 Chr.17: 39165708-39165708 on Build GRCh38
rs16527 Chr.17: 39165827-39165827 on Build GRCh38
rs73304948 Chr.17: 39165777-39165777 on Build GRCh38
rs779814640 Chr.17: 39165997-39165997 on Build GRCh38
rs762365899 Chr.17: 39165805-39165805 on Build GRCh38
rs185706581 Chr.17: 39165830-39165830 on Build GRCh38
rs75055280 Chr.17: 39165801-39165801 on Build GRCh38
rs955234561 Chr.17: 39165759-39165759 on Build GRCh38
rs575079347 Chr.17: 39165700-39165700 on Build GRCh38
rs529318420 Chr.17: 39165829-39165829 on Build GRCh38
rs373649789 Chr.17: 39166098-39166098 on Build GRCh38
rs373730174 Chr.17: 39165770-39165770 on Build GRCh38
rs958078325 Chr.17: 39165816-39165816 on Build GRCh38
rs943588013 Chr.17: 39165724-39165724 on Build GRCh38
rs1002054849 Chr.17: 39165803-39165803 on Build GRCh38
rs964390662 Chr.17: 39165953-39165953 on Build GRCh38
rs966478402 Chr.17: 39165840-39165841 on Build GRCh38
rs986724830 Chr.17: 39165768-39165768 on Build GRCh38
rs987131565 Chr.17: 39165706-39165706 on Build GRCh38
rs1032788881 Chr.17: 39165846-39165846 on Build GRCh38
rs540143741 Chr.17: 39165802-39165802 on Build GRCh38
rs372247327 Chr.17: 39165729-39165729 on Build GRCh38
rs551958365 Chr.17: 39166061-39166061 on Build GRCh38
rs920735713 Chr.17: 39165765-39165765 on Build GRCh38
rs1047217293 Chr.17: 39165817-39165817 on Build GRCh38
rs533564516 Chr.17: 39165914-39165914 on Build GRCh38
rs1045813568 Chr.17: 39166070-39166070 on Build GRCh38
rs966740474 Chr.17: 39165701-39165701 on Build GRCh38
rs929762852 Chr.17: 39165811-39165811 on Build GRCh38
rs762426874 Chr.17: 39165772-39165772 on Build GRCh38
rs974961051 Chr.17: 39166002-39166002 on Build GRCh38
rs530877967 Chr.17: 39165929-39165929 on Build GRCh38
rs375252863 Chr.17: 39165688-39165688 on Build GRCh38
rs892550567 Chr.17: 39165890-39165890 on Build GRCh38
rs920862218 Chr.17: 39166031-39166032 on Build GRCh38
rs757217440 Chr.17: 39165705-39165705 on Build GRCh38
rs1020094298 Chr.17: 39166040-39166040 on Build GRCh38
rs911541071 Chr.17: 39165900-39165900 on Build GRCh38
rs776370721 Chr.17: 39165670-39165670 on Build GRCh38
rs955205771 Chr.17: 39166059-39166059 on Build GRCh38
rs905465304 Chr.17: 39165843-39165843 on Build GRCh38
rs1008538921 Chr.17: 39166060-39166060 on Build GRCh38
rs758292043 Chr.17: 39165924-39165924 on Build GRCh38
rs1009656752 Chr.17: 39166038-39166038 on Build GRCh38
rs1000860254 Chr.17: 39165845-39165845 on Build GRCh38
rs535173380 Chr.17: 39165653-39165653 on Build GRCh38
rs939415196 Chr.17: 39165732-39165732 on Build GRCh38
rs908067055 Chr.17: 39166054-39166054 on Build GRCh38

View Full Product Details