Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 65011881-65012390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNA13
Entrez Gene ID
10672
Gene Name
G protein subunit alpha 13
Gene Aliases
G13
UniGene
Hs.515018
Gene Chromosome Location
Chr.17: 65009289-65056802 on Build GRCh38
Transcripts
NM_001282425.1 NM_006572.5

SNP Detail

SNP ID Chromosome Location
rs1027877618 Chr.17: 65012161-65012161 on Build GRCh38
rs935500851 Chr.17: 65011995-65011995 on Build GRCh38
rs768520601 Chr.17: 65012062-65012062 on Build GRCh38
rs570458317 Chr.17: 65012266-65012266 on Build GRCh38
rs773359531 Chr.17: 65012009-65012010 on Build GRCh38
rs960797473 Chr.17: 65012229-65012229 on Build GRCh38
rs148137657 Chr.17: 65012230-65012230 on Build GRCh38
rs983297124 Chr.17: 65012189-65012189 on Build GRCh38
rs1052666659 Chr.17: 65012299-65012299 on Build GRCh38
rs939015156 Chr.17: 65012298-65012298 on Build GRCh38
rs891323938 Chr.17: 65012306-65012306 on Build GRCh38
rs907726510 Chr.17: 65012198-65012198 on Build GRCh38
rs754282267 Chr.17: 65012291-65012291 on Build GRCh38
rs184858377 Chr.17: 65012277-65012277 on Build GRCh38
rs956983027 Chr.17: 65012265-65012265 on Build GRCh38
rs116270472 Chr.17: 65012095-65012095 on Build GRCh38
rs1027284111 Chr.17: 65012359-65012359 on Build GRCh38
rs986064556 Chr.17: 65011948-65011948 on Build GRCh38
rs965354519 Chr.17: 65012196-65012196 on Build GRCh38
rs1052614145 Chr.17: 65012081-65012081 on Build GRCh38
rs915377742 Chr.17: 65012269-65012269 on Build GRCh38
rs566434115 Chr.17: 65011996-65011996 on Build GRCh38
rs561406494 Chr.17: 65012189-65012189 on Build GRCh38
rs1007448367 Chr.17: 65012165-65012165 on Build GRCh38
rs1036092355 Chr.17: 65012092-65012092 on Build GRCh38
rs939052615 Chr.17: 65012091-65012091 on Build GRCh38
rs1027566485 Chr.17: 65012166-65012166 on Build GRCh38
rs924046186 Chr.17: 65011990-65011990 on Build GRCh38
rs1045300082 Chr.17: 65012038-65012038 on Build GRCh38
rs749501534 Chr.17: 65011946-65011946 on Build GRCh38
rs977187767 Chr.17: 65012212-65012212 on Build GRCh38
rs760780020 Chr.17: 65012021-65012025 on Build GRCh38
rs573793130 Chr.17: 65012326-65012326 on Build GRCh38
rs142033484 Chr.17: 65012333-65012333 on Build GRCh38
rs181465721 Chr.17: 65012088-65012088 on Build GRCh38
rs964439470 Chr.17: 65012130-65012130 on Build GRCh38
rs560556225 Chr.17: 65012008-65012008 on Build GRCh38
rs1027557476 Chr.17: 65011919-65011919 on Build GRCh38
rs994607192 Chr.17: 65012101-65012101 on Build GRCh38
rs866737383 Chr.17: 65011962-65011962 on Build GRCh38
rs920500012 Chr.17: 65011923-65011923 on Build GRCh38
rs548378531 Chr.17: 65012179-65012179 on Build GRCh38
rs531834236 Chr.17: 65012142-65012142 on Build GRCh38
rs980988881 Chr.17: 65012279-65012279 on Build GRCh38
rs965377644 Chr.17: 65011967-65011967 on Build GRCh38
rs1056922038 Chr.17: 65011972-65011972 on Build GRCh38
rs983245020 Chr.17: 65011937-65011937 on Build GRCh38
rs540225537 Chr.17: 65011983-65011983 on Build GRCh38
rs928241556 Chr.17: 65012285-65012285 on Build GRCh38
rs1048850498 Chr.17: 65012365-65012365 on Build GRCh38
rs999740615 Chr.17: 65012070-65012070 on Build GRCh38
rs951965542 Chr.17: 65012187-65012187 on Build GRCh38
rs1018248171 Chr.17: 65011956-65011956 on Build GRCh38
rs888954207 Chr.17: 65012163-65012163 on Build GRCh38
rs550140767 Chr.17: 65012354-65012354 on Build GRCh38
rs532672804 Chr.17: 65012028-65012028 on Build GRCh38
rs1019092362 Chr.17: 65012115-65012115 on Build GRCh38
rs944190224 Chr.17: 65012307-65012307 on Build GRCh38
rs75200830 Chr.17: 65012355-65012355 on Build GRCh38
rs544378372 Chr.17: 65012345-65012345 on Build GRCh38
rs527396584 Chr.17: 65012190-65012190 on Build GRCh38
rs935584821 Chr.17: 65012294-65012294 on Build GRCh38
rs953245538 Chr.17: 65011930-65011930 on Build GRCh38
rs976853921 Chr.17: 65011970-65011970 on Build GRCh38
rs964709008 Chr.17: 65011918-65011918 on Build GRCh38

View Full Product Details