Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75272766-75273265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100287042
Entrez Gene ID
100287042
Gene Name
uncharacterized LOC100287042
Gene Aliases
-
UniGene
Hs.514470
Gene Chromosome Location
Chr.17: 75271299-75273895 on Build GRCh38
Transcripts
NR_036520.1 NM_001126121.1 NM_001126122.1 NM_021734.4 XM_005257559.3 XM_005257560.2 XM_005257561.3 XM_005257562.2 XM_006722007.2 XM_011525098.1 XM_017024926.1 XM_017024927.1 XM_017024928.1

Gene Symbol
SLC25A19
Entrez Gene ID
60386
Gene Name
solute carrier family 25 member 19
Gene Aliases
DNC, MCPHA, MUP1, THMD3, THMD4, TPC
UniGene
Hs.514470
Gene Chromosome Location
Chr.17: 75272980-75289957 on Build GRCh38
Transcripts
NR_036520.1 NM_001126121.1 NM_001126122.1 NM_021734.4 XM_005257559.3 XM_005257560.2 XM_005257561.3 XM_005257562.2 XM_006722007.2 XM_011525098.1 XM_017024926.1 XM_017024927.1 XM_017024928.1

SNP Detail

SNP ID Chromosome Location
rs547940324 Chr.17: 75273186-75273186 on Build GRCh38
rs3744230 Chr.17: 75272823-75272823 on Build GRCh38
rs62089320 Chr.17: 75272976-75272976 on Build GRCh38
rs932280155 Chr.17: 75273020-75273020 on Build GRCh38
rs539840492 Chr.17: 75272938-75272938 on Build GRCh38
rs574512861 Chr.17: 75273113-75273113 on Build GRCh38
rs1049306534 Chr.17: 75273156-75273156 on Build GRCh38
rs951995785 Chr.17: 75272862-75272862 on Build GRCh38
rs966483821 Chr.17: 75273233-75273233 on Build GRCh38
rs897971230 Chr.17: 75273142-75273142 on Build GRCh38
rs889293661 Chr.17: 75272809-75272809 on Build GRCh38
rs751644411 Chr.17: 75273081-75273081 on Build GRCh38
rs912336789 Chr.17: 75273071-75273071 on Build GRCh38
rs1034140674 Chr.17: 75272942-75272942 on Build GRCh38
rs760328618 Chr.17: 75272950-75272950 on Build GRCh38
rs543638381 Chr.17: 75273150-75273150 on Build GRCh38
rs866088396 Chr.17: 75272818-75272818 on Build GRCh38
rs557354128 Chr.17: 75272894-75272894 on Build GRCh38
rs1029690572 Chr.17: 75272846-75272846 on Build GRCh38
rs996844855 Chr.17: 75272824-75272824 on Build GRCh38
rs192494814 Chr.17: 75273067-75273067 on Build GRCh38
rs35911256 Chr.17: 75272925-75272926 on Build GRCh38
rs928277355 Chr.17: 75272988-75272988 on Build GRCh38
rs563237505 Chr.17: 75273158-75273158 on Build GRCh38
rs541911861 Chr.17: 75273073-75273073 on Build GRCh38
rs532311879 Chr.17: 75273162-75273162 on Build GRCh38
rs1009521638 Chr.17: 75272810-75272810 on Build GRCh38
rs1053408093 Chr.17: 75272996-75272996 on Build GRCh38
rs3180439 Chr.17: 75272991-75272991 on Build GRCh38
rs966618278 Chr.17: 75272989-75272989 on Build GRCh38
rs747664256 Chr.17: 75272787-75272787 on Build GRCh38
rs913723283 Chr.17: 75272983-75272983 on Build GRCh38
rs1027674581 Chr.17: 75272861-75272861 on Build GRCh38
rs970213671 Chr.17: 75273228-75273228 on Build GRCh38
rs577641912 Chr.17: 75272917-75272917 on Build GRCh38
rs568179839 Chr.17: 75273193-75273193 on Build GRCh38
rs896591116 Chr.17: 75273203-75273203 on Build GRCh38
rs1020757872 Chr.17: 75273231-75273231 on Build GRCh38
rs979290129 Chr.17: 75273010-75273010 on Build GRCh38
rs568530325 Chr.17: 75272845-75272845 on Build GRCh38
rs893062616 Chr.17: 75273009-75273009 on Build GRCh38
rs562233396 Chr.17: 75273110-75273110 on Build GRCh38
rs7198 Chr.17: 75273177-75273177 on Build GRCh38
rs915660545 Chr.17: 75272918-75272918 on Build GRCh38
rs552061911 Chr.17: 75273163-75273163 on Build GRCh38
rs774360327 Chr.17: 75272979-75272980 on Build GRCh38
rs908811460 Chr.17: 75272882-75272882 on Build GRCh38
rs1004218259 Chr.17: 75273159-75273159 on Build GRCh38
rs771746604 Chr.17: 75272792-75272792 on Build GRCh38
rs766111465 Chr.17: 75272972-75272972 on Build GRCh38
rs73356384 Chr.17: 75273014-75273014 on Build GRCh38
rs1033677225 Chr.17: 75273183-75273183 on Build GRCh38
rs1013652498 Chr.17: 75273230-75273230 on Build GRCh38
rs3744229 Chr.17: 75272825-75272825 on Build GRCh38
rs558874774 Chr.17: 75273167-75273167 on Build GRCh38
rs537665302 Chr.17: 75272863-75272863 on Build GRCh38
rs761864925 Chr.17: 75273132-75273132 on Build GRCh38

View Full Product Details