Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 50663778-50664283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPK4
Entrez Gene ID
5596
Gene Name
mitogen-activated protein kinase 4
Gene Aliases
ERK-4, ERK4, PRKM4, p63-MAPK, p63MAPK
UniGene
Hs.433728
Gene Chromosome Location
Chr.18: 50559781-50731826 on Build GRCh38
Transcripts
NM_001292039.1 NM_001292040.1 NM_002747.3 XM_005258299.3 XM_011526074.2 XM_011526075.2 XM_011526076.2 XM_017025839.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6233221 c.282G>T p.E94D Chr.18: 50664240-50664240 on Build GRCh38
473900 c.4G>A p.A2T Chr.18: 50663962-50663962 on Build GRCh38
6707856 c.280G>A p.E94K Chr.18: 50664238-50664238 on Build GRCh38
1750542 c.74C>T p.P25L Chr.18: 50664032-50664032 on Build GRCh38
4000421 c.112G>A p.V38M Chr.18: 50664070-50664070 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1040441186 Chr.18: 50664196-50664196 on Build GRCh38
rs944821568 Chr.18: 50663914-50663914 on Build GRCh38
rs762992386 Chr.18: 50664089-50664089 on Build GRCh38
rs548264569 Chr.18: 50664193-50664194 on Build GRCh38
rs750239947 Chr.18: 50664186-50664188 on Build GRCh38
rs900728295 Chr.18: 50663964-50663964 on Build GRCh38
rs368717366 Chr.18: 50664261-50664261 on Build GRCh38
rs565759866 Chr.18: 50664125-50664125 on Build GRCh38
rs992713532 Chr.18: 50663853-50663853 on Build GRCh38
rs920644770 Chr.18: 50664088-50664088 on Build GRCh38
rs745586840 Chr.18: 50664143-50664143 on Build GRCh38
rs536964018 Chr.18: 50664171-50664171 on Build GRCh38
rs866862776 Chr.18: 50664162-50664162 on Build GRCh38
rs757694990 Chr.18: 50664200-50664200 on Build GRCh38
rs767365680 Chr.18: 50664169-50664169 on Build GRCh38
rs201113430 Chr.18: 50663959-50663959 on Build GRCh38
rs199714690 Chr.18: 50664256-50664256 on Build GRCh38
rs767319099 Chr.18: 50664065-50664066 on Build GRCh38
rs756144988 Chr.18: 50663957-50663957 on Build GRCh38
rs781287223 Chr.18: 50663940-50663940 on Build GRCh38
rs576351233 Chr.18: 50663895-50663895 on Build GRCh38
rs958449532 Chr.18: 50663852-50663852 on Build GRCh38
rs55800528 Chr.18: 50664141-50664141 on Build GRCh38
rs199879399 Chr.18: 50664080-50664080 on Build GRCh38
rs367914884 Chr.18: 50664066-50664066 on Build GRCh38
rs3752087 Chr.18: 50664070-50664070 on Build GRCh38
rs762751290 Chr.18: 50664149-50664149 on Build GRCh38
rs916938136 Chr.18: 50663877-50663877 on Build GRCh38
rs775288765 Chr.18: 50664086-50664086 on Build GRCh38
rs56335862 Chr.18: 50664210-50664210 on Build GRCh38
rs972025834 Chr.18: 50663878-50663878 on Build GRCh38
rs763775905 Chr.18: 50664096-50664096 on Build GRCh38
rs200603915 Chr.18: 50664166-50664166 on Build GRCh38
rs755853041 Chr.18: 50664204-50664204 on Build GRCh38
rs140972647 Chr.18: 50664113-50664113 on Build GRCh38
rs947030584 Chr.18: 50664138-50664138 on Build GRCh38
rs1042753382 Chr.18: 50663987-50663987 on Build GRCh38
rs556250636 Chr.18: 50664260-50664260 on Build GRCh38
rs748473611 Chr.18: 50663975-50663975 on Build GRCh38
rs764672737 Chr.18: 50664190-50664190 on Build GRCh38
rs765073743 Chr.18: 50663920-50663920 on Build GRCh38
rs776211637 Chr.18: 50664079-50664079 on Build GRCh38
rs779378577 Chr.18: 50664046-50664046 on Build GRCh38
rs765961979 Chr.18: 50663991-50663991 on Build GRCh38
rs1047768083 Chr.18: 50664243-50664243 on Build GRCh38
rs950749935 Chr.18: 50664244-50664244 on Build GRCh38
rs755010351 Chr.18: 50663919-50663919 on Build GRCh38
rs755045853 Chr.18: 50664114-50664114 on Build GRCh38
rs905689204 Chr.18: 50664205-50664205 on Build GRCh38
rs770735277 Chr.18: 50664075-50664075 on Build GRCh38
rs34207571 Chr.18: 50664008-50664008 on Build GRCh38
rs765272318 Chr.18: 50664003-50664003 on Build GRCh38
rs541638701 Chr.18: 50663986-50663986 on Build GRCh38
rs373566179 Chr.18: 50664128-50664128 on Build GRCh38
rs768671292 Chr.18: 50664163-50664163 on Build GRCh38
rs765979945 Chr.18: 50663911-50663911 on Build GRCh38
rs61747609 Chr.18: 50664121-50664121 on Build GRCh38
rs370578537 Chr.18: 50663936-50663936 on Build GRCh38
rs772545058 Chr.18: 50663980-50663980 on Build GRCh38
rs749490740 Chr.18: 50663856-50663856 on Build GRCh38
rs369990219 Chr.18: 50664127-50664127 on Build GRCh38
rs754028426 Chr.18: 50664170-50664170 on Build GRCh38
rs370265083 Chr.18: 50663982-50663982 on Build GRCh38
rs367684848 Chr.18: 50664097-50664097 on Build GRCh38
rs559455057 Chr.18: 50664131-50664131 on Build GRCh38
rs959869255 Chr.18: 50664038-50664038 on Build GRCh38
rs749775353 Chr.18: 50663966-50663966 on Build GRCh38
rs751209391 Chr.18: 50663932-50663932 on Build GRCh38
rs781721089 Chr.18: 50664201-50664201 on Build GRCh38
rs746164583 Chr.18: 50663954-50663954 on Build GRCh38
rs779393411 Chr.18: 50664209-50664209 on Build GRCh38
rs536389282 Chr.18: 50664014-50664014 on Build GRCh38
rs774550171 Chr.18: 50663900-50663900 on Build GRCh38
rs759503690 Chr.18: 50664001-50664001 on Build GRCh38
rs977690745 Chr.18: 50663894-50663894 on Build GRCh38
rs3752085 Chr.18: 50663901-50663901 on Build GRCh38
rs3752086 Chr.18: 50664042-50664042 on Build GRCh38
rs760849669 Chr.18: 50664100-50664100 on Build GRCh38
rs372835108 Chr.18: 50664148-50664148 on Build GRCh38
rs758963755 Chr.18: 50664123-50664123 on Build GRCh38
rs267605201 Chr.18: 50663961-50663961 on Build GRCh38
rs374524649 Chr.18: 50664007-50664007 on Build GRCh38
rs200651462 Chr.18: 50664255-50664255 on Build GRCh38
rs772293615 Chr.18: 50664065-50664065 on Build GRCh38
rs761581511 Chr.18: 50664167-50664167 on Build GRCh38
rs56259916 Chr.18: 50664120-50664120 on Build GRCh38
rs752849769 Chr.18: 50664181-50664181 on Build GRCh38
rs943420702 Chr.18: 50664180-50664180 on Build GRCh38
rs572593041 Chr.18: 50663971-50663971 on Build GRCh38
rs774339790 Chr.18: 50664228-50664228 on Build GRCh38
rs750579920 Chr.18: 50664013-50664013 on Build GRCh38
rs983563131 Chr.18: 50664117-50664117 on Build GRCh38
rs1027949763 Chr.18: 50664235-50664235 on Build GRCh38
rs973397591 Chr.18: 50663889-50663889 on Build GRCh38
rs748756367 Chr.18: 50664058-50664058 on Build GRCh38
rs756949521 Chr.18: 50664063-50664064 on Build GRCh38
rs867229676 Chr.18: 50664069-50664069 on Build GRCh38
rs556292726 Chr.18: 50663876-50663876 on Build GRCh38
rs755417970 Chr.18: 50664043-50664043 on Build GRCh38
rs748123352 Chr.18: 50664252-50664252 on Build GRCh38
rs768107488 Chr.18: 50664216-50664216 on Build GRCh38
rs532114080 Chr.18: 50664107-50664107 on Build GRCh38
rs758762362 Chr.18: 50664186-50664186 on Build GRCh38
rs558995002 Chr.18: 50663916-50663916 on Build GRCh38
rs770553702 Chr.18: 50663823-50663823 on Build GRCh38
rs886982579 Chr.18: 50664253-50664253 on Build GRCh38
rs372936554 Chr.18: 50663937-50663937 on Build GRCh38
rs745497107 Chr.18: 50664202-50664202 on Build GRCh38
rs769175436 Chr.18: 50663967-50663967 on Build GRCh38
rs1005426120 Chr.18: 50663825-50663825 on Build GRCh38
rs370695586 Chr.18: 50664030-50664030 on Build GRCh38
rs1016684852 Chr.18: 50663828-50663828 on Build GRCh38
rs752066432 Chr.18: 50664194-50664194 on Build GRCh38
rs769489864 Chr.18: 50664081-50664081 on Build GRCh38
rs768790539 Chr.18: 50663879-50663879 on Build GRCh38
rs752485103 Chr.18: 50664006-50664006 on Build GRCh38
rs56328353 Chr.18: 50664144-50664144 on Build GRCh38

View Full Product Details