Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35310455-35310951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAG
Entrez Gene ID
4099
Gene Name
myelin associated glycoprotein
Gene Aliases
GMA, S-MAG, SIGLEC-4A, SIGLEC4A, SPG75
UniGene
Hs.643440
Gene Chromosome Location
Chr.19: 35292086-35313807 on Build GRCh38
Transcripts
NM_001199216.1 NM_002361.3 NM_080600.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5449251 c.1538A>C p.K513T Chr.19: 35310565-35310565 on Build GRCh38
344695 c.1575C>T p.I525I Chr.19: 35310602-35310602 on Build GRCh38
1392825 c.1554C>T p.G518G Chr.19: 35310581-35310581 on Build GRCh38
994941 c.1542C>T p.I514I Chr.19: 35310569-35310569 on Build GRCh38
1680893 c.1522C>T p.R508* Chr.19: 35310549-35310549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776158188 Chr.19: 35310640-35310640 on Build GRCh38
rs375849472 Chr.19: 35310585-35310585 on Build GRCh38
rs1026479960 Chr.19: 35310863-35310863 on Build GRCh38
rs763492004 Chr.19: 35310647-35310647 on Build GRCh38
rs200163072 Chr.19: 35310524-35310524 on Build GRCh38
rs199804955 Chr.19: 35310584-35310584 on Build GRCh38
rs185732027 Chr.19: 35310617-35310617 on Build GRCh38
rs199740655 Chr.19: 35310520-35310520 on Build GRCh38
rs543427177 Chr.19: 35310740-35310740 on Build GRCh38
rs776901974 Chr.19: 35310630-35310630 on Build GRCh38
rs773454882 Chr.19: 35310514-35310514 on Build GRCh38
rs769951488 Chr.19: 35310637-35310637 on Build GRCh38
rs201344002 Chr.19: 35310582-35310582 on Build GRCh38
rs1009777258 Chr.19: 35310707-35310707 on Build GRCh38
rs202032308 Chr.19: 35310521-35310521 on Build GRCh38
rs576709324 Chr.19: 35310753-35310753 on Build GRCh38
rs556153832 Chr.19: 35310509-35310509 on Build GRCh38
rs1015640630 Chr.19: 35310763-35310763 on Build GRCh38
rs35237014 Chr.19: 35310636-35310636 on Build GRCh38
rs557895821 Chr.19: 35310673-35310673 on Build GRCh38
rs750061140 Chr.19: 35310675-35310675 on Build GRCh38
rs1056944771 Chr.19: 35310666-35310666 on Build GRCh38
rs564105713 Chr.19: 35310671-35310671 on Build GRCh38
rs200859403 Chr.19: 35310589-35310589 on Build GRCh38
rs753222649 Chr.19: 35310545-35310545 on Build GRCh38
rs201518817 Chr.19: 35310615-35310615 on Build GRCh38
rs751935497 Chr.19: 35310653-35310653 on Build GRCh38
rs764556817 Chr.19: 35310648-35310648 on Build GRCh38
rs771128534 Chr.19: 35310628-35310628 on Build GRCh38
rs765892219 Chr.19: 35310542-35310542 on Build GRCh38
rs1028516837 Chr.19: 35310875-35310875 on Build GRCh38
rs202241678 Chr.19: 35310501-35310501 on Build GRCh38
rs201465791 Chr.19: 35310529-35310529 on Build GRCh38
rs574568807 Chr.19: 35310877-35310877 on Build GRCh38
rs200514323 Chr.19: 35310683-35310683 on Build GRCh38
rs200469368 Chr.19: 35310504-35310504 on Build GRCh38
rs939398041 Chr.19: 35310644-35310644 on Build GRCh38
rs774895076 Chr.19: 35310855-35310855 on Build GRCh38
rs200994145 Chr.19: 35310497-35310497 on Build GRCh38
rs950498060 Chr.19: 35310871-35310871 on Build GRCh38
rs939347266 Chr.19: 35310925-35310925 on Build GRCh38
rs983321114 Chr.19: 35310876-35310876 on Build GRCh38
rs762223278 Chr.19: 35310672-35310672 on Build GRCh38
rs533479961 Chr.19: 35310929-35310929 on Build GRCh38
rs769008895 Chr.19: 35310844-35310844 on Build GRCh38
rs542000108 Chr.19: 35310892-35310892 on Build GRCh38
rs768463699 Chr.19: 35310598-35310598 on Build GRCh38
rs367555758 Chr.19: 35310670-35310670 on Build GRCh38
rs750172142 Chr.19: 35310864-35310864 on Build GRCh38
rs201687816 Chr.19: 35310590-35310590 on Build GRCh38
rs898636130 Chr.19: 35310789-35310789 on Build GRCh38
rs756792757 Chr.19: 35310580-35310580 on Build GRCh38
rs751237118 Chr.19: 35310574-35310574 on Build GRCh38
rs373591774 Chr.19: 35310543-35310543 on Build GRCh38
rs983008757 Chr.19: 35310553-35310553 on Build GRCh38
rs762279596 Chr.19: 35310922-35310922 on Build GRCh38
rs199748603 Chr.19: 35310569-35310569 on Build GRCh38
rs1056383879 Chr.19: 35310700-35310700 on Build GRCh38
rs768410611 Chr.19: 35310511-35310511 on Build GRCh38
rs561994079 Chr.19: 35310898-35310898 on Build GRCh38
rs749402827 Chr.19: 35310799-35310799 on Build GRCh38
rs986448856 Chr.19: 35310513-35310513 on Build GRCh38
rs757044952 Chr.19: 35310523-35310523 on Build GRCh38
rs938636150 Chr.19: 35310692-35310692 on Build GRCh38
rs965687534 Chr.19: 35310824-35310824 on Build GRCh38
rs896530603 Chr.19: 35310725-35310725 on Build GRCh38
rs75496944 Chr.19: 35310744-35310744 on Build GRCh38
rs200430942 Chr.19: 35310581-35310581 on Build GRCh38
rs746073131 Chr.19: 35310593-35310593 on Build GRCh38
rs910066595 Chr.19: 35310886-35310886 on Build GRCh38

View Full Product Details