Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36546364-36546912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF529
Entrez Gene ID
57711
Gene Name
zinc finger protein 529
Gene Aliases
-
UniGene
Hs.654960
Gene Chromosome Location
Chr.19: 36534491-36605411 on Build GRCh38
Transcripts
NM_001145649.1 NM_001145650.1 NM_001321351.1 NM_020951.4 XM_006723302.3 XM_006723304.2 XM_011527164.2 XM_011527165.2 XM_011527166.2 XM_011527167.2 XM_011527169.2 XM_011527170.1 XM_017027039.1 XM_017027040.1 XM_017027041.1 XM_017027042.1 XM_017027043.1 XM_017027044.1 XM_017027045.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4569332 c.1674T>C p.G558G Chr.19: 36546884-36546884 on Build GRCh38
278346 c.1659G>A p.P553P Chr.19: 36546899-36546899 on Build GRCh38
4077386 c.1646T>C p.L549P Chr.19: 36546912-36546912 on Build GRCh38
5520273 c.1648_1651delACTT p.T550fs*>13 Chr.19: 36546907-36546910 on Build GRCh38
4569333 c.1674T>C p.G558G Chr.19: 36546884-36546884 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535609484 Chr.19: 36546673-36546673 on Build GRCh38
rs112368357 Chr.19: 36546403-36546403 on Build GRCh38
rs1007020032 Chr.19: 36546648-36546648 on Build GRCh38
rs10678581 Chr.19: 36546403-36546404 on Build GRCh38
rs750759014 Chr.19: 36546871-36546871 on Build GRCh38
rs767990256 Chr.19: 36546869-36546869 on Build GRCh38
rs531109591 Chr.19: 36546677-36546677 on Build GRCh38
rs764694417 Chr.19: 36546856-36546856 on Build GRCh38
rs950644746 Chr.19: 36546735-36546735 on Build GRCh38
rs909061566 Chr.19: 36546487-36546490 on Build GRCh38
rs923189114 Chr.19: 36546452-36546452 on Build GRCh38
rs557256658 Chr.19: 36546679-36546679 on Build GRCh38
rs746857805 Chr.19: 36546462-36546462 on Build GRCh38
rs776457645 Chr.19: 36546823-36546823 on Build GRCh38
rs553572270 Chr.19: 36546497-36546497 on Build GRCh38
rs775571924 Chr.19: 36546685-36546685 on Build GRCh38
rs759049681 Chr.19: 36546848-36546848 on Build GRCh38
rs938218556 Chr.19: 36546695-36546695 on Build GRCh38
rs756569304 Chr.19: 36546878-36546878 on Build GRCh38
rs1043213998 Chr.19: 36546593-36546599 on Build GRCh38
rs898608300 Chr.19: 36546409-36546409 on Build GRCh38
rs766951206 Chr.19: 36546883-36546883 on Build GRCh38
rs68121045 Chr.19: 36546407-36546408 on Build GRCh38
rs751988790 Chr.19: 36546747-36546747 on Build GRCh38
rs1021374016 Chr.19: 36546597-36546597 on Build GRCh38
rs1028290489 Chr.19: 36546448-36546448 on Build GRCh38
rs770607214 Chr.19: 36546496-36546496 on Build GRCh38
rs59616372 Chr.19: 36546408-36546409 on Build GRCh38
rs1057446884 Chr.19: 36546536-36546536 on Build GRCh38
rs1019515325 Chr.19: 36546499-36546499 on Build GRCh38
rs1030439615 Chr.19: 36546732-36546733 on Build GRCh38
rs568570715 Chr.19: 36546590-36546590 on Build GRCh38
rs142899174 Chr.19: 36546447-36546447 on Build GRCh38
rs77850123 Chr.19: 36546406-36546406 on Build GRCh38
rs386809011 Chr.19: 36546404-36546406 on Build GRCh38
rs149720452 Chr.19: 36546406-36546407 on Build GRCh38
rs765389144 Chr.19: 36546684-36546684 on Build GRCh38
rs938625942 Chr.19: 36546532-36546532 on Build GRCh38
rs892020960 Chr.19: 36546566-36546566 on Build GRCh38
rs752321921 Chr.19: 36546494-36546494 on Build GRCh38
rs997110371 Chr.19: 36546717-36546717 on Build GRCh38
rs775187673 Chr.19: 36546858-36546858 on Build GRCh38
rs983291079 Chr.19: 36546481-36546481 on Build GRCh38
rs182684905 Chr.19: 36546702-36546702 on Build GRCh38
rs570952298 Chr.19: 36546429-36546429 on Build GRCh38
rs762571412 Chr.19: 36546867-36546867 on Build GRCh38
rs796530855 Chr.19: 36546404-36546405 on Build GRCh38
rs759686280 Chr.19: 36546626-36546626 on Build GRCh38
rs1049694675 Chr.19: 36546788-36546788 on Build GRCh38
rs979935018 Chr.19: 36546759-36546759 on Build GRCh38
rs770801284 Chr.19: 36546818-36546818 on Build GRCh38
rs767769701 Chr.19: 36546758-36546758 on Build GRCh38
rs375236315 Chr.19: 36546764-36546764 on Build GRCh38
rs955888667 Chr.19: 36546511-36546511 on Build GRCh38
rs938764193 Chr.19: 36546798-36546798 on Build GRCh38
rs1057121736 Chr.19: 36546811-36546811 on Build GRCh38
rs776965385 Chr.19: 36546534-36546534 on Build GRCh38
rs558124097 Chr.19: 36546725-36546725 on Build GRCh38
rs918192759 Chr.19: 36546719-36546719 on Build GRCh38
rs369725168 Chr.19: 36546768-36546775 on Build GRCh38
rs989086112 Chr.19: 36546528-36546528 on Build GRCh38
rs764627640 Chr.19: 36546746-36546746 on Build GRCh38
rs964310362 Chr.19: 36546434-36546434 on Build GRCh38
rs889672198 Chr.19: 36546472-36546472 on Build GRCh38
rs575501230 Chr.19: 36546691-36546691 on Build GRCh38
rs757699878 Chr.19: 36546748-36546748 on Build GRCh38
rs371856197 Chr.19: 36546726-36546726 on Build GRCh38
rs927340215 Chr.19: 36546795-36546795 on Build GRCh38

View Full Product Details