Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 37466982-37467473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF569
Entrez Gene ID
148266
Gene Name
zinc finger protein 569
Gene Aliases
ZAP1, ZNF
UniGene
Hs.511848
Gene Chromosome Location
Chr.19: 37411146-37469268 on Build GRCh38
Transcripts
NM_152484.2 XM_006723046.2 XM_006723047.3 XM_006723048.3 XM_011526538.2 XM_017026376.1 XM_017026377.1 XM_017026378.1 XM_017026379.1 XM_017026380.1 XM_017026381.1 XM_017026382.1 XM_011526544.2

Gene Symbol
ZNF570
Entrez Gene ID
148268
Gene Name
zinc finger protein 570
Gene Aliases
-
UniGene
Hs.126962
Gene Chromosome Location
Chr.19: 37467465-37488652 on Build GRCh38
Transcripts
NM_152484.2 XM_006723046.2 XM_006723047.3 XM_006723048.3 XM_011526538.2 XM_017026376.1 XM_017026377.1 XM_017026378.1 XM_017026379.1 XM_017026380.1 XM_017026381.1 XM_017026382.1 XM_011526544.2

SNP Detail

SNP ID Chromosome Location
rs146090495 Chr.19: 37467442-37467442 on Build GRCh38
rs927481778 Chr.19: 37467228-37467228 on Build GRCh38
rs927400217 Chr.19: 37467439-37467439 on Build GRCh38
rs776315555 Chr.19: 37467336-37467336 on Build GRCh38
rs759497921 Chr.19: 37467363-37467363 on Build GRCh38
rs993160289 Chr.19: 37467241-37467241 on Build GRCh38
rs990325445 Chr.19: 37467443-37467443 on Build GRCh38
rs1020594564 Chr.19: 37467086-37467086 on Build GRCh38
rs1054752148 Chr.19: 37467277-37467277 on Build GRCh38
rs981533794 Chr.19: 37467224-37467224 on Build GRCh38
rs528131601 Chr.19: 37467211-37467211 on Build GRCh38
rs1027518857 Chr.19: 37467152-37467152 on Build GRCh38
rs769663795 Chr.19: 37467374-37467374 on Build GRCh38
rs902153298 Chr.19: 37467301-37467301 on Build GRCh38
rs906009993 Chr.19: 37467012-37467012 on Build GRCh38
rs934609231 Chr.19: 37467091-37467091 on Build GRCh38
rs965818534 Chr.19: 37467232-37467232 on Build GRCh38
rs1042087927 Chr.19: 37467294-37467294 on Build GRCh38
rs907842347 Chr.19: 37467215-37467215 on Build GRCh38
rs946245652 Chr.19: 37467286-37467286 on Build GRCh38
rs1010575451 Chr.19: 37467068-37467068 on Build GRCh38
rs1019211857 Chr.19: 37467096-37467096 on Build GRCh38
rs1009086893 Chr.19: 37467334-37467334 on Build GRCh38
rs183043610 Chr.19: 37467448-37467448 on Build GRCh38
rs756289934 Chr.19: 37467414-37467416 on Build GRCh38
rs914713146 Chr.19: 37467446-37467446 on Build GRCh38
rs893510811 Chr.19: 37467051-37467051 on Build GRCh38
rs568392783 Chr.19: 37467316-37467316 on Build GRCh38
rs928660769 Chr.19: 37467080-37467080 on Build GRCh38
rs981970809 Chr.19: 37467406-37467406 on Build GRCh38
rs952084941 Chr.19: 37467165-37467165 on Build GRCh38
rs1040201881 Chr.19: 37467409-37467409 on Build GRCh38
rs190849133 Chr.19: 37467130-37467130 on Build GRCh38
rs1054701681 Chr.19: 37467025-37467025 on Build GRCh38
rs544821467 Chr.19: 37467015-37467015 on Build GRCh38
rs1027404392 Chr.19: 37467381-37467381 on Build GRCh38
rs950130872 Chr.19: 37467005-37467005 on Build GRCh38
rs901632317 Chr.19: 37467419-37467419 on Build GRCh38
rs983308548 Chr.19: 37467384-37467384 on Build GRCh38
rs893396117 Chr.19: 37467282-37467282 on Build GRCh38
rs937517614 Chr.19: 37467258-37467258 on Build GRCh38
rs564127714 Chr.19: 37467210-37467210 on Build GRCh38
rs983426115 Chr.19: 37467212-37467212 on Build GRCh38
rs993190775 Chr.19: 37467427-37467427 on Build GRCh38
rs1039713900 Chr.19: 37467391-37467392 on Build GRCh38
rs568978802 Chr.19: 37467379-37467379 on Build GRCh38
rs1023147545 Chr.19: 37467333-37467333 on Build GRCh38
rs546613437 Chr.19: 37467248-37467248 on Build GRCh38
rs533978958 Chr.19: 37467436-37467436 on Build GRCh38
rs545938055 Chr.19: 37467176-37467176 on Build GRCh38
rs937616125 Chr.19: 37467018-37467018 on Build GRCh38
rs888916199 Chr.19: 37467188-37467188 on Build GRCh38
rs533536249 Chr.19: 37467144-37467144 on Build GRCh38
rs1041952809 Chr.19: 37467452-37467452 on Build GRCh38
rs1043584041 Chr.19: 37467177-37467177 on Build GRCh38
rs540960010 Chr.19: 37467392-37467392 on Build GRCh38
rs566618240 Chr.19: 37467407-37467407 on Build GRCh38
rs550817439 Chr.19: 37467350-37467350 on Build GRCh38
rs902268219 Chr.19: 37467089-37467089 on Build GRCh38
rs145508409 Chr.19: 37467256-37467256 on Build GRCh38
rs887139938 Chr.19: 37467222-37467222 on Build GRCh38
rs1010809543 Chr.19: 37467147-37467147 on Build GRCh38
rs539574648 Chr.19: 37467382-37467382 on Build GRCh38
rs948850846 Chr.19: 37467050-37467050 on Build GRCh38
rs1008618280 Chr.19: 37467094-37467094 on Build GRCh38
rs775064989 Chr.19: 37467396-37467396 on Build GRCh38
rs1046335742 Chr.19: 37467010-37467010 on Build GRCh38
rs529159436 Chr.19: 37467325-37467325 on Build GRCh38
rs916047664 Chr.19: 37467003-37467003 on Build GRCh38
rs3840939 Chr.19: 37467351-37467352 on Build GRCh38

View Full Product Details