Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38218225-38218765 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPF1
Entrez Gene ID
8193
Gene Name
double PHD fingers 1
Gene Aliases
BAF45b, NEUD4, neuro-d4
UniGene
Hs.631576
Gene Chromosome Location
Chr.19: 38211006-38229714 on Build GRCh38
Transcripts
NM_001135155.2 NM_001135156.2 NM_001289978.1 NM_004647.3 XM_005259289.2 XM_005259290.3 XM_005259292.4 XM_006723407.2 XM_006723408.3 XM_006723409.2 XM_006723410.2 XM_011527356.1 XM_011527357.2 XM_011527358.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5817567 c.467A>G p.E156G Chr.19: 38218622-38218622 on Build GRCh38
4428214 c.541G>C p.E181Q Chr.19: 38218629-38218629 on Build GRCh38
5817566 c.548A>G p.E183G Chr.19: 38218622-38218622 on Build GRCh38
4753169 c.536A>T p.D179V Chr.19: 38218634-38218634 on Build GRCh38
4077675 c.562G>A p.D188N Chr.19: 38218608-38218608 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs929349724 Chr.19: 38218277-38218277 on Build GRCh38
rs201255893 Chr.19: 38218630-38218630 on Build GRCh38
rs149830028 Chr.19: 38218449-38218449 on Build GRCh38
rs778823204 Chr.19: 38218698-38218698 on Build GRCh38
rs563545590 Chr.19: 38218408-38218408 on Build GRCh38
rs377405216 Chr.19: 38218545-38218545 on Build GRCh38
rs561010474 Chr.19: 38218541-38218541 on Build GRCh38
rs961170623 Chr.19: 38218488-38218488 on Build GRCh38
rs773162200 Chr.19: 38218702-38218702 on Build GRCh38
rs1011224302 Chr.19: 38218341-38218341 on Build GRCh38
rs776366953 Chr.19: 38218606-38218606 on Build GRCh38
rs926545404 Chr.19: 38218666-38218666 on Build GRCh38
rs201650538 Chr.19: 38218544-38218544 on Build GRCh38
rs780430565 Chr.19: 38218535-38218535 on Build GRCh38
rs10421241 Chr.19: 38218512-38218512 on Build GRCh38
rs374160204 Chr.19: 38218543-38218543 on Build GRCh38
rs375057978 Chr.19: 38218633-38218633 on Build GRCh38
rs568445989 Chr.19: 38218394-38218394 on Build GRCh38
rs936347631 Chr.19: 38218352-38218352 on Build GRCh38
rs370460157 Chr.19: 38218582-38218582 on Build GRCh38
rs755128250 Chr.19: 38218695-38218695 on Build GRCh38
rs775010543 Chr.19: 38218688-38218688 on Build GRCh38
rs1009295508 Chr.19: 38218438-38218438 on Build GRCh38
rs980595542 Chr.19: 38218650-38218650 on Build GRCh38
rs757077741 Chr.19: 38218322-38218322 on Build GRCh38
rs1028448372 Chr.19: 38218419-38218419 on Build GRCh38
rs948723709 Chr.19: 38218637-38218637 on Build GRCh38
rs565731054 Chr.19: 38218565-38218565 on Build GRCh38
rs771980344 Chr.19: 38218548-38218548 on Build GRCh38
rs992891373 Chr.19: 38218327-38218327 on Build GRCh38
rs780236041 Chr.19: 38218685-38218685 on Build GRCh38
rs917336577 Chr.19: 38218599-38218599 on Build GRCh38
rs375418730 Chr.19: 38218331-38218331 on Build GRCh38
rs866512855 Chr.19: 38218601-38218601 on Build GRCh38
rs769938876 Chr.19: 38218534-38218534 on Build GRCh38
rs1018277862 Chr.19: 38218491-38218491 on Build GRCh38
rs773073890 Chr.19: 38218573-38218573 on Build GRCh38
rs1053355721 Chr.19: 38218358-38218358 on Build GRCh38
rs2384780 Chr.19: 38218330-38218330 on Build GRCh38
rs764559263 Chr.19: 38218627-38218627 on Build GRCh38
rs750895914 Chr.19: 38218639-38218639 on Build GRCh38
rs749577825 Chr.19: 38218686-38218686 on Build GRCh38
rs747904896 Chr.19: 38218699-38218699 on Build GRCh38
rs374316921 Chr.19: 38218684-38218684 on Build GRCh38
rs746897267 Chr.19: 38218710-38218710 on Build GRCh38
rs892234047 Chr.19: 38218363-38218363 on Build GRCh38
rs986682557 Chr.19: 38218463-38218463 on Build GRCh38
rs200837988 Chr.19: 38218616-38218616 on Build GRCh38
rs527488188 Chr.19: 38218299-38218299 on Build GRCh38
rs945009175 Chr.19: 38218725-38218725 on Build GRCh38
rs750372963 Chr.19: 38218460-38218460 on Build GRCh38
rs1045936533 Chr.19: 38218682-38218682 on Build GRCh38
rs963630832 Chr.19: 38218504-38218504 on Build GRCh38
rs936568294 Chr.19: 38218674-38218674 on Build GRCh38
rs1045424982 Chr.19: 38218344-38218344 on Build GRCh38
rs571297388 Chr.19: 38218474-38218474 on Build GRCh38
rs764626423 Chr.19: 38218669-38218669 on Build GRCh38
rs905069809 Chr.19: 38218348-38218348 on Build GRCh38
rs375239664 Chr.19: 38218610-38218610 on Build GRCh38
rs771786140 Chr.19: 38218701-38218701 on Build GRCh38

View Full Product Details