Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 48231456-48231952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC5A9
Entrez Gene ID
200010
Gene Name
solute carrier family 5 member 9
Gene Aliases
SGLT4
UniGene
Hs.37890
Gene Chromosome Location
Chr.1: 48221113-48248644 on Build GRCh38
Transcripts
NM_001011547.2 NM_001135181.1 XM_011540924.2 XM_011540925.2 XM_011540926.2 XM_011540927.1 XM_011540928.1 XM_011540929.2 XM_017000558.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4754680 c.725C>T p.T242M Chr.1: 48231584-48231584 on Build GRCh38
7314847 c.692_696delTCATG p.L231fs*44 Chr.1: 48231551-48231555 on Build GRCh38
4008653 c.726G>A p.T242T Chr.1: 48231585-48231585 on Build GRCh38
4637721 c.688G>A p.G230S Chr.1: 48231547-48231547 on Build GRCh38
7238881 c.695T>C p.M232T Chr.1: 48231554-48231554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201595035 Chr.1: 48231606-48231606 on Build GRCh38
rs776516604 Chr.1: 48231509-48231509 on Build GRCh38
rs754326718 Chr.1: 48231661-48231661 on Build GRCh38
rs1038658287 Chr.1: 48231520-48231520 on Build GRCh38
rs973959793 Chr.1: 48231875-48231875 on Build GRCh38
rs921948183 Chr.1: 48231790-48231790 on Build GRCh38
rs760386213 Chr.1: 48231501-48231501 on Build GRCh38
rs932580229 Chr.1: 48231810-48231810 on Build GRCh38
rs577296499 Chr.1: 48231647-48231648 on Build GRCh38
rs868586444 Chr.1: 48231601-48231602 on Build GRCh38
rs973443534 Chr.1: 48231781-48231781 on Build GRCh38
rs1011563062 Chr.1: 48231705-48231705 on Build GRCh38
rs747660405 Chr.1: 48231574-48231574 on Build GRCh38
rs770636365 Chr.1: 48231600-48231600 on Build GRCh38
rs115427846 Chr.1: 48231729-48231729 on Build GRCh38
rs780114984 Chr.1: 48231928-48231928 on Build GRCh38
rs780749249 Chr.1: 48231572-48231572 on Build GRCh38
rs890631136 Chr.1: 48231627-48231627 on Build GRCh38
rs12047252 Chr.1: 48231554-48231554 on Build GRCh38
rs907963227 Chr.1: 48231753-48231753 on Build GRCh38
rs144495507 Chr.1: 48231517-48231517 on Build GRCh38
rs777133054 Chr.1: 48231925-48231928 on Build GRCh38
rs776268093 Chr.1: 48231604-48231604 on Build GRCh38
rs746598985 Chr.1: 48231592-48231592 on Build GRCh38
rs777095034 Chr.1: 48231901-48231901 on Build GRCh38
rs764095817 Chr.1: 48231625-48231625 on Build GRCh38
rs547903283 Chr.1: 48231782-48231782 on Build GRCh38
rs61730945 Chr.1: 48231605-48231605 on Build GRCh38
rs368190593 Chr.1: 48231507-48231507 on Build GRCh38
rs900778116 Chr.1: 48231702-48231702 on Build GRCh38
rs367794847 Chr.1: 48231617-48231617 on Build GRCh38
rs147266128 Chr.1: 48231584-48231584 on Build GRCh38
rs373152448 Chr.1: 48231890-48231890 on Build GRCh38
rs990691368 Chr.1: 48231779-48231779 on Build GRCh38
rs74077907 Chr.1: 48231870-48231870 on Build GRCh38
rs200959917 Chr.1: 48231898-48231898 on Build GRCh38
rs766983760 Chr.1: 48231653-48231653 on Build GRCh38
rs771820828 Chr.1: 48231495-48231495 on Build GRCh38
rs61730938 Chr.1: 48231585-48231585 on Build GRCh38
rs960840935 Chr.1: 48231757-48231757 on Build GRCh38
rs376514818 Chr.1: 48231631-48231631 on Build GRCh38
rs751675519 Chr.1: 48231548-48231548 on Build GRCh38
rs781372055 Chr.1: 48231558-48231558 on Build GRCh38
rs779817676 Chr.1: 48231837-48231842 on Build GRCh38
rs529558167 Chr.1: 48231746-48231746 on Build GRCh38
rs764286277 Chr.1: 48231534-48231534 on Build GRCh38
rs912359782 Chr.1: 48231845-48231845 on Build GRCh38
rs773068561 Chr.1: 48231500-48231500 on Build GRCh38
rs560935221 Chr.1: 48231712-48231712 on Build GRCh38
rs970323469 Chr.1: 48231695-48231695 on Build GRCh38
rs1049665476 Chr.1: 48231837-48231837 on Build GRCh38
rs910067442 Chr.1: 48231776-48231776 on Build GRCh38
rs756560321 Chr.1: 48231564-48231564 on Build GRCh38
rs141963205 Chr.1: 48231579-48231579 on Build GRCh38
rs756616323 Chr.1: 48231651-48231651 on Build GRCh38
rs144771107 Chr.1: 48231561-48231561 on Build GRCh38
rs747327959 Chr.1: 48231602-48231603 on Build GRCh38
rs202041931 Chr.1: 48231628-48231628 on Build GRCh38
rs756827507 Chr.1: 48231911-48231911 on Build GRCh38
rs1035858591 Chr.1: 48231703-48231703 on Build GRCh38
rs750500646 Chr.1: 48231724-48231724 on Build GRCh38
rs763085884 Chr.1: 48231532-48231532 on Build GRCh38
rs551776772 Chr.1: 48231881-48231881 on Build GRCh38
rs767833098 Chr.1: 48231636-48231636 on Build GRCh38
rs566215874 Chr.1: 48231849-48231849 on Build GRCh38
rs746575504 Chr.1: 48231902-48231902 on Build GRCh38
rs745634985 Chr.1: 48231925-48231925 on Build GRCh38
rs571691782 Chr.1: 48231559-48231559 on Build GRCh38
rs1034736788 Chr.1: 48231751-48231751 on Build GRCh38
rs762892060 Chr.1: 48231618-48231618 on Build GRCh38
rs1039713981 Chr.1: 48231648-48231648 on Build GRCh38
rs920598623 Chr.1: 48231878-48231878 on Build GRCh38
rs752742028 Chr.1: 48231522-48231522 on Build GRCh38
rs868702520 Chr.1: 48231688-48231688 on Build GRCh38

View Full Product Details