Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51823648-51824142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FPR3
Entrez Gene ID
2359
Gene Name
formyl peptide receptor 3
Gene Aliases
FML2_HUMAN, FMLP-R-II, FMLPY, FPRH1, FPRH2, FPRL2, RMLP-R-I
UniGene
Hs.445466
Gene Chromosome Location
Chr.19: 51795146-51826207 on Build GRCh38
Transcripts
NM_002030.4 XM_011526687.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5856535 c.160C>T p.R54W Chr.19: 51823908-51823908 on Build GRCh38
3537914 c.382C>T p.L128L Chr.19: 51824130-51824130 on Build GRCh38
1712706 c.236C>T p.P79L Chr.19: 51823984-51823984 on Build GRCh38
4472706 c.17C>T p.S6F Chr.19: 51823765-51823765 on Build GRCh38
5926024 c.101G>A p.G34E Chr.19: 51823849-51823849 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368542589 Chr.19: 51823794-51823794 on Build GRCh38
rs769430733 Chr.19: 51823983-51823983 on Build GRCh38
rs372060962 Chr.19: 51823943-51823943 on Build GRCh38
rs761856901 Chr.19: 51823977-51823977 on Build GRCh38
rs142506547 Chr.19: 51823887-51823887 on Build GRCh38
rs149153203 Chr.19: 51823847-51823847 on Build GRCh38
rs922564216 Chr.19: 51824100-51824100 on Build GRCh38
rs768724344 Chr.19: 51824044-51824044 on Build GRCh38
rs770401673 Chr.19: 51823778-51823778 on Build GRCh38
rs751196865 Chr.19: 51823998-51823998 on Build GRCh38
rs752886660 Chr.19: 51823835-51823835 on Build GRCh38
rs1013081896 Chr.19: 51823930-51823930 on Build GRCh38
rs932828527 Chr.19: 51823682-51823682 on Build GRCh38
rs780689685 Chr.19: 51824003-51824003 on Build GRCh38
rs61738412 Chr.19: 51823803-51823803 on Build GRCh38
rs143231858 Chr.19: 51823918-51823918 on Build GRCh38
rs753706002 Chr.19: 51823712-51823712 on Build GRCh38
rs267605625 Chr.19: 51823966-51823966 on Build GRCh38
rs764140414 Chr.19: 51823842-51823842 on Build GRCh38
rs186794108 Chr.19: 51823949-51823949 on Build GRCh38
rs1033841357 Chr.19: 51823855-51823855 on Build GRCh38
rs991733070 Chr.19: 51823693-51823693 on Build GRCh38
rs952881243 Chr.19: 51824008-51824008 on Build GRCh38
rs747654600 Chr.19: 51823694-51823694 on Build GRCh38
rs752241328 Chr.19: 51824101-51824101 on Build GRCh38
rs771642655 Chr.19: 51823728-51823728 on Build GRCh38
rs374854764 Chr.19: 51823800-51823800 on Build GRCh38
rs746571721 Chr.19: 51823880-51823880 on Build GRCh38
rs147419483 Chr.19: 51823767-51823767 on Build GRCh38
rs745546213 Chr.19: 51823725-51823725 on Build GRCh38
rs751161506 Chr.19: 51824079-51824079 on Build GRCh38
rs775409263 Chr.19: 51823912-51823912 on Build GRCh38
rs145113785 Chr.19: 51824066-51824066 on Build GRCh38
rs779630767 Chr.19: 51823731-51823731 on Build GRCh38
rs775740175 Chr.19: 51824036-51824036 on Build GRCh38
rs941355937 Chr.19: 51823723-51823723 on Build GRCh38
rs529548789 Chr.19: 51823718-51823718 on Build GRCh38
rs766108887 Chr.19: 51823995-51823995 on Build GRCh38
rs1050257388 Chr.19: 51823684-51823684 on Build GRCh38
rs753762728 Chr.19: 51823954-51823954 on Build GRCh38
rs746392669 Chr.19: 51824119-51824119 on Build GRCh38
rs770182384 Chr.19: 51823882-51823882 on Build GRCh38
rs774486058 Chr.19: 51823818-51823818 on Build GRCh38
rs779428648 Chr.19: 51823877-51823877 on Build GRCh38
rs755006976 Chr.19: 51823698-51823698 on Build GRCh38
rs772344801 Chr.19: 51824022-51824022 on Build GRCh38
rs777573101 Chr.19: 51823701-51823717 on Build GRCh38
rs1029947307 Chr.19: 51824115-51824115 on Build GRCh38
rs759748060 Chr.19: 51823908-51823908 on Build GRCh38
rs767007741 Chr.19: 51824087-51824087 on Build GRCh38
rs765308727 Chr.19: 51824065-51824065 on Build GRCh38
rs754439930 Chr.19: 51824001-51824001 on Build GRCh38
rs758185337 Chr.19: 51823866-51823866 on Build GRCh38
rs150944177 Chr.19: 51823899-51823899 on Build GRCh38
rs532790458 Chr.19: 51823840-51823840 on Build GRCh38
rs777424542 Chr.19: 51823710-51823710 on Build GRCh38
rs757959263 Chr.19: 51823990-51823990 on Build GRCh38
rs868395367 Chr.19: 51824052-51824052 on Build GRCh38
rs779342164 Chr.19: 51824116-51824116 on Build GRCh38
rs201759424 Chr.19: 51823848-51823848 on Build GRCh38
rs1035349596 Chr.19: 51823687-51823687 on Build GRCh38
rs1002074327 Chr.19: 51823845-51823845 on Build GRCh38
rs182683513 Chr.19: 51823917-51823917 on Build GRCh38
rs553351178 Chr.19: 51824085-51824085 on Build GRCh38
rs761840519 Chr.19: 51824062-51824062 on Build GRCh38
rs868227413 Chr.19: 51823689-51823689 on Build GRCh38
rs902930803 Chr.19: 51823891-51823891 on Build GRCh38
rs763361125 Chr.19: 51823796-51823796 on Build GRCh38
rs146700831 Chr.19: 51823989-51823989 on Build GRCh38
rs559612922 Chr.19: 51823829-51823829 on Build GRCh38
rs766122928 Chr.19: 51824068-51824068 on Build GRCh38
rs541289763 Chr.19: 51823737-51823737 on Build GRCh38
rs756132271 Chr.19: 51823707-51823707 on Build GRCh38
rs762763346 Chr.19: 51824067-51824067 on Build GRCh38
rs140768137 Chr.19: 51823909-51823909 on Build GRCh38
rs778856602 Chr.19: 51823862-51823862 on Build GRCh38
rs779215430 Chr.19: 51824009-51824009 on Build GRCh38
rs767479545 Chr.19: 51823831-51823831 on Build GRCh38
rs976730222 Chr.19: 51823772-51823772 on Build GRCh38
rs879188379 Chr.19: 51823741-51823741 on Build GRCh38
rs201037671 Chr.19: 51824054-51824054 on Build GRCh38
rs143250626 Chr.19: 51824004-51824004 on Build GRCh38
rs375494728 Chr.19: 51823720-51823720 on Build GRCh38
rs993344600 Chr.19: 51823817-51823817 on Build GRCh38
rs752516033 Chr.19: 51823705-51823705 on Build GRCh38
rs767584609 Chr.19: 51823699-51823699 on Build GRCh38
rs778298008 Chr.19: 51823884-51823884 on Build GRCh38
rs771224132 Chr.19: 51823890-51823890 on Build GRCh38
rs746923030 Chr.19: 51823939-51823940 on Build GRCh38
rs746279537 Chr.19: 51824013-51824013 on Build GRCh38
rs369595664 Chr.19: 51823865-51823865 on Build GRCh38
rs141324188 Chr.19: 51824039-51824039 on Build GRCh38

View Full Product Details