Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 52405828-52406341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF528
Entrez Gene ID
84436
Gene Name
zinc finger protein 528
Gene Aliases
-
UniGene
Hs.662043
Gene Chromosome Location
Chr.19: 52392771-52418408 on Build GRCh38
Transcripts
NM_032423.2 XM_005259323.1 XM_006723418.2 XM_011527392.1 XM_017027360.1 XM_017027361.1 XM_017027362.1 XM_017027363.1 XM_017027364.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4856448 c.80C>G p.P27R Chr.19: 52405971-52405971 on Build GRCh38
6995068 c.139C>A p.L47M Chr.19: 52406030-52406030 on Build GRCh38
3371460 c.34G>T p.D12Y Chr.19: 52405925-52405925 on Build GRCh38
4775388 c.112T>A p.L38M Chr.19: 52406003-52406003 on Build GRCh38
3389300 c.47A>T p.E16V Chr.19: 52405938-52405938 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771082741 Chr.19: 52405980-52405980 on Build GRCh38
rs545805854 Chr.19: 52406314-52406314 on Build GRCh38
rs373691668 Chr.19: 52405975-52405975 on Build GRCh38
rs2245620 Chr.19: 52406205-52406205 on Build GRCh38
rs374660359 Chr.19: 52405867-52405867 on Build GRCh38
rs551703205 Chr.19: 52406040-52406040 on Build GRCh38
rs983870911 Chr.19: 52406301-52406301 on Build GRCh38
rs370656674 Chr.19: 52405947-52405947 on Build GRCh38
rs377651156 Chr.19: 52405878-52405878 on Build GRCh38
rs562711165 Chr.19: 52406176-52406176 on Build GRCh38
rs974320673 Chr.19: 52405909-52405909 on Build GRCh38
rs549979976 Chr.19: 52406104-52406104 on Build GRCh38
rs143434257 Chr.19: 52406072-52406072 on Build GRCh38
rs746831899 Chr.19: 52406060-52406060 on Build GRCh38
rs141013163 Chr.19: 52405974-52405974 on Build GRCh38
rs375475150 Chr.19: 52406249-52406249 on Build GRCh38
rs982587373 Chr.19: 52406306-52406306 on Build GRCh38
rs749566600 Chr.19: 52406035-52406035 on Build GRCh38
rs761042020 Chr.19: 52406012-52406012 on Build GRCh38
rs368856577 Chr.19: 52406059-52406059 on Build GRCh38
rs778159726 Chr.19: 52406026-52406026 on Build GRCh38
rs907928425 Chr.19: 52406309-52406309 on Build GRCh38
rs774464653 Chr.19: 52405981-52405981 on Build GRCh38
rs200590539 Chr.19: 52406036-52406036 on Build GRCh38
rs113027615 Chr.19: 52406125-52406125 on Build GRCh38
rs768490189 Chr.19: 52406062-52406062 on Build GRCh38
rs568296092 Chr.19: 52406122-52406122 on Build GRCh38
rs1036797161 Chr.19: 52405876-52405876 on Build GRCh38
rs182066556 Chr.19: 52406183-52406183 on Build GRCh38
rs536020300 Chr.19: 52406256-52406256 on Build GRCh38
rs377324364 Chr.19: 52405880-52405880 on Build GRCh38
rs780562677 Chr.19: 52405905-52405905 on Build GRCh38
rs531677584 Chr.19: 52406073-52406073 on Build GRCh38
rs939962086 Chr.19: 52406118-52406118 on Build GRCh38
rs998043248 Chr.19: 52405912-52405912 on Build GRCh38
rs2290744 Chr.19: 52405996-52405996 on Build GRCh38
rs370548074 Chr.19: 52406002-52406002 on Build GRCh38
rs758482288 Chr.19: 52406257-52406257 on Build GRCh38
rs753443394 Chr.19: 52405997-52405997 on Build GRCh38
rs759395532 Chr.19: 52405860-52405860 on Build GRCh38
rs370994436 Chr.19: 52406043-52406043 on Build GRCh38
rs2290746 Chr.19: 52406228-52406228 on Build GRCh38
rs2290747 Chr.19: 52406254-52406254 on Build GRCh38
rs1023039030 Chr.19: 52406274-52406274 on Build GRCh38
rs915638246 Chr.19: 52406038-52406038 on Build GRCh38
rs763066728 Chr.19: 52405856-52405856 on Build GRCh38
rs553541314 Chr.19: 52406284-52406284 on Build GRCh38
rs376391054 Chr.19: 52405978-52405978 on Build GRCh38
rs762380642 Chr.19: 52405910-52405910 on Build GRCh38
rs2290745 Chr.19: 52406050-52406050 on Build GRCh38
rs752057936 Chr.19: 52405933-52405933 on Build GRCh38
rs764931627 Chr.19: 52406078-52406078 on Build GRCh38
rs1056873226 Chr.19: 52406192-52406192 on Build GRCh38
rs756505790 Chr.19: 52406024-52406024 on Build GRCh38
rs137874809 Chr.19: 52405987-52405987 on Build GRCh38
rs974056713 Chr.19: 52406099-52406099 on Build GRCh38
rs189459884 Chr.19: 52406174-52406174 on Build GRCh38
rs556326049 Chr.19: 52406252-52406252 on Build GRCh38
rs753116369 Chr.19: 52406021-52406021 on Build GRCh38
rs776380510 Chr.19: 52406069-52406069 on Build GRCh38
rs1030777920 Chr.19: 52406049-52406049 on Build GRCh38
rs964307487 Chr.19: 52406092-52406092 on Build GRCh38
rs889171283 Chr.19: 52406202-52406202 on Build GRCh38
rs759428575 Chr.19: 52405982-52405982 on Build GRCh38
rs2608512 Chr.19: 52405857-52405857 on Build GRCh38
rs146475361 Chr.19: 52405971-52405971 on Build GRCh38
rs572117514 Chr.19: 52406289-52406289 on Build GRCh38
rs903353137 Chr.19: 52406248-52406248 on Build GRCh38
rs775165507 Chr.19: 52405993-52405993 on Build GRCh38
rs752502222 Chr.19: 52405873-52405873 on Build GRCh38
rs756719785 Chr.19: 52405892-52405892 on Build GRCh38
rs760381869 Chr.19: 52405877-52405877 on Build GRCh38
rs886997205 Chr.19: 52405862-52405862 on Build GRCh38
rs933297468 Chr.19: 52406142-52406142 on Build GRCh38
rs781681476 Chr.19: 52406020-52406020 on Build GRCh38
rs556032800 Chr.19: 52405963-52405963 on Build GRCh38
rs760400169 Chr.19: 52405994-52405994 on Build GRCh38
rs371918549 Chr.19: 52406259-52406259 on Build GRCh38
rs373661269 Chr.19: 52406014-52406014 on Build GRCh38
rs895396840 Chr.19: 52406015-52406015 on Build GRCh38
rs999261103 Chr.19: 52406048-52406048 on Build GRCh38

View Full Product Details