Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54354960-54355455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LAIR1
Entrez Gene ID
3903
Gene Name
leukocyte associated immunoglobulin like receptor 1
Gene Aliases
CD305, LAIR-1
UniGene
Hs.572535
Gene Chromosome Location
Chr.19: 54353624-54370556 on Build GRCh38
Transcripts
NM_001289023.2 NM_001289025.2 NM_001289026.2 NM_001289027.2 NM_002287.5 NM_021706.4 NR_110279.2 NR_110280.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6698984 c.776T>C p.L259P Chr.19: 54355356-54355356 on Build GRCh38
4697186 c.722T>C p.L241P Chr.19: 54355410-54355410 on Build GRCh38
1172660 c.741G>C p.Q247H Chr.19: 54355391-54355391 on Build GRCh38
6698983 c.850G>A p.V284I Chr.19: 54355282-54355282 on Build GRCh38
282341 c.831G>T p.E277D Chr.19: 54355301-54355301 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200353736 Chr.19: 54355277-54355277 on Build GRCh38
rs749608668 Chr.19: 54355286-54355296 on Build GRCh38
rs954350465 Chr.19: 54355042-54355042 on Build GRCh38
rs769013974 Chr.19: 54355410-54355411 on Build GRCh38
rs554172383 Chr.19: 54355304-54355304 on Build GRCh38
rs760167614 Chr.19: 54355309-54355309 on Build GRCh38
rs929548414 Chr.19: 54355182-54355182 on Build GRCh38
rs749650618 Chr.19: 54355382-54355382 on Build GRCh38
rs760303811 Chr.19: 54355250-54355250 on Build GRCh38
rs374608831 Chr.19: 54355245-54355245 on Build GRCh38
rs928143203 Chr.19: 54355064-54355064 on Build GRCh38
rs938393917 Chr.19: 54355065-54355065 on Build GRCh38
rs752521470 Chr.19: 54355332-54355332 on Build GRCh38
rs945647718 Chr.19: 54355026-54355026 on Build GRCh38
rs568061484 Chr.19: 54355283-54355283 on Build GRCh38
rs765167924 Chr.19: 54355319-54355319 on Build GRCh38
rs559968360 Chr.19: 54355091-54355091 on Build GRCh38
rs775426251 Chr.19: 54355413-54355413 on Build GRCh38
rs376569529 Chr.19: 54355377-54355377 on Build GRCh38
rs546662558 Chr.19: 54355067-54355067 on Build GRCh38
rs754763759 Chr.19: 54355260-54355260 on Build GRCh38
rs146736409 Chr.19: 54355398-54355398 on Build GRCh38
rs186759639 Chr.19: 54355219-54355219 on Build GRCh38
rs766052042 Chr.19: 54355257-54355257 on Build GRCh38
rs888334531 Chr.19: 54355127-54355127 on Build GRCh38
rs776252557 Chr.19: 54355404-54355404 on Build GRCh38
rs888124179 Chr.19: 54355336-54355336 on Build GRCh38
rs571716940 Chr.19: 54355421-54355421 on Build GRCh38
rs759439899 Chr.19: 54355317-54355317 on Build GRCh38
rs759315445 Chr.19: 54355405-54355405 on Build GRCh38
rs948414195 Chr.19: 54355109-54355109 on Build GRCh38
rs757423062 Chr.19: 54355282-54355282 on Build GRCh38
rs532201686 Chr.19: 54355092-54355092 on Build GRCh38
rs368044931 Chr.19: 54355299-54355299 on Build GRCh38
rs61742614 Chr.19: 54355383-54355383 on Build GRCh38
rs770294341 Chr.19: 54355228-54355228 on Build GRCh38
rs777767149 Chr.19: 54355276-54355276 on Build GRCh38
rs144745396 Chr.19: 54354986-54354986 on Build GRCh38
rs998743529 Chr.19: 54355322-54355322 on Build GRCh38
rs545341120 Chr.19: 54355125-54355125 on Build GRCh38
rs1031307101 Chr.19: 54355025-54355025 on Build GRCh38
rs755352799 Chr.19: 54355263-54355263 on Build GRCh38
rs762746926 Chr.19: 54355416-54355416 on Build GRCh38
rs775923414 Chr.19: 54355232-54355232 on Build GRCh38
rs964937847 Chr.19: 54355095-54355095 on Build GRCh38
rs1027830644 Chr.19: 54355435-54355435 on Build GRCh38
rs973974328 Chr.19: 54355102-54355102 on Build GRCh38
rs763958281 Chr.19: 54355427-54355427 on Build GRCh38
rs776374278 Chr.19: 54355316-54355316 on Build GRCh38
rs372322548 Chr.19: 54355312-54355312 on Build GRCh38
rs920888066 Chr.19: 54355124-54355124 on Build GRCh38
rs529702973 Chr.19: 54355021-54355021 on Build GRCh38
rs901043908 Chr.19: 54355172-54355172 on Build GRCh38
rs55799769 Chr.19: 54355259-54355259 on Build GRCh38
rs756307025 Chr.19: 54355354-54355354 on Build GRCh38
rs987256924 Chr.19: 54355053-54355053 on Build GRCh38
rs200961101 Chr.19: 54355303-54355303 on Build GRCh38
rs769560138 Chr.19: 54355407-54355407 on Build GRCh38
rs1004142441 Chr.19: 54355133-54355133 on Build GRCh38
rs542269242 Chr.19: 54355221-54355221 on Build GRCh38
rs552011678 Chr.19: 54355110-54355110 on Build GRCh38
rs746161447 Chr.19: 54355227-54355227 on Build GRCh38
rs746249343 Chr.19: 54355292-54355292 on Build GRCh38
rs1015398241 Chr.19: 54355143-54355143 on Build GRCh38
rs772755587 Chr.19: 54355248-54355248 on Build GRCh38
rs748673200 Chr.19: 54355349-54355349 on Build GRCh38
rs1042502766 Chr.19: 54355055-54355055 on Build GRCh38
rs192330346 Chr.19: 54355265-54355265 on Build GRCh38
rs141309380 Chr.19: 54355351-54355351 on Build GRCh38
rs539893856 Chr.19: 54355338-54355338 on Build GRCh38
rs146566429 Chr.19: 54355267-54355267 on Build GRCh38
rs761342733 Chr.19: 54355241-54355241 on Build GRCh38
rs983667885 Chr.19: 54355197-54355197 on Build GRCh38
rs757309719 Chr.19: 54355340-54355340 on Build GRCh38
rs780452555 Chr.19: 54355378-54355378 on Build GRCh38
rs1048503323 Chr.19: 54355328-54355328 on Build GRCh38
rs751471800 Chr.19: 54355431-54355431 on Build GRCh38
rs750683919 Chr.19: 54355352-54355352 on Build GRCh38
rs200218727 Chr.19: 54355339-54355339 on Build GRCh38
rs770103913 Chr.19: 54355293-54355293 on Build GRCh38
rs1008969074 Chr.19: 54355379-54355379 on Build GRCh38

View Full Product Details